Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159192338-159192858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADM3
Entrez Gene ID
57863
Gene Name
cell adhesion molecule 3
Gene Aliases
BIgR, IGSF4B, NECL1, Necl-1, TSLL1, synCAM3
UniGene
Hs.365689
Gene Chromosome Location
Chr.1: 159171587-159203142 on Build GRCh38
Transcripts
NM_001127173.1 NM_021189.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6850003 c.384C>A p.S128R Chr.1: 159192630-159192630 on Build GRCh38
3934172 c.396C>T p.S132S Chr.1: 159192642-159192642 on Build GRCh38
1688884 c.337C>T p.R113* Chr.1: 159192583-159192583 on Build GRCh38
3376791 c.418G>A p.G140S Chr.1: 159192664-159192664 on Build GRCh38
3399869 c.346C>T p.R116* Chr.1: 159192592-159192592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776693432 Chr.1: 159192540-159192540 on Build GRCh38
rs755756300 Chr.1: 159192609-159192609 on Build GRCh38
rs990517248 Chr.1: 159192393-159192393 on Build GRCh38
rs765272968 Chr.1: 159192651-159192651 on Build GRCh38
rs983781182 Chr.1: 159192700-159192700 on Build GRCh38
rs369946971 Chr.1: 159192718-159192718 on Build GRCh38
rs952612749 Chr.1: 159192696-159192696 on Build GRCh38
rs753491750 Chr.1: 159192723-159192723 on Build GRCh38
rs867156364 Chr.1: 159192549-159192549 on Build GRCh38
rs372345467 Chr.1: 159192701-159192701 on Build GRCh38
rs992950448 Chr.1: 159192726-159192726 on Build GRCh38
rs375999159 Chr.1: 159192619-159192619 on Build GRCh38
rs926440497 Chr.1: 159192509-159192509 on Build GRCh38
rs373357445 Chr.1: 159192778-159192778 on Build GRCh38
rs537290057 Chr.1: 159192438-159192438 on Build GRCh38
rs777454778 Chr.1: 159192612-159192612 on Build GRCh38
rs759032980 Chr.1: 159192418-159192418 on Build GRCh38
rs769071503 Chr.1: 159192748-159192748 on Build GRCh38
rs888060211 Chr.1: 159192622-159192622 on Build GRCh38
rs777176359 Chr.1: 159192629-159192629 on Build GRCh38
rs4329510 Chr.1: 159192605-159192605 on Build GRCh38
rs186510837 Chr.1: 159192481-159192481 on Build GRCh38
rs967995441 Chr.1: 159192816-159192816 on Build GRCh38
rs775746546 Chr.1: 159192618-159192618 on Build GRCh38
rs568923586 Chr.1: 159192422-159192422 on Build GRCh38
rs752726122 Chr.1: 159192705-159192705 on Build GRCh38
rs371824931 Chr.1: 159192599-159192599 on Build GRCh38
rs868159331 Chr.1: 159192740-159192740 on Build GRCh38
rs193920866 Chr.1: 159192730-159192730 on Build GRCh38
rs1032328444 Chr.1: 159192488-159192488 on Build GRCh38
rs762258608 Chr.1: 159192741-159192742 on Build GRCh38
rs766135173 Chr.1: 159192573-159192573 on Build GRCh38
rs750677129 Chr.1: 159192565-159192565 on Build GRCh38
rs375465939 Chr.1: 159192704-159192704 on Build GRCh38
rs780792191 Chr.1: 159192588-159192588 on Build GRCh38
rs892565338 Chr.1: 159192513-159192513 on Build GRCh38
rs141333594 Chr.1: 159192443-159192443 on Build GRCh38
rs747358243 Chr.1: 159192746-159192746 on Build GRCh38
rs777170455 Chr.1: 159192572-159192572 on Build GRCh38
rs896625322 Chr.1: 159192375-159192375 on Build GRCh38
rs2814767 Chr.1: 159192384-159192384 on Build GRCh38
rs780404957 Chr.1: 159192744-159192744 on Build GRCh38
rs966175397 Chr.1: 159192672-159192672 on Build GRCh38
rs751476207 Chr.1: 159192578-159192578 on Build GRCh38
rs956074562 Chr.1: 159192388-159192388 on Build GRCh38
rs751327260 Chr.1: 159192690-159192690 on Build GRCh38
rs754971531 Chr.1: 159192579-159192579 on Build GRCh38
rs761961875 Chr.1: 159192550-159192550 on Build GRCh38
rs200275600 Chr.1: 159192682-159192682 on Build GRCh38
rs35133972 Chr.1: 159192456-159192456 on Build GRCh38
rs918102000 Chr.1: 159192729-159192729 on Build GRCh38
rs975265753 Chr.1: 159192663-159192663 on Build GRCh38
rs748945624 Chr.1: 159192614-159192614 on Build GRCh38
rs146412590 Chr.1: 159192615-159192615 on Build GRCh38
rs764116647 Chr.1: 159192531-159192531 on Build GRCh38
rs765427974 Chr.1: 159192564-159192564 on Build GRCh38
rs554543604 Chr.1: 159192758-159192758 on Build GRCh38
rs182428055 Chr.1: 159192572-159192572 on Build GRCh38
rs761876244 Chr.1: 159192634-159192634 on Build GRCh38
rs557217087 Chr.1: 159192660-159192660 on Build GRCh38
rs949620314 Chr.1: 159192741-159192741 on Build GRCh38
rs369601885 Chr.1: 159192667-159192667 on Build GRCh38
rs769161367 Chr.1: 159192621-159192621 on Build GRCh38
rs187132036 Chr.1: 159192601-159192601 on Build GRCh38
rs189768245 Chr.1: 159192498-159192498 on Build GRCh38
rs748702027 Chr.1: 159192762-159192762 on Build GRCh38
rs755666560 Chr.1: 159192714-159192714 on Build GRCh38

View Full Product Details