Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 108496307-108496837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNPLA8
Entrez Gene ID
50640
Gene Name
patatin like phospholipase domain containing 8
Gene Aliases
IPLA2-2, IPLA2G, MMLA, PNPLA-gamma, iPLA2gamma
UniGene
Hs.617340
Gene Chromosome Location
Chr.7: 108470417-108528161 on Build GRCh38
Transcripts
NM_001256007.2 NM_001256008.2 NM_001256009.2 NM_001256010.2 NM_001256011.2 NM_015723.4 XM_005250396.4 XM_011516274.2 XM_011516275.2 XM_017012261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4716282 c.1457C>T p.A486V Chr.7: 108496752-108496752 on Build GRCh38
6349859 c.1454-6T>C p.? Chr.7: 108496761-108496761 on Build GRCh38
1447245 c.1519C>T p.R507* Chr.7: 108496690-108496690 on Build GRCh38
6315303 c.1492C>A p.P498T Chr.7: 108496717-108496717 on Build GRCh38
1447244 c.1520G>A p.R507Q Chr.7: 108496689-108496689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764595710 Chr.7: 108496736-108496736 on Build GRCh38
rs929693359 Chr.7: 108496345-108496345 on Build GRCh38
rs886583880 Chr.7: 108496346-108496346 on Build GRCh38
rs913750346 Chr.7: 108496397-108496397 on Build GRCh38
rs746978363 Chr.7: 108496662-108496662 on Build GRCh38
rs764931626 Chr.7: 108496766-108496766 on Build GRCh38
rs758248611 Chr.7: 108496655-108496655 on Build GRCh38
rs943257637 Chr.7: 108496431-108496431 on Build GRCh38
rs1001860426 Chr.7: 108496377-108496377 on Build GRCh38
rs753107099 Chr.7: 108496543-108496543 on Build GRCh38
rs997974156 Chr.7: 108496331-108496331 on Build GRCh38
rs958071797 Chr.7: 108496504-108496504 on Build GRCh38
rs1006479318 Chr.7: 108496386-108496386 on Build GRCh38
rs79896302 Chr.7: 108496499-108496499 on Build GRCh38
rs777654236 Chr.7: 108496659-108496659 on Build GRCh38
rs552298126 Chr.7: 108496467-108496467 on Build GRCh38
rs747971764 Chr.7: 108496654-108496654 on Build GRCh38
rs35773218 Chr.7: 108496594-108496594 on Build GRCh38
rs925227622 Chr.7: 108496421-108496421 on Build GRCh38
rs777566130 Chr.7: 108496796-108496796 on Build GRCh38
rs757191532 Chr.7: 108496801-108496801 on Build GRCh38
rs923227600 Chr.7: 108496485-108496485 on Build GRCh38
rs775630045 Chr.7: 108496719-108496719 on Build GRCh38
rs554676664 Chr.7: 108496573-108496573 on Build GRCh38
rs72570598 Chr.7: 108496808-108496809 on Build GRCh38
rs11448779 Chr.7: 108496809-108496810 on Build GRCh38
rs754513143 Chr.7: 108496765-108496765 on Build GRCh38
rs1026484631 Chr.7: 108496337-108496338 on Build GRCh38
rs970282078 Chr.7: 108496372-108496372 on Build GRCh38
rs572980597 Chr.7: 108496477-108496477 on Build GRCh38
rs895180127 Chr.7: 108496399-108496399 on Build GRCh38
rs1033274321 Chr.7: 108496385-108496385 on Build GRCh38
rs925072131 Chr.7: 108496720-108496720 on Build GRCh38
rs74677391 Chr.7: 108496349-108496349 on Build GRCh38
rs372867841 Chr.7: 108496711-108496711 on Build GRCh38
rs774129321 Chr.7: 108496569-108496574 on Build GRCh38
rs369590150 Chr.7: 108496705-108496705 on Build GRCh38
rs1046444209 Chr.7: 108496810-108496810 on Build GRCh38
rs761621799 Chr.7: 108496578-108496581 on Build GRCh38
rs79415311 Chr.7: 108496389-108496389 on Build GRCh38
rs906493221 Chr.7: 108496811-108496811 on Build GRCh38
rs375293468 Chr.7: 108496653-108496653 on Build GRCh38
rs763291396 Chr.7: 108496546-108496546 on Build GRCh38
rs992295923 Chr.7: 108496390-108496390 on Build GRCh38
rs900912943 Chr.7: 108496332-108496332 on Build GRCh38
rs879185889 Chr.7: 108496652-108496654 on Build GRCh38
rs776793393 Chr.7: 108496539-108496539 on Build GRCh38
rs34792119 Chr.7: 108496729-108496730 on Build GRCh38
rs758612557 Chr.7: 108496411-108496411 on Build GRCh38
rs929751854 Chr.7: 108496619-108496619 on Build GRCh38
rs763351460 Chr.7: 108496730-108496730 on Build GRCh38
rs372287503 Chr.7: 108496576-108496576 on Build GRCh38
rs1016495366 Chr.7: 108496363-108496363 on Build GRCh38
rs886574385 Chr.7: 108496380-108496380 on Build GRCh38
rs747060333 Chr.7: 108496542-108496542 on Build GRCh38
rs537267381 Chr.7: 108496799-108496799 on Build GRCh38
rs770781117 Chr.7: 108496606-108496606 on Build GRCh38
rs754672564 Chr.7: 108496628-108496628 on Build GRCh38
rs774961886 Chr.7: 108496738-108496738 on Build GRCh38
rs375676360 Chr.7: 108496672-108496672 on Build GRCh38
rs759826233 Chr.7: 108496541-108496541 on Build GRCh38
rs764681645 Chr.7: 108496548-108496548 on Build GRCh38
rs758148091 Chr.7: 108496789-108496789 on Build GRCh38
rs886651935 Chr.7: 108496735-108496735 on Build GRCh38
rs376656732 Chr.7: 108496750-108496750 on Build GRCh38
rs76741982 Chr.7: 108496424-108496424 on Build GRCh38
rs186772108 Chr.7: 108496406-108496406 on Build GRCh38
rs989395466 Chr.7: 108496531-108496531 on Build GRCh38
rs752344118 Chr.7: 108496768-108496768 on Build GRCh38
rs777742719 Chr.7: 108496489-108496489 on Build GRCh38
rs746006408 Chr.7: 108496710-108496710 on Build GRCh38
rs1012479281 Chr.7: 108496793-108496793 on Build GRCh38
rs190830973 Chr.7: 108496535-108496535 on Build GRCh38
rs781176391 Chr.7: 108496737-108496737 on Build GRCh38
rs970853330 Chr.7: 108496478-108496478 on Build GRCh38
rs760775644 Chr.7: 108496810-108496811 on Build GRCh38
rs974633836 Chr.7: 108496809-108496809 on Build GRCh38
rs1039059984 Chr.7: 108496470-108496470 on Build GRCh38
rs753386955 Chr.7: 108496599-108496599 on Build GRCh38
rs770948284 Chr.7: 108496689-108496689 on Build GRCh38

View Full Product Details