Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 120951024-120951548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ING3
Entrez Gene ID
54556
Gene Name
inhibitor of growth family member 3
Gene Aliases
Eaf4, ING2, MEAF4, p47ING3
UniGene
Hs.489811
Gene Chromosome Location
Chr.7: 120950755-120975657 on Build GRCh38
Transcripts
NM_019071.2 NM_198267.1 XM_017012369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5033190 c.84G>A p.M28I Chr.7: 120951219-120951219 on Build GRCh38
1254834 c.96G>T p.V32V Chr.7: 120951231-120951231 on Build GRCh38
292021 c.72A>T p.E24D Chr.7: 120951207-120951207 on Build GRCh38
3257617 c.70G>T p.E24* Chr.7: 120951205-120951205 on Build GRCh38
4422880 c.34G>C p.E12Q Chr.7: 120951169-120951169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754081978 Chr.7: 120951256-120951256 on Build GRCh38
rs11548053 Chr.7: 120951192-120951192 on Build GRCh38
rs765005996 Chr.7: 120951146-120951146 on Build GRCh38
rs201878594 Chr.7: 120951137-120951137 on Build GRCh38
rs778984071 Chr.7: 120951135-120951136 on Build GRCh38
rs188626385 Chr.7: 120951066-120951066 on Build GRCh38
rs750273807 Chr.7: 120951251-120951251 on Build GRCh38
rs553563387 Chr.7: 120951498-120951498 on Build GRCh38
rs747705476 Chr.7: 120951088-120951088 on Build GRCh38
rs914955246 Chr.7: 120951370-120951370 on Build GRCh38
rs536782017 Chr.7: 120951375-120951375 on Build GRCh38
rs114383422 Chr.7: 120951195-120951195 on Build GRCh38
rs376829981 Chr.7: 120951125-120951125 on Build GRCh38
rs769127768 Chr.7: 120951220-120951220 on Build GRCh38
rs932425797 Chr.7: 120951352-120951352 on Build GRCh38
rs772673743 Chr.7: 120951203-120951203 on Build GRCh38
rs369134261 Chr.7: 120951301-120951301 on Build GRCh38
rs770097111 Chr.7: 120951122-120951122 on Build GRCh38
rs377106574 Chr.7: 120951245-120951245 on Build GRCh38
rs769040781 Chr.7: 120951185-120951185 on Build GRCh38
rs559672718 Chr.7: 120951409-120951409 on Build GRCh38
rs779077907 Chr.7: 120951265-120951265 on Build GRCh38
rs370568348 Chr.7: 120951072-120951073 on Build GRCh38
rs540289134 Chr.7: 120951397-120951397 on Build GRCh38
rs749105998 Chr.7: 120951393-120951393 on Build GRCh38
rs758676507 Chr.7: 120951274-120951274 on Build GRCh38
rs758937516 Chr.7: 120951140-120951140 on Build GRCh38
rs762651592 Chr.7: 120951228-120951228 on Build GRCh38
rs752278803 Chr.7: 120951147-120951147 on Build GRCh38
rs897864187 Chr.7: 120951100-120951100 on Build GRCh38
rs762268894 Chr.7: 120951136-120951136 on Build GRCh38
rs897415536 Chr.7: 120951081-120951081 on Build GRCh38
rs113378963 Chr.7: 120951247-120951247 on Build GRCh38
rs571215475 Chr.7: 120951332-120951332 on Build GRCh38
rs925199145 Chr.7: 120951069-120951069 on Build GRCh38
rs994145798 Chr.7: 120951102-120951102 on Build GRCh38
rs997692342 Chr.7: 120951391-120951391 on Build GRCh38
rs929302053 Chr.7: 120951318-120951318 on Build GRCh38
rs749714257 Chr.7: 120951126-120951126 on Build GRCh38
rs762850940 Chr.7: 120951148-120951148 on Build GRCh38
rs553727631 Chr.7: 120951088-120951088 on Build GRCh38
rs757210404 Chr.7: 120951153-120951153 on Build GRCh38
rs958753859 Chr.7: 120951103-120951103 on Build GRCh38
rs764000713 Chr.7: 120951242-120951242 on Build GRCh38
rs567223826 Chr.7: 120951427-120951427 on Build GRCh38
rs773960425 Chr.7: 120951504-120951504 on Build GRCh38
rs574163629 Chr.7: 120951083-120951083 on Build GRCh38
rs748577257 Chr.7: 120951267-120951267 on Build GRCh38
rs780341286 Chr.7: 120951183-120951183 on Build GRCh38
rs371143231 Chr.7: 120951285-120951285 on Build GRCh38
rs996525599 Chr.7: 120951065-120951065 on Build GRCh38
rs921084044 Chr.7: 120951333-120951333 on Build GRCh38
rs965647576 Chr.7: 120951314-120951315 on Build GRCh38
rs903456307 Chr.7: 120951418-120951418 on Build GRCh38
rs748425080 Chr.7: 120951256-120951258 on Build GRCh38
rs1011819494 Chr.7: 120951081-120951082 on Build GRCh38
rs750586764 Chr.7: 120951171-120951171 on Build GRCh38
rs902048121 Chr.7: 120951382-120951382 on Build GRCh38
rs908466076 Chr.7: 120951313-120951313 on Build GRCh38
rs367660853 Chr.7: 120951252-120951252 on Build GRCh38
rs571017006 Chr.7: 120951057-120951057 on Build GRCh38
rs888024330 Chr.7: 120951350-120951350 on Build GRCh38
rs75104085 Chr.7: 120951408-120951408 on Build GRCh38
rs1045688417 Chr.7: 120951099-120951099 on Build GRCh38
rs982716258 Chr.7: 120951129-120951129 on Build GRCh38
rs537100843 Chr.7: 120951059-120951059 on Build GRCh38
rs373986234 Chr.7: 120951151-120951151 on Build GRCh38
rs763910956 Chr.7: 120951149-120951149 on Build GRCh38
rs768955019 Chr.7: 120951127-120951127 on Build GRCh38
rs745844011 Chr.7: 120951517-120951517 on Build GRCh38
rs1035797518 Chr.7: 120951264-120951264 on Build GRCh38
rs775282901 Chr.7: 120951225-120951225 on Build GRCh38
rs1039219046 Chr.7: 120951380-120951380 on Build GRCh38
rs141835756 Chr.7: 120951086-120951086 on Build GRCh38
rs528328263 Chr.7: 120951306-120951306 on Build GRCh38
rs756138808 Chr.7: 120951181-120951181 on Build GRCh38
rs573544219 Chr.7: 120951094-120951094 on Build GRCh38
rs545429239 Chr.7: 120951131-120951131 on Build GRCh38
rs780962589 Chr.7: 120951166-120951166 on Build GRCh38
rs983155034 Chr.7: 120951310-120951310 on Build GRCh38
rs201054245 Chr.7: 120951208-120951208 on Build GRCh38
rs572964406 Chr.7: 120951322-120951322 on Build GRCh38
rs1032450392 Chr.7: 120951459-120951459 on Build GRCh38
rs113365551 Chr.7: 120951447-120951447 on Build GRCh38
rs967027106 Chr.7: 120951062-120951062 on Build GRCh38
rs989228334 Chr.7: 120951366-120951366 on Build GRCh38
rs938915946 Chr.7: 120951453-120951453 on Build GRCh38
rs902278303 Chr.7: 120951446-120951446 on Build GRCh38
rs770765336 Chr.7: 120951478-120951478 on Build GRCh38
rs932370241 Chr.7: 120951198-120951198 on Build GRCh38
rs553490361 Chr.7: 120951279-120951289 on Build GRCh38
rs776252766 Chr.7: 120951139-120951139 on Build GRCh38
rs943823958 Chr.7: 120951048-120951048 on Build GRCh38
rs919341013 Chr.7: 120951317-120951317 on Build GRCh38
rs536868779 Chr.7: 120951443-120951443 on Build GRCh38
rs759143530 Chr.7: 120951218-120951218 on Build GRCh38
rs1020663887 Chr.7: 120951415-120951415 on Build GRCh38
rs950165363 Chr.7: 120951096-120951096 on Build GRCh38
rs991468175 Chr.7: 120951104-120951104 on Build GRCh38
rs200539948 Chr.7: 120951250-120951250 on Build GRCh38
rs937820282 Chr.7: 120951077-120951077 on Build GRCh38

View Full Product Details