Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 120968999-120969531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ING3
Entrez Gene ID
54556
Gene Name
inhibitor of growth family member 3
Gene Aliases
Eaf4, ING2, MEAF4, p47ING3
UniGene
Hs.489811
Gene Chromosome Location
Chr.7: 120950755-120975657 on Build GRCh38
Transcripts
NM_019071.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7100378 c.836C>T p.S279L Chr.7: 120969132-120969132 on Build GRCh38
3632395 c.896G>T p.G299V Chr.7: 120969192-120969192 on Build GRCh38
3632396 c.898C>T p.R300* Chr.7: 120969194-120969194 on Build GRCh38
1084549 c.728G>A p.R243Q Chr.7: 120969024-120969024 on Build GRCh38
71166 c.809G>C p.R270T Chr.7: 120969105-120969105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553130683 Chr.7: 120969178-120969178 on Build GRCh38
rs112519217 Chr.7: 120969478-120969478 on Build GRCh38
rs938061161 Chr.7: 120969275-120969275 on Build GRCh38
rs2721372 Chr.7: 120969339-120969339 on Build GRCh38
rs989107358 Chr.7: 120969184-120969184 on Build GRCh38
rs745335917 Chr.7: 120969069-120969069 on Build GRCh38
rs758866749 Chr.7: 120969162-120969162 on Build GRCh38
rs200011060 Chr.7: 120969141-120969141 on Build GRCh38
rs753065874 Chr.7: 120969251-120969251 on Build GRCh38
rs548979831 Chr.7: 120969468-120969468 on Build GRCh38
rs762854929 Chr.7: 120969098-120969098 on Build GRCh38
rs953828612 Chr.7: 120969109-120969109 on Build GRCh38
rs779481523 Chr.7: 120969060-120969060 on Build GRCh38
rs1056888165 Chr.7: 120969304-120969304 on Build GRCh38
rs754084011 Chr.7: 120969231-120969231 on Build GRCh38
rs759722688 Chr.7: 120969246-120969246 on Build GRCh38
rs540514781 Chr.7: 120969486-120969486 on Build GRCh38
rs956892697 Chr.7: 120969143-120969143 on Build GRCh38
rs749714240 Chr.7: 120969029-120969029 on Build GRCh38
rs536061422 Chr.7: 120969133-120969133 on Build GRCh38
rs753080716 Chr.7: 120969161-120969161 on Build GRCh38
rs568506964 Chr.7: 120969104-120969104 on Build GRCh38
rs750568317 Chr.7: 120969117-120969117 on Build GRCh38
rs570602732 Chr.7: 120969354-120969354 on Build GRCh38
rs763727104 Chr.7: 120969099-120969099 on Build GRCh38
rs1005442027 Chr.7: 120969403-120969403 on Build GRCh38
rs998638571 Chr.7: 120969493-120969493 on Build GRCh38
rs538921370 Chr.7: 120969221-120969221 on Build GRCh38
rs766408610 Chr.7: 120969226-120969226 on Build GRCh38
rs200877009 Chr.7: 120969154-120969154 on Build GRCh38
rs184401227 Chr.7: 120969223-120969223 on Build GRCh38
rs745425792 Chr.7: 120969171-120969171 on Build GRCh38
rs778334927 Chr.7: 120969169-120969169 on Build GRCh38
rs557398817 Chr.7: 120969445-120969447 on Build GRCh38
rs937054698 Chr.7: 120969281-120969281 on Build GRCh38
rs768419875 Chr.7: 120969285-120969285 on Build GRCh38
rs769633520 Chr.7: 120969070-120969070 on Build GRCh38
rs116554148 Chr.7: 120969272-120969272 on Build GRCh38
rs766605352 Chr.7: 120969130-120969130 on Build GRCh38
rs761664024 Chr.7: 120969189-120969189 on Build GRCh38
rs774056554 Chr.7: 120969187-120969187 on Build GRCh38
rs141452615 Chr.7: 120969091-120969091 on Build GRCh38
rs781337340 Chr.7: 120969365-120969365 on Build GRCh38
rs554717118 Chr.7: 120969326-120969326 on Build GRCh38
rs150854781 Chr.7: 120969115-120969115 on Build GRCh38
rs140563514 Chr.7: 120969212-120969212 on Build GRCh38
rs148770498 Chr.7: 120969052-120969052 on Build GRCh38
rs897448437 Chr.7: 120969489-120969489 on Build GRCh38
rs995655632 Chr.7: 120969103-120969103 on Build GRCh38
rs569713487 Chr.7: 120969284-120969285 on Build GRCh38
rs560724979 Chr.7: 120969494-120969494 on Build GRCh38
rs756187407 Chr.7: 120969066-120969066 on Build GRCh38
rs761469817 Chr.7: 120969431-120969431 on Build GRCh38
rs1006647546 Chr.7: 120969128-120969128 on Build GRCh38
rs929816281 Chr.7: 120969331-120969331 on Build GRCh38
rs925352138 Chr.7: 120969274-120969274 on Build GRCh38
rs1018252472 Chr.7: 120969086-120969086 on Build GRCh38
rs748565699 Chr.7: 120969063-120969063 on Build GRCh38
rs866714436 Chr.7: 120969195-120969195 on Build GRCh38
rs765465420 Chr.7: 120969247-120969247 on Build GRCh38
rs748907453 Chr.7: 120969185-120969185 on Build GRCh38
rs988749099 Chr.7: 120969166-120969166 on Build GRCh38
rs753997717 Chr.7: 120969132-120969132 on Build GRCh38
rs776051200 Chr.7: 120969067-120969067 on Build GRCh38
rs769371186 Chr.7: 120969173-120969173 on Build GRCh38
rs758772364 Chr.7: 120969255-120969255 on Build GRCh38
rs115314385 Chr.7: 120969277-120969277 on Build GRCh38
rs768333923 Chr.7: 120969186-120969186 on Build GRCh38

View Full Product Details