Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 121339083-121339592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNT16
Entrez Gene ID
51384
Gene Name
Wnt family member 16
Gene Aliases
-
UniGene
Hs.272375
Gene Chromosome Location
Chr.7: 121325367-121341104 on Build GRCh38
Transcripts
NM_016087.2 NM_057168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7448983 c.1046A>G p.Y349C Chr.7: 121339293-121339293 on Build GRCh38
3831732 c.942C>T p.C314C Chr.7: 121339219-121339219 on Build GRCh38
6251342 c.1083C>A p.V361V Chr.7: 121339330-121339330 on Build GRCh38
6175909 c.886G>C p.E296Q Chr.7: 121339133-121339133 on Build GRCh38
387819 c.850C>G p.L284V Chr.7: 121339097-121339097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772403126 Chr.7: 121339256-121339256 on Build GRCh38
rs370654533 Chr.7: 121339288-121339288 on Build GRCh38
rs750894625 Chr.7: 121339162-121339162 on Build GRCh38
rs545777684 Chr.7: 121339404-121339404 on Build GRCh38
rs10632884 Chr.7: 121339473-121339474 on Build GRCh38
rs963498125 Chr.7: 121339497-121339497 on Build GRCh38
rs995673016 Chr.7: 121339556-121339556 on Build GRCh38
rs554766007 Chr.7: 121339182-121339182 on Build GRCh38
rs766409939 Chr.7: 121339274-121339274 on Build GRCh38
rs772492425 Chr.7: 121339394-121339394 on Build GRCh38
rs1046643988 Chr.7: 121339477-121339477 on Build GRCh38
rs757180081 Chr.7: 121339321-121339321 on Build GRCh38
rs780879168 Chr.7: 121339494-121339494 on Build GRCh38
rs181300392 Chr.7: 121339423-121339423 on Build GRCh38
rs773392753 Chr.7: 121339125-121339125 on Build GRCh38
rs931887257 Chr.7: 121339543-121339543 on Build GRCh38
rs768871697 Chr.7: 121339241-121339241 on Build GRCh38
rs762201457 Chr.7: 121339253-121339253 on Build GRCh38
rs768687126 Chr.7: 121339360-121339360 on Build GRCh38
rs773765415 Chr.7: 121339258-121339258 on Build GRCh38
rs763235672 Chr.7: 121339126-121339126 on Build GRCh38
rs757088066 Chr.7: 121339197-121339197 on Build GRCh38
rs538184954 Chr.7: 121339163-121339163 on Build GRCh38
rs373376386 Chr.7: 121339298-121339298 on Build GRCh38
rs539923811 Chr.7: 121339223-121339223 on Build GRCh38
rs576902188 Chr.7: 121339396-121339396 on Build GRCh38
rs780017797 Chr.7: 121339347-121339347 on Build GRCh38
rs533305922 Chr.7: 121339530-121339530 on Build GRCh38
rs761238720 Chr.7: 121339270-121339270 on Build GRCh38
rs769835842 Chr.7: 121339226-121339226 on Build GRCh38
rs547703351 Chr.7: 121339491-121339491 on Build GRCh38
rs750335090 Chr.7: 121339346-121339346 on Build GRCh38
rs143771072 Chr.7: 121339110-121339110 on Build GRCh38
rs17143305 Chr.7: 121339458-121339458 on Build GRCh38
rs748356342 Chr.7: 121339383-121339383 on Build GRCh38
rs752358411 Chr.7: 121339181-121339181 on Build GRCh38
rs147181856 Chr.7: 121339151-121339151 on Build GRCh38
rs1053840110 Chr.7: 121339447-121339447 on Build GRCh38
rs764950129 Chr.7: 121339376-121339377 on Build GRCh38
rs911828836 Chr.7: 121339567-121339567 on Build GRCh38
rs145703812 Chr.7: 121339426-121339426 on Build GRCh38
rs553849899 Chr.7: 121339356-121339356 on Build GRCh38
rs3832519 Chr.7: 121339471-121339472 on Build GRCh38
rs764373597 Chr.7: 121339136-121339136 on Build GRCh38
rs370474988 Chr.7: 121339304-121339304 on Build GRCh38
rs116618588 Chr.7: 121339190-121339190 on Build GRCh38
rs190011371 Chr.7: 121339514-121339514 on Build GRCh38
rs542118860 Chr.7: 121339427-121339427 on Build GRCh38
rs751234120 Chr.7: 121339315-121339315 on Build GRCh38
rs374417329 Chr.7: 121339172-121339172 on Build GRCh38
rs139571008 Chr.7: 121339224-121339224 on Build GRCh38
rs752203937 Chr.7: 121339292-121339292 on Build GRCh38
rs754785335 Chr.7: 121339420-121339420 on Build GRCh38
rs896937711 Chr.7: 121339537-121339537 on Build GRCh38
rs774637352 Chr.7: 121339144-121339144 on Build GRCh38
rs1051656684 Chr.7: 121339561-121339561 on Build GRCh38
rs774725194 Chr.7: 121339247-121339247 on Build GRCh38
rs781133598 Chr.7: 121339335-121339335 on Build GRCh38
rs773982060 Chr.7: 121339448-121339448 on Build GRCh38
rs545472355 Chr.7: 121339510-121339510 on Build GRCh38
rs570067881 Chr.7: 121339542-121339542 on Build GRCh38
rs185320709 Chr.7: 121339456-121339456 on Build GRCh38
rs377683444 Chr.7: 121339237-121339237 on Build GRCh38
rs936741463 Chr.7: 121339440-121339440 on Build GRCh38
rs1012918160 Chr.7: 121339149-121339149 on Build GRCh38
rs373809618 Chr.7: 121339371-121339371 on Build GRCh38
rs775576446 Chr.7: 121339227-121339227 on Build GRCh38
rs771323229 Chr.7: 121339524-121339524 on Build GRCh38
rs777735313 Chr.7: 121339183-121339183 on Build GRCh38
rs376388838 Chr.7: 121339299-121339299 on Build GRCh38
rs906780980 Chr.7: 121339487-121339487 on Build GRCh38
rs181639902 Chr.7: 121339532-121339532 on Build GRCh38
rs767969036 Chr.7: 121339154-121339154 on Build GRCh38
rs983482831 Chr.7: 121339488-121339488 on Build GRCh38

View Full Product Details