Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151175617-151176119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB10
Entrez Gene ID
136371
Gene Name
ankyrin repeat and SOCS box containing 10
Gene Aliases
GLC1F
UniGene
Hs.647081
Gene Chromosome Location
Chr.7: 151175698-151187832 on Build GRCh38
Transcripts
NM_001142459.1 NM_001142460.1 NM_080871.3

SNP Detail

SNP ID Chromosome Location
rs973524161 Chr.7: 151176013-151176013 on Build GRCh38
rs549435570 Chr.7: 151175725-151175725 on Build GRCh38
rs310597 Chr.7: 151175959-151175959 on Build GRCh38
rs188161385 Chr.7: 151175912-151175912 on Build GRCh38
rs772012596 Chr.7: 151175966-151175966 on Build GRCh38
rs956169774 Chr.7: 151176051-151176051 on Build GRCh38
rs1024240570 Chr.7: 151175958-151175958 on Build GRCh38
rs754279733 Chr.7: 151176095-151176095 on Build GRCh38
rs144447072 Chr.7: 151175647-151175647 on Build GRCh38
rs754530729 Chr.7: 151175811-151175811 on Build GRCh38
rs1013238466 Chr.7: 151175943-151175943 on Build GRCh38
rs750963428 Chr.7: 151176092-151176092 on Build GRCh38
rs1041423937 Chr.7: 151175857-151175857 on Build GRCh38
rs890604694 Chr.7: 151175823-151175823 on Build GRCh38
rs778997476 Chr.7: 151175921-151175921 on Build GRCh38
rs10251269 Chr.7: 151175657-151175657 on Build GRCh38
rs1044026199 Chr.7: 151175828-151175828 on Build GRCh38
rs932028985 Chr.7: 151176079-151176079 on Build GRCh38
rs914298363 Chr.7: 151175812-151175812 on Build GRCh38
rs552296281 Chr.7: 151175660-151175660 on Build GRCh38
rs926986076 Chr.7: 151175851-151175851 on Build GRCh38
rs534918152 Chr.7: 151175993-151175993 on Build GRCh38
rs902940814 Chr.7: 151175872-151175872 on Build GRCh38
rs1024542270 Chr.7: 151175643-151175643 on Build GRCh38
rs973409805 Chr.7: 151175761-151175761 on Build GRCh38
rs572360917 Chr.7: 151175996-151175996 on Build GRCh38
rs982614561 Chr.7: 151175967-151175967 on Build GRCh38
rs576489815 Chr.7: 151175950-151175950 on Build GRCh38
rs751615686 Chr.7: 151176073-151176073 on Build GRCh38
rs756697942 Chr.7: 151176094-151176094 on Build GRCh38
rs554606785 Chr.7: 151175822-151175822 on Build GRCh38
rs988573690 Chr.7: 151176064-151176064 on Build GRCh38
rs996287982 Chr.7: 151175970-151175970 on Build GRCh38
rs1029029540 Chr.7: 151175668-151175668 on Build GRCh38
rs932222046 Chr.7: 151175816-151175816 on Build GRCh38
rs761834358 Chr.7: 151176074-151176074 on Build GRCh38
rs539814940 Chr.7: 151175910-151175910 on Build GRCh38
rs1028998224 Chr.7: 151176006-151176006 on Build GRCh38
rs753346018 Chr.7: 151175649-151175649 on Build GRCh38
rs565475894 Chr.7: 151176007-151176007 on Build GRCh38
rs574073787 Chr.7: 151175862-151175862 on Build GRCh38
rs1019488712 Chr.7: 151176018-151176018 on Build GRCh38
rs764319277 Chr.7: 151176071-151176071 on Build GRCh38
rs920639874 Chr.7: 151175784-151175784 on Build GRCh38
rs920573014 Chr.7: 151176043-151176043 on Build GRCh38
rs768043608 Chr.7: 151176089-151176089 on Build GRCh38
rs894167089 Chr.7: 151175937-151175937 on Build GRCh38
rs955865993 Chr.7: 151175805-151175805 on Build GRCh38
rs1016321236 Chr.7: 151175656-151175656 on Build GRCh38
rs538138706 Chr.7: 151175821-151175821 on Build GRCh38
rs944888095 Chr.7: 151175838-151175838 on Build GRCh38

View Full Product Details