Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151466947-151467445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHEB
Entrez Gene ID
6009
Gene Name
Ras homolog enriched in brain
Gene Aliases
RHEB2
UniGene
Hs.283521
Gene Chromosome Location
Chr.7: 151466012-151519924 on Build GRCh38
Transcripts
NM_005614.3 XM_011516457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6940364 c.505A>C p.K169Q Chr.7: 151467169-151467169 on Build GRCh38
5656380 c.539_540insT p.C181fs*>5 Chr.7: 151467134-151467135 on Build GRCh38
6306520 c.536C>A p.S179Y Chr.7: 151467138-151467138 on Build GRCh38
400688 c.517G>A p.A173T Chr.7: 151467157-151467157 on Build GRCh38
3262185 c.503A>G p.E168G Chr.7: 151467171-151467171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893076765 Chr.7: 151467311-151467311 on Build GRCh38
rs781512533 Chr.7: 151467192-151467192 on Build GRCh38
rs752009019 Chr.7: 151467166-151467166 on Build GRCh38
rs767187355 Chr.7: 151467144-151467144 on Build GRCh38
rs773101365 Chr.7: 151467242-151467242 on Build GRCh38
rs1022807069 Chr.7: 151467361-151467361 on Build GRCh38
rs745538266 Chr.7: 151467223-151467223 on Build GRCh38
rs1026251059 Chr.7: 151467416-151467416 on Build GRCh38
rs757654395 Chr.7: 151467263-151467263 on Build GRCh38
rs774411561 Chr.7: 151467229-151467229 on Build GRCh38
rs1039678062 Chr.7: 151467417-151467417 on Build GRCh38
rs9768124 Chr.7: 151467185-151467185 on Build GRCh38
rs370458727 Chr.7: 151467101-151467101 on Build GRCh38
rs142694904 Chr.7: 151467274-151467274 on Build GRCh38
rs373682217 Chr.7: 151467246-151467246 on Build GRCh38
rs766642352 Chr.7: 151467239-151467240 on Build GRCh38
rs1031574426 Chr.7: 151467054-151467054 on Build GRCh38
rs949853261 Chr.7: 151467141-151467141 on Build GRCh38
rs998819478 Chr.7: 151467052-151467052 on Build GRCh38
rs148315772 Chr.7: 151467163-151467163 on Build GRCh38
rs774927561 Chr.7: 151467100-151467100 on Build GRCh38
rs184654781 Chr.7: 151467035-151467035 on Build GRCh38
rs1035498972 Chr.7: 151467364-151467364 on Build GRCh38
rs768036177 Chr.7: 151467228-151467228 on Build GRCh38
rs529019735 Chr.7: 151467301-151467301 on Build GRCh38
rs745486349 Chr.7: 151467096-151467096 on Build GRCh38
rs773554563 Chr.7: 151467070-151467072 on Build GRCh38
rs368150276 Chr.7: 151467220-151467220 on Build GRCh38
rs755720023 Chr.7: 151467225-151467225 on Build GRCh38
rs769340964 Chr.7: 151467099-151467099 on Build GRCh38
rs777044574 Chr.7: 151467154-151467154 on Build GRCh38
rs201092245 Chr.7: 151467238-151467238 on Build GRCh38
rs9770800 Chr.7: 151467079-151467079 on Build GRCh38
rs571546215 Chr.7: 151467226-151467226 on Build GRCh38
rs760898792 Chr.7: 151467221-151467221 on Build GRCh38
rs752762612 Chr.7: 151467162-151467162 on Build GRCh38
rs115795498 Chr.7: 151467386-151467386 on Build GRCh38
rs17635434 Chr.7: 151467262-151467262 on Build GRCh38
rs764052422 Chr.7: 151467259-151467259 on Build GRCh38
rs946291733 Chr.7: 151467062-151467062 on Build GRCh38
rs551387324 Chr.7: 151467160-151467160 on Build GRCh38
rs550467806 Chr.7: 151467305-151467305 on Build GRCh38
rs995128475 Chr.7: 151467215-151467216 on Build GRCh38
rs9768956 Chr.7: 151467103-151467103 on Build GRCh38
rs952722238 Chr.7: 151467027-151467027 on Build GRCh38
rs928021516 Chr.7: 151467249-151467249 on Build GRCh38
rs775845378 Chr.7: 151467253-151467253 on Build GRCh38
rs960769743 Chr.7: 151467365-151467365 on Build GRCh38
rs949830614 Chr.7: 151467355-151467356 on Build GRCh38
rs1007680098 Chr.7: 151466972-151466972 on Build GRCh38
rs977312227 Chr.7: 151467055-151467055 on Build GRCh38
rs748013851 Chr.7: 151467232-151467232 on Build GRCh38
rs969504133 Chr.7: 151467214-151467214 on Build GRCh38
rs559587654 Chr.7: 151467117-151467117 on Build GRCh38
rs937006710 Chr.7: 151467085-151467085 on Build GRCh38
rs1011459770 Chr.7: 151467326-151467326 on Build GRCh38
rs913424846 Chr.7: 151467059-151467059 on Build GRCh38
rs776828341 Chr.7: 151467253-151467253 on Build GRCh38
rs188394856 Chr.7: 151467275-151467275 on Build GRCh38
rs1026934560 Chr.7: 151467012-151467012 on Build GRCh38
rs764723561 Chr.7: 151467164-151467164 on Build GRCh38
rs768571063 Chr.7: 151467111-151467111 on Build GRCh38
rs910676422 Chr.7: 151467288-151467288 on Build GRCh38
rs965828523 Chr.7: 151467240-151467240 on Build GRCh38
rs758899248 Chr.7: 151467078-151467078 on Build GRCh38
rs146509587 Chr.7: 151467294-151467294 on Build GRCh38
rs573203748 Chr.7: 151467289-151467289 on Build GRCh38
rs9770802 Chr.7: 151467203-151467203 on Build GRCh38
rs528741864 Chr.7: 151467134-151467134 on Build GRCh38
rs905685763 Chr.7: 151467362-151467362 on Build GRCh38
rs978982983 Chr.7: 151467285-151467285 on Build GRCh38
rs972314680 Chr.7: 151467261-151467261 on Build GRCh38
rs916987578 Chr.7: 151467128-151467128 on Build GRCh38
rs888423835 Chr.7: 151467235-151467235 on Build GRCh38
rs1040522918 Chr.7: 151467293-151467293 on Build GRCh38
rs527268685 Chr.7: 151467363-151467363 on Build GRCh38
rs186659694 Chr.7: 151467004-151467004 on Build GRCh38
rs143352740 Chr.7: 151467161-151467161 on Build GRCh38
rs994803138 Chr.7: 151467005-151467005 on Build GRCh38
rs763161814 Chr.7: 151467254-151467254 on Build GRCh38
rs544300269 Chr.7: 151467415-151467415 on Build GRCh38
rs771926122 Chr.7: 151467233-151467233 on Build GRCh38
rs1036120265 Chr.7: 151467356-151467356 on Build GRCh38
rs376087347 Chr.7: 151467084-151467084 on Build GRCh38
rs916957918 Chr.7: 151467321-151467321 on Build GRCh38
rs373922071 Chr.7: 151467080-151467080 on Build GRCh38
rs991133349 Chr.7: 151467309-151467309 on Build GRCh38
rs759843219 Chr.7: 151467245-151467245 on Build GRCh38
rs113627586 Chr.7: 151467028-151467028 on Build GRCh38
rs563918409 Chr.7: 151467027-151467027 on Build GRCh38

View Full Product Details