Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161099256-161099752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHDC9
Entrez Gene ID
126823
Gene Name
kelch domain containing 9
Gene Aliases
KARCA1
UniGene
Hs.507290
Gene Chromosome Location
Chr.1: 161098361-161100348 on Build GRCh38
Transcripts
NM_001007255.2 NM_152366.4 NR_033385.1 NR_033386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1335615 c.779A>G p.E260G Chr.1: 161099670-161099670 on Build GRCh38
6320120 c.759G>T p.Q253H Chr.1: 161099669-161099669 on Build GRCh38
1255951 c.561_562insT p.S188fs*29 Chr.1: 161099379-161099380 on Build GRCh38
6248935 c.763G>T p.G255W Chr.1: 161099673-161099673 on Build GRCh38
4024309 c.716T>C p.M239T Chr.1: 161099626-161099626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150493322 Chr.1: 161099487-161099487 on Build GRCh38
rs200796951 Chr.1: 161099408-161099408 on Build GRCh38
rs750341230 Chr.1: 161099565-161099565 on Build GRCh38
rs760903002 Chr.1: 161099421-161099421 on Build GRCh38
rs758973669 Chr.1: 161099593-161099593 on Build GRCh38
rs765156889 Chr.1: 161099512-161099512 on Build GRCh38
rs771401029 Chr.1: 161099299-161099299 on Build GRCh38
rs142593956 Chr.1: 161099493-161099493 on Build GRCh38
rs768750539 Chr.1: 161099597-161099597 on Build GRCh38
rs376790387 Chr.1: 161099381-161099381 on Build GRCh38
rs771398646 Chr.1: 161099399-161099399 on Build GRCh38
rs201362134 Chr.1: 161099326-161099326 on Build GRCh38
rs751172376 Chr.1: 161099563-161099563 on Build GRCh38
rs778286163 Chr.1: 161099394-161099394 on Build GRCh38
rs964948804 Chr.1: 161099465-161099465 on Build GRCh38
rs576638208 Chr.1: 161099286-161099286 on Build GRCh38
rs761897468 Chr.1: 161099538-161099538 on Build GRCh38
rs749658678 Chr.1: 161099397-161099397 on Build GRCh38
rs755674250 Chr.1: 161099575-161099575 on Build GRCh38
rs766413650 Chr.1: 161099558-161099558 on Build GRCh38
rs374767652 Chr.1: 161099572-161099572 on Build GRCh38
rs772822250 Chr.1: 161099542-161099542 on Build GRCh38
rs771296778 Chr.1: 161099496-161099496 on Build GRCh38
rs201726019 Chr.1: 161099643-161099643 on Build GRCh38
rs747145725 Chr.1: 161099596-161099596 on Build GRCh38
rs375259316 Chr.1: 161099688-161099688 on Build GRCh38
rs142312611 Chr.1: 161099436-161099436 on Build GRCh38
rs746154278 Chr.1: 161099591-161099591 on Build GRCh38
rs961885573 Chr.1: 161099424-161099424 on Build GRCh38
rs749947043 Chr.1: 161099377-161099377 on Build GRCh38
rs752092667 Chr.1: 161099678-161099678 on Build GRCh38
rs541844589 Chr.1: 161099346-161099346 on Build GRCh38
rs752095931 Chr.1: 161099712-161099712 on Build GRCh38
rs757150236 Chr.1: 161099351-161099351 on Build GRCh38
rs369595659 Chr.1: 161099673-161099673 on Build GRCh38
rs775555319 Chr.1: 161099520-161099520 on Build GRCh38
rs374276703 Chr.1: 161099297-161099298 on Build GRCh38
rs750860573 Chr.1: 161099689-161099689 on Build GRCh38
rs372802596 Chr.1: 161099681-161099681 on Build GRCh38
rs776520482 Chr.1: 161099604-161099604 on Build GRCh38
rs781348591 Chr.1: 161099729-161099729 on Build GRCh38
rs753690320 Chr.1: 161099405-161099405 on Build GRCh38
rs78500642 Chr.1: 161099656-161099656 on Build GRCh38
rs1054597674 Chr.1: 161099709-161099709 on Build GRCh38
rs773659781 Chr.1: 161099293-161099293 on Build GRCh38
rs556236171 Chr.1: 161099292-161099292 on Build GRCh38
rs751045637 Chr.1: 161099476-161099476 on Build GRCh38
rs769839703 Chr.1: 161099615-161099615 on Build GRCh38
rs752019069 Chr.1: 161099299-161099299 on Build GRCh38
rs373092571 Chr.1: 161099546-161099546 on Build GRCh38
rs750402122 Chr.1: 161099449-161099449 on Build GRCh38
rs777604391 Chr.1: 161099706-161099733 on Build GRCh38
rs753702777 Chr.1: 161099428-161099428 on Build GRCh38
rs562159459 Chr.1: 161099445-161099445 on Build GRCh38
rs775500503 Chr.1: 161099414-161099414 on Build GRCh38
rs763565517 Chr.1: 161099297-161099297 on Build GRCh38
rs760848121 Chr.1: 161099342-161099342 on Build GRCh38
rs772512875 Chr.1: 161099514-161099514 on Build GRCh38
rs868821453 Chr.1: 161099544-161099544 on Build GRCh38
rs748559836 Chr.1: 161099291-161099291 on Build GRCh38
rs186325496 Chr.1: 161099288-161099288 on Build GRCh38
rs778909363 Chr.1: 161099372-161099372 on Build GRCh38
rs745320353 Chr.1: 161099280-161099280 on Build GRCh38
rs761650296 Chr.1: 161099505-161099505 on Build GRCh38
rs200211462 Chr.1: 161099573-161099573 on Build GRCh38
rs755347288 Chr.1: 161099614-161099616 on Build GRCh38
rs768928324 Chr.1: 161099534-161099534 on Build GRCh38
rs140541574 Chr.1: 161099451-161099451 on Build GRCh38
rs562131728 Chr.1: 161099378-161099378 on Build GRCh38
rs758744538 Chr.1: 161099691-161099691 on Build GRCh38
rs929144879 Chr.1: 161099495-161099495 on Build GRCh38
rs752886941 Chr.1: 161099333-161099333 on Build GRCh38
rs765099802 Chr.1: 161099540-161099540 on Build GRCh38
rs754743396 Chr.1: 161099484-161099484 on Build GRCh38
rs747090793 Chr.1: 161099527-161099527 on Build GRCh38
rs759648961 Chr.1: 161099325-161099325 on Build GRCh38
rs772226634 Chr.1: 161099410-161099410 on Build GRCh38
rs776877369 Chr.1: 161099536-161099536 on Build GRCh38
rs764237325 Chr.1: 161099422-161099422 on Build GRCh38
rs867968350 Chr.1: 161099296-161099297 on Build GRCh38
rs755442434 Chr.1: 161099393-161099393 on Build GRCh38
rs779722431 Chr.1: 161099466-161099466 on Build GRCh38
rs753574944 Chr.1: 161099348-161099348 on Build GRCh38
rs773124583 Chr.1: 161099622-161099622 on Build GRCh38
rs758456974 Chr.1: 161099720-161099720 on Build GRCh38
rs201094088 Chr.1: 161099711-161099711 on Build GRCh38
rs866213235 Chr.1: 161099598-161099598 on Build GRCh38
rs779351519 Chr.1: 161099576-161099576 on Build GRCh38
rs748413149 Chr.1: 161099607-161099607 on Build GRCh38

View Full Product Details