Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 162854555-162855095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC190
Entrez Gene ID
339512
Gene Name
coiled-coil domain containing 190
Gene Aliases
C1orf110
UniGene
Hs.407631
Gene Chromosome Location
Chr.1: 162851291-162868918 on Build GRCh38
Transcripts
NM_178550.4 XM_005245125.3 XM_006711289.3 XM_011509477.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6246015 c.711C>A p.F237L Chr.1: 162854963-162854963 on Build GRCh38
4024621 c.607A>T p.M203L Chr.1: 162855067-162855067 on Build GRCh38
3976440 c.887C>T p.P296L Chr.1: 162854787-162854787 on Build GRCh38
3732574 c.854C>G p.S285C Chr.1: 162854820-162854820 on Build GRCh38
5889452 c.679C>T p.P227S Chr.1: 162854995-162854995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775727372 Chr.1: 162854900-162854900 on Build GRCh38
rs772401179 Chr.1: 162855018-162855018 on Build GRCh38
rs766804954 Chr.1: 162854793-162854793 on Build GRCh38
rs544714388 Chr.1: 162854937-162854937 on Build GRCh38
rs752247770 Chr.1: 162855061-162855061 on Build GRCh38
rs201358928 Chr.1: 162854957-162854957 on Build GRCh38
rs756251822 Chr.1: 162854821-162854821 on Build GRCh38
rs745690653 Chr.1: 162854875-162854875 on Build GRCh38
rs746489294 Chr.1: 162854884-162854884 on Build GRCh38
rs765990205 Chr.1: 162854880-162854880 on Build GRCh38
rs749768117 Chr.1: 162854894-162854894 on Build GRCh38
rs763391319 Chr.1: 162854784-162854784 on Build GRCh38
rs745470607 Chr.1: 162854756-162854756 on Build GRCh38
rs768879364 Chr.1: 162854989-162854989 on Build GRCh38
rs754616520 Chr.1: 162855038-162855038 on Build GRCh38
rs1053374775 Chr.1: 162854712-162854712 on Build GRCh38
rs374875454 Chr.1: 162854892-162854892 on Build GRCh38
rs374822482 Chr.1: 162854997-162854997 on Build GRCh38
rs779553524 Chr.1: 162854946-162854946 on Build GRCh38
rs771361068 Chr.1: 162854867-162854867 on Build GRCh38
rs752704285 Chr.1: 162854816-162854816 on Build GRCh38
rs192651726 Chr.1: 162854880-162854880 on Build GRCh38
rs368571297 Chr.1: 162854837-162854837 on Build GRCh38
rs368241445 Chr.1: 162854942-162854942 on Build GRCh38
rs750426484 Chr.1: 162855013-162855013 on Build GRCh38
rs1041279719 Chr.1: 162854818-162854818 on Build GRCh38
rs372036844 Chr.1: 162854956-162854956 on Build GRCh38
rs184192552 Chr.1: 162855072-162855072 on Build GRCh38
rs145929601 Chr.1: 162854623-162854623 on Build GRCh38
rs373424324 Chr.1: 162855059-162855059 on Build GRCh38
rs370149150 Chr.1: 162854885-162854885 on Build GRCh38
rs377391335 Chr.1: 162854762-162854762 on Build GRCh38
rs367988830 Chr.1: 162854904-162854904 on Build GRCh38
rs78851184 Chr.1: 162854763-162854763 on Build GRCh38
rs777124304 Chr.1: 162854750-162854762 on Build GRCh38
rs200349261 Chr.1: 162854983-162854983 on Build GRCh38
rs762021462 Chr.1: 162855007-162855007 on Build GRCh38
rs201168794 Chr.1: 162854963-162854963 on Build GRCh38
rs990666922 Chr.1: 162854598-162854598 on Build GRCh38
rs373112550 Chr.1: 162854754-162854754 on Build GRCh38
rs565529537 Chr.1: 162854614-162854614 on Build GRCh38
rs765058279 Chr.1: 162855009-162855009 on Build GRCh38
rs200708375 Chr.1: 162854891-162854891 on Build GRCh38
rs907796949 Chr.1: 162854633-162854633 on Build GRCh38
rs773898696 Chr.1: 162854781-162854781 on Build GRCh38
rs1027668801 Chr.1: 162854840-162854840 on Build GRCh38
rs770770533 Chr.1: 162854626-162854626 on Build GRCh38
rs749076058 Chr.1: 162854733-162854733 on Build GRCh38
rs778506006 Chr.1: 162854868-162854868 on Build GRCh38
rs1037026424 Chr.1: 162854681-162854681 on Build GRCh38
rs774519632 Chr.1: 162854895-162854895 on Build GRCh38
rs765279608 Chr.1: 162854934-162854934 on Build GRCh38
rs760000255 Chr.1: 162854896-162854896 on Build GRCh38
rs148693034 Chr.1: 162854686-162854686 on Build GRCh38
rs746186019 Chr.1: 162854972-162854972 on Build GRCh38
rs774805555 Chr.1: 162854804-162854804 on Build GRCh38
rs372511486 Chr.1: 162854986-162854986 on Build GRCh38
rs375511140 Chr.1: 162854922-162854922 on Build GRCh38
rs551401189 Chr.1: 162854853-162854853 on Build GRCh38
rs369842895 Chr.1: 162854985-162854985 on Build GRCh38
rs753821131 Chr.1: 162854844-162854844 on Build GRCh38
rs750181807 Chr.1: 162854941-162854941 on Build GRCh38
rs769746551 Chr.1: 162855020-162855020 on Build GRCh38
rs528968419 Chr.1: 162854697-162854697 on Build GRCh38
rs916410705 Chr.1: 162854608-162854608 on Build GRCh38
rs762526915 Chr.1: 162854810-162854811 on Build GRCh38
rs773768727 Chr.1: 162854879-162854880 on Build GRCh38
rs372194632 Chr.1: 162854920-162854920 on Build GRCh38
rs920019610 Chr.1: 162854715-162854715 on Build GRCh38
rs369176927 Chr.1: 162855030-162855030 on Build GRCh38
rs187742957 Chr.1: 162854772-162854772 on Build GRCh38
rs767790051 Chr.1: 162854812-162854812 on Build GRCh38
rs757083415 Chr.1: 162854734-162854734 on Build GRCh38
rs760721206 Chr.1: 162854901-162854901 on Build GRCh38
rs751401369 Chr.1: 162855031-162855031 on Build GRCh38
rs779538866 Chr.1: 162854971-162854971 on Build GRCh38
rs78906366 Chr.1: 162854589-162854589 on Build GRCh38
rs533306514 Chr.1: 162854631-162854631 on Build GRCh38
rs780778113 Chr.1: 162854962-162854962 on Build GRCh38
rs961972943 Chr.1: 162855055-162855055 on Build GRCh38
rs759608496 Chr.1: 162854806-162854806 on Build GRCh38
rs75707515 Chr.1: 162854677-162854677 on Build GRCh38
rs545000326 Chr.1: 162855074-162855074 on Build GRCh38

View Full Product Details