Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 166918516-166919017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ILDR2
Entrez Gene ID
387597
Gene Name
immunoglobulin like domain containing receptor 2
Gene Aliases
C1orf32, dJ782G3.1
UniGene
Hs.444835
Gene Chromosome Location
Chr.1: 166913204-166975324 on Build GRCh38
Transcripts
NM_199351.2 XM_006711314.3 XM_017001252.1 XM_017001253.1 XM_017001254.1 XM_017001255.1 XM_017001256.1

SNP Detail

SNP ID Chromosome Location
rs907520738 Chr.1: 166918583-166918583 on Build GRCh38
rs1010871635 Chr.1: 166918796-166918796 on Build GRCh38
rs973925989 Chr.1: 166918811-166918811 on Build GRCh38
rs973010297 Chr.1: 166918982-166918982 on Build GRCh38
rs1018123498 Chr.1: 166918742-166918742 on Build GRCh38
rs1029812139 Chr.1: 166918898-166918898 on Build GRCh38
rs1008285774 Chr.1: 166918733-166918733 on Build GRCh38
rs192015835 Chr.1: 166918677-166918677 on Build GRCh38
rs997034058 Chr.1: 166918835-166918835 on Build GRCh38
rs961968143 Chr.1: 166918604-166918604 on Build GRCh38
rs755922393 Chr.1: 166918852-166918852 on Build GRCh38
rs144718048 Chr.1: 166918605-166918605 on Build GRCh38
rs951314312 Chr.1: 166918888-166918888 on Build GRCh38
rs866991125 Chr.1: 166918869-166918869 on Build GRCh38
rs112726172 Chr.1: 166918735-166918735 on Build GRCh38
rs765257682 Chr.1: 166918792-166918798 on Build GRCh38
rs1008639304 Chr.1: 166918927-166918927 on Build GRCh38
rs928058751 Chr.1: 166918657-166918657 on Build GRCh38
rs753648063 Chr.1: 166918976-166918976 on Build GRCh38
rs1030983134 Chr.1: 166918710-166918710 on Build GRCh38
rs1012454833 Chr.1: 166918634-166918634 on Build GRCh38
rs563258367 Chr.1: 166918616-166918616 on Build GRCh38
rs879018125 Chr.1: 166918699-166918699 on Build GRCh38
rs1052895138 Chr.1: 166918737-166918737 on Build GRCh38
rs564187097 Chr.1: 166918929-166918929 on Build GRCh38
rs954643877 Chr.1: 166918902-166918902 on Build GRCh38
rs892459842 Chr.1: 166918794-166918794 on Build GRCh38
rs904094918 Chr.1: 166918649-166918649 on Build GRCh38
rs954461714 Chr.1: 166918566-166918566 on Build GRCh38
rs545888981 Chr.1: 166918656-166918656 on Build GRCh38
rs928399538 Chr.1: 166918549-166918549 on Build GRCh38
rs963838742 Chr.1: 166918763-166918763 on Build GRCh38
rs938423483 Chr.1: 166918565-166918565 on Build GRCh38
rs891243495 Chr.1: 166918714-166918714 on Build GRCh38
rs928294212 Chr.1: 166918975-166918975 on Build GRCh38
rs1027356783 Chr.1: 166918824-166918824 on Build GRCh38
rs987566125 Chr.1: 166918567-166918567 on Build GRCh38
rs900054378 Chr.1: 166918832-166918834 on Build GRCh38
rs1029170184 Chr.1: 166918564-166918564 on Build GRCh38
rs939435718 Chr.1: 166918713-166918713 on Build GRCh38
rs575137967 Chr.1: 166918617-166918617 on Build GRCh38
rs1044008773 Chr.1: 166918819-166918819 on Build GRCh38
rs759601802 Chr.1: 166918790-166918790 on Build GRCh38
rs1044332744 Chr.1: 166918636-166918636 on Build GRCh38

View Full Product Details