Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 80637268-80637773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF704
Entrez Gene ID
619279
Gene Name
zinc finger protein 704
Gene Aliases
Gig1
UniGene
Hs.434957
Gene Chromosome Location
Chr.8: 80628451-80884416 on Build GRCh38
Transcripts
NM_001033723.2 XM_005251279.4 XM_017013725.1

SNP Detail

SNP ID Chromosome Location
rs1048472741 Chr.8: 80637484-80637484 on Build GRCh38
rs912139931 Chr.8: 80637505-80637505 on Build GRCh38
rs776800073 Chr.8: 80637308-80637308 on Build GRCh38
rs935709193 Chr.8: 80637575-80637575 on Build GRCh38
rs973393766 Chr.8: 80637367-80637367 on Build GRCh38
rs58984500 Chr.8: 80637712-80637712 on Build GRCh38
rs936012138 Chr.8: 80637462-80637462 on Build GRCh38
rs181027435 Chr.8: 80637373-80637373 on Build GRCh38
rs539529030 Chr.8: 80637540-80637540 on Build GRCh38
rs1039165372 Chr.8: 80637567-80637567 on Build GRCh38
rs116587489 Chr.8: 80637369-80637369 on Build GRCh38
rs907340523 Chr.8: 80637520-80637521 on Build GRCh38
rs866626240 Chr.8: 80637676-80637676 on Build GRCh38
rs975261232 Chr.8: 80637331-80637331 on Build GRCh38
rs1041786119 Chr.8: 80637427-80637427 on Build GRCh38
rs1055137116 Chr.8: 80637477-80637477 on Build GRCh38
rs900272073 Chr.8: 80637568-80637568 on Build GRCh38
rs551682833 Chr.8: 80637413-80637413 on Build GRCh38
rs942552371 Chr.8: 80637307-80637308 on Build GRCh38
rs114153483 Chr.8: 80637675-80637675 on Build GRCh38
rs922403715 Chr.8: 80637350-80637350 on Build GRCh38
rs569648166 Chr.8: 80637701-80637701 on Build GRCh38
rs1013556820 Chr.8: 80637485-80637485 on Build GRCh38
rs151265170 Chr.8: 80637449-80637449 on Build GRCh38
rs533276200 Chr.8: 80637727-80637727 on Build GRCh38
rs1017294255 Chr.8: 80637682-80637682 on Build GRCh38
rs1024674633 Chr.8: 80637692-80637692 on Build GRCh38
rs373901063 Chr.8: 80637603-80637604 on Build GRCh38
rs559810294 Chr.8: 80637372-80637372 on Build GRCh38
rs944877314 Chr.8: 80637551-80637551 on Build GRCh38
rs775537974 Chr.8: 80637691-80637691 on Build GRCh38
rs35675165 Chr.8: 80637453-80637454 on Build GRCh38

View Full Product Details