Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 88041546-88041973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP16
Entrez Gene ID
4325
Gene Name
matrix metallopeptidase 16
Gene Aliases
C8orf57, MMP-X2, MT-MMP2, MT-MMP3, MT3-MMP
UniGene
Hs.492187
Gene Chromosome Location
Chr.8: 88037232-88327488 on Build GRCh38
Transcripts
NM_005941.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6041723 c.1642G>T p.D548Y Chr.8: 88041643-88041643 on Build GRCh38
2790062 c.1560A>T p.G520G Chr.8: 88041725-88041725 on Build GRCh38
354658 c.1636C>A p.P546T Chr.8: 88041649-88041649 on Build GRCh38
5988111 c.1603C>A p.P535T Chr.8: 88041682-88041682 on Build GRCh38
2790060 c.1590G>A p.M530I Chr.8: 88041695-88041695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911606002 Chr.8: 88041584-88041584 on Build GRCh38
rs751418739 Chr.8: 88041846-88041846 on Build GRCh38
rs921880030 Chr.8: 88041735-88041735 on Build GRCh38
rs553868301 Chr.8: 88041576-88041576 on Build GRCh38
rs775987201 Chr.8: 88041788-88041788 on Build GRCh38
rs748797692 Chr.8: 88041810-88041811 on Build GRCh38
rs780135772 Chr.8: 88041832-88041832 on Build GRCh38
rs1003657586 Chr.8: 88041890-88041890 on Build GRCh38
rs762633102 Chr.8: 88041745-88041745 on Build GRCh38
rs779385399 Chr.8: 88041807-88041811 on Build GRCh38
rs754316702 Chr.8: 88041804-88041804 on Build GRCh38
rs924381693 Chr.8: 88041573-88041573 on Build GRCh38
rs200606225 Chr.8: 88041601-88041601 on Build GRCh38
rs141851128 Chr.8: 88041825-88041825 on Build GRCh38
rs772326236 Chr.8: 88041767-88041767 on Build GRCh38
rs1056564013 Chr.8: 88041928-88041928 on Build GRCh38
rs764500669 Chr.8: 88041571-88041571 on Build GRCh38
rs758151246 Chr.8: 88041606-88041606 on Build GRCh38
rs542654507 Chr.8: 88041678-88041678 on Build GRCh38
rs749270258 Chr.8: 88041929-88041929 on Build GRCh38
rs1051789 Chr.8: 88041724-88041724 on Build GRCh38
rs920620102 Chr.8: 88041811-88041811 on Build GRCh38
rs768097456 Chr.8: 88041812-88041813 on Build GRCh38
rs147103821 Chr.8: 88041635-88041635 on Build GRCh38
rs747698919 Chr.8: 88041684-88041684 on Build GRCh38
rs762525559 Chr.8: 88041808-88041808 on Build GRCh38
rs768707886 Chr.8: 88041671-88041671 on Build GRCh38
rs1004177288 Chr.8: 88041807-88041809 on Build GRCh38
rs181678413 Chr.8: 88041816-88041816 on Build GRCh38
rs750607147 Chr.8: 88041604-88041604 on Build GRCh38
rs746926391 Chr.8: 88041656-88041656 on Build GRCh38
rs773017409 Chr.8: 88041719-88041719 on Build GRCh38
rs199542125 Chr.8: 88041707-88041707 on Build GRCh38
rs955897945 Chr.8: 88041633-88041633 on Build GRCh38
rs764713557 Chr.8: 88041800-88041800 on Build GRCh38
rs551470820 Chr.8: 88041844-88041844 on Build GRCh38
rs886561185 Chr.8: 88041842-88041842 on Build GRCh38
rs866625908 Chr.8: 88041774-88041774 on Build GRCh38
rs28988879 Chr.8: 88041804-88041804 on Build GRCh38
rs565147285 Chr.8: 88041874-88041874 on Build GRCh38
rs1011153208 Chr.8: 88041894-88041894 on Build GRCh38
rs987337699 Chr.8: 88041738-88041738 on Build GRCh38
rs760938151 Chr.8: 88041789-88041789 on Build GRCh38
rs757308005 Chr.8: 88041599-88041599 on Build GRCh38

View Full Product Details