Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167917192-167917708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPC2
Entrez Gene ID
25874
Gene Name
mitochondrial pyruvate carrier 2
Gene Aliases
BRP44
UniGene
Hs.517768
Gene Chromosome Location
Chr.1: 167916675-167937069 on Build GRCh38
Transcripts
NM_001143674.3 NM_015415.3 NR_026550.2

SNP Detail

SNP ID Chromosome Location
rs967208839 Chr.1: 167917528-167917528 on Build GRCh38
rs555468967 Chr.1: 167917515-167917515 on Build GRCh38
rs1057290648 Chr.1: 167917364-167917364 on Build GRCh38
rs192370215 Chr.1: 167917456-167917456 on Build GRCh38
rs556305297 Chr.1: 167917614-167917614 on Build GRCh38
rs971140835 Chr.1: 167917425-167917425 on Build GRCh38
rs550588178 Chr.1: 167917368-167917368 on Build GRCh38
rs909199310 Chr.1: 167917578-167917578 on Build GRCh38
rs398039787 Chr.1: 167917618-167917619 on Build GRCh38
rs1014185239 Chr.1: 167917400-167917400 on Build GRCh38
rs932060690 Chr.1: 167917564-167917564 on Build GRCh38
rs186665961 Chr.1: 167917559-167917559 on Build GRCh38
rs528060819 Chr.1: 167917630-167917630 on Build GRCh38
rs528912207 Chr.1: 167917335-167917335 on Build GRCh38
rs112396873 Chr.1: 167917490-167917490 on Build GRCh38
rs2420 Chr.1: 167917264-167917264 on Build GRCh38
rs1019623436 Chr.1: 167917492-167917492 on Build GRCh38
rs990064599 Chr.1: 167917491-167917491 on Build GRCh38
rs923370443 Chr.1: 167917239-167917239 on Build GRCh38
rs140354583 Chr.1: 167917369-167917369 on Build GRCh38
rs1021444637 Chr.1: 167917383-167917383 on Build GRCh38
rs185009049 Chr.1: 167917483-167917483 on Build GRCh38
rs36120795 Chr.1: 167917597-167917598 on Build GRCh38
rs561666558 Chr.1: 167917314-167917314 on Build GRCh38
rs1028881632 Chr.1: 167917479-167917479 on Build GRCh38
rs868839408 Chr.1: 167917421-167917421 on Build GRCh38
rs1024039286 Chr.1: 167917419-167917419 on Build GRCh38
rs927532645 Chr.1: 167917633-167917633 on Build GRCh38
rs140887485 Chr.1: 167917546-167917546 on Build GRCh38
rs528155067 Chr.1: 167917225-167917225 on Build GRCh38
rs1050510918 Chr.1: 167917577-167917577 on Build GRCh38
rs942057383 Chr.1: 167917589-167917589 on Build GRCh38
rs887813156 Chr.1: 167917327-167917327 on Build GRCh38
rs1036588576 Chr.1: 167917597-167917597 on Build GRCh38
rs144107237 Chr.1: 167917396-167917396 on Build GRCh38
rs938932194 Chr.1: 167917667-167917667 on Build GRCh38
rs867718274 Chr.1: 167917623-167917623 on Build GRCh38
rs200797846 Chr.1: 167917395-167917395 on Build GRCh38
rs71301003 Chr.1: 167917598-167917599 on Build GRCh38
rs769883174 Chr.1: 167917316-167917316 on Build GRCh38
rs1001164709 Chr.1: 167917440-167917440 on Build GRCh38
rs996515632 Chr.1: 167917397-167917397 on Build GRCh38

View Full Product Details