Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169921251-169921760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIFAP3
Entrez Gene ID
22920
Gene Name
kinesin associated protein 3
Gene Aliases
FLA3, KAP-1, KAP-3, KAP3, SMAP, Smg-GDS, dJ190I16.1
UniGene
Hs.433442
Gene Chromosome Location
Chr.1: 169921329-170074741 on Build GRCh38
Transcripts
NM_001204514.1 NM_001204516.1 NM_001204517.1 NM_014970.3 XM_005244970.1 XM_011509307.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3376024 c.2116G>T p.E706* Chr.1: 169921705-169921705 on Build GRCh38
6695734 c.2342G>A p.R781H Chr.1: 169921713-169921713 on Build GRCh38
4961200 c.2081G>A p.G694E Chr.1: 169921740-169921740 on Build GRCh38
4961199 c.2315G>A p.G772E Chr.1: 169921740-169921740 on Build GRCh38
6695735 c.2108G>A p.R703H Chr.1: 169921713-169921713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778694011 Chr.1: 169921656-169921656 on Build GRCh38
rs368164572 Chr.1: 169921733-169921733 on Build GRCh38
rs771132189 Chr.1: 169921671-169921671 on Build GRCh38
rs964169632 Chr.1: 169921404-169921404 on Build GRCh38
rs775816268 Chr.1: 169921695-169921695 on Build GRCh38
rs780379824 Chr.1: 169921654-169921655 on Build GRCh38
rs1030007475 Chr.1: 169921329-169921329 on Build GRCh38
rs559856517 Chr.1: 169921442-169921442 on Build GRCh38
rs7674 Chr.1: 169921482-169921482 on Build GRCh38
rs373947122 Chr.1: 169921662-169921662 on Build GRCh38
rs200602323 Chr.1: 169921726-169921726 on Build GRCh38
rs753412995 Chr.1: 169921655-169921656 on Build GRCh38
rs898197417 Chr.1: 169921316-169921316 on Build GRCh38
rs753569409 Chr.1: 169921635-169921635 on Build GRCh38
rs549698923 Chr.1: 169921476-169921476 on Build GRCh38
rs769209419 Chr.1: 169921551-169921551 on Build GRCh38
rs891030787 Chr.1: 169921357-169921361 on Build GRCh38
rs1006704553 Chr.1: 169921345-169921345 on Build GRCh38
rs747178735 Chr.1: 169921668-169921668 on Build GRCh38
rs1050767561 Chr.1: 169921706-169921706 on Build GRCh38
rs776908705 Chr.1: 169921686-169921686 on Build GRCh38
rs916746916 Chr.1: 169921557-169921557 on Build GRCh38
rs754842097 Chr.1: 169921645-169921645 on Build GRCh38
rs928224739 Chr.1: 169921495-169921495 on Build GRCh38
rs554145598 Chr.1: 169921703-169921703 on Build GRCh38
rs4519 Chr.1: 169921433-169921433 on Build GRCh38
rs184581509 Chr.1: 169921738-169921738 on Build GRCh38
rs760327497 Chr.1: 169921713-169921713 on Build GRCh38
rs935011071 Chr.1: 169921690-169921690 on Build GRCh38
rs747696606 Chr.1: 169921655-169921655 on Build GRCh38
rs916022917 Chr.1: 169921641-169921645 on Build GRCh38
rs1012373360 Chr.1: 169921281-169921281 on Build GRCh38
rs902181498 Chr.1: 169921287-169921287 on Build GRCh38
rs766097860 Chr.1: 169921714-169921714 on Build GRCh38
rs567896336 Chr.1: 169921480-169921480 on Build GRCh38
rs114938904 Chr.1: 169921546-169921546 on Build GRCh38
rs1018051747 Chr.1: 169921378-169921378 on Build GRCh38
rs199699281 Chr.1: 169921737-169921737 on Build GRCh38
rs770213276 Chr.1: 169921694-169921694 on Build GRCh38
rs753576167 Chr.1: 169921722-169921722 on Build GRCh38
rs964801815 Chr.1: 169921383-169921383 on Build GRCh38
rs532860667 Chr.1: 169921354-169921354 on Build GRCh38
rs57164758 Chr.1: 169921696-169921696 on Build GRCh38
rs3841695 Chr.1: 169921315-169921316 on Build GRCh38
rs1027704509 Chr.1: 169921441-169921441 on Build GRCh38
rs192958712 Chr.1: 169921519-169921519 on Build GRCh38
rs991677854 Chr.1: 169921571-169921571 on Build GRCh38
rs946828589 Chr.1: 169921601-169921601 on Build GRCh38
rs772832933 Chr.1: 169921712-169921712 on Build GRCh38
rs1038563857 Chr.1: 169921678-169921678 on Build GRCh38
rs550323977 Chr.1: 169921514-169921514 on Build GRCh38
rs767075823 Chr.1: 169921710-169921710 on Build GRCh38
rs746192151 Chr.1: 169921689-169921689 on Build GRCh38
rs967380277 Chr.1: 169921451-169921451 on Build GRCh38
rs144656623 Chr.1: 169921458-169921458 on Build GRCh38
rs990679035 Chr.1: 169921410-169921410 on Build GRCh38
rs777836246 Chr.1: 169921663-169921663 on Build GRCh38
rs35534461 Chr.1: 169921666-169921667 on Build GRCh38
rs999165886 Chr.1: 169921289-169921289 on Build GRCh38
rs891021146 Chr.1: 169921344-169921344 on Build GRCh38

View Full Product Details