Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 170539068-170539614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GORAB
Entrez Gene ID
92344
Gene Name
golgin, RAB6 interacting
Gene Aliases
GO, NTKLBP1, SCYL1BP1
UniGene
Hs.183702
Gene Chromosome Location
Chr.1: 170532122-170553834 on Build GRCh38
Transcripts
NM_001146039.1 NM_001320252.1 NM_152281.2 NR_027397.1 XM_006711628.3 XM_011510149.2 XM_011510150.2 XM_011510151.1 XM_017002807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2074138 c.166G>A p.A56T Chr.1: 170539239-170539239 on Build GRCh38
5727936 c.238G>A p.D80N Chr.1: 170539311-170539311 on Build GRCh38
2074139 c.184C>T p.Q62* Chr.1: 170539257-170539257 on Build GRCh38
7369781 c.167C>T p.A56V Chr.1: 170539240-170539240 on Build GRCh38
2074145 c.372A>C p.Q124H Chr.1: 170539445-170539445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780894393 Chr.1: 170539481-170539481 on Build GRCh38
rs958641328 Chr.1: 170539098-170539098 on Build GRCh38
rs377643172 Chr.1: 170539370-170539370 on Build GRCh38
rs774304692 Chr.1: 170539516-170539516 on Build GRCh38
rs147859830 Chr.1: 170539432-170539432 on Build GRCh38
rs119455951 Chr.1: 170539515-170539515 on Build GRCh38
rs138420081 Chr.1: 170539240-170539240 on Build GRCh38
rs200082761 Chr.1: 170539385-170539385 on Build GRCh38
rs374614330 Chr.1: 170539405-170539405 on Build GRCh38
rs200333798 Chr.1: 170539336-170539336 on Build GRCh38
rs751708902 Chr.1: 170539371-170539371 on Build GRCh38
rs760397381 Chr.1: 170539327-170539327 on Build GRCh38
rs745488711 Chr.1: 170539487-170539487 on Build GRCh38
rs759352018 Chr.1: 170539422-170539422 on Build GRCh38
rs766190900 Chr.1: 170539334-170539334 on Build GRCh38
rs369967804 Chr.1: 170539266-170539266 on Build GRCh38
rs377442498 Chr.1: 170539406-170539406 on Build GRCh38
rs902247084 Chr.1: 170539146-170539146 on Build GRCh38
rs372377857 Chr.1: 170539210-170539210 on Build GRCh38
rs754286398 Chr.1: 170539436-170539436 on Build GRCh38
rs755379894 Chr.1: 170539447-170539447 on Build GRCh38
rs764152549 Chr.1: 170539428-170539428 on Build GRCh38
rs991261881 Chr.1: 170539140-170539140 on Build GRCh38
rs769560414 Chr.1: 170539490-170539490 on Build GRCh38
rs770583915 Chr.1: 170539477-170539477 on Build GRCh38
rs772817185 Chr.1: 170539315-170539315 on Build GRCh38
rs534302027 Chr.1: 170539303-170539303 on Build GRCh38
rs761888047 Chr.1: 170539429-170539429 on Build GRCh38
rs914443995 Chr.1: 170539142-170539142 on Build GRCh38
rs561578722 Chr.1: 170539194-170539194 on Build GRCh38
rs537712038 Chr.1: 170539096-170539096 on Build GRCh38
rs765177742 Chr.1: 170539340-170539340 on Build GRCh38
rs777858963 Chr.1: 170539187-170539187 on Build GRCh38
rs373666279 Chr.1: 170539427-170539427 on Build GRCh38
rs545064167 Chr.1: 170539526-170539526 on Build GRCh38
rs772275964 Chr.1: 170539178-170539178 on Build GRCh38
rs370933035 Chr.1: 170539387-170539387 on Build GRCh38
rs916210169 Chr.1: 170539421-170539421 on Build GRCh38
rs756398250 Chr.1: 170539431-170539431 on Build GRCh38
rs767563774 Chr.1: 170539529-170539529 on Build GRCh38
rs775398638 Chr.1: 170539426-170539426 on Build GRCh38
rs946528163 Chr.1: 170539122-170539122 on Build GRCh38
rs757514096 Chr.1: 170539260-170539260 on Build GRCh38
rs781408413 Chr.1: 170539382-170539382 on Build GRCh38
rs920179398 Chr.1: 170539577-170539577 on Build GRCh38
rs868380549 Chr.1: 170539525-170539525 on Build GRCh38
rs779129692 Chr.1: 170539171-170539171 on Build GRCh38
rs150870759 Chr.1: 170539333-170539333 on Build GRCh38
rs370271267 Chr.1: 170539424-170539424 on Build GRCh38
rs770637285 Chr.1: 170539407-170539407 on Build GRCh38
rs746474929 Chr.1: 170539472-170539472 on Build GRCh38
rs573300781 Chr.1: 170539276-170539276 on Build GRCh38
rs746165481 Chr.1: 170539383-170539383 on Build GRCh38
rs770230615 Chr.1: 170539286-170539286 on Build GRCh38
rs891309886 Chr.1: 170539482-170539482 on Build GRCh38
rs762156742 Chr.1: 170539548-170539548 on Build GRCh38
rs73029138 Chr.1: 170539207-170539207 on Build GRCh38
rs779477245 Chr.1: 170539457-170539457 on Build GRCh38
rs1047104786 Chr.1: 170539317-170539317 on Build GRCh38
rs765276217 Chr.1: 170539241-170539241 on Build GRCh38
rs1036122529 Chr.1: 170539349-170539349 on Build GRCh38
rs775867092 Chr.1: 170539290-170539290 on Build GRCh38
rs375138675 Chr.1: 170539252-170539252 on Build GRCh38
rs894822983 Chr.1: 170539206-170539206 on Build GRCh38
rs778007699 Chr.1: 170539255-170539255 on Build GRCh38
rs770296573 Chr.1: 170539127-170539127 on Build GRCh38
rs146212101 Chr.1: 170539375-170539375 on Build GRCh38
rs762822921 Chr.1: 170539499-170539499 on Build GRCh38
rs766634981 Chr.1: 170539580-170539580 on Build GRCh38
rs977675431 Chr.1: 170539419-170539419 on Build GRCh38
rs149866830 Chr.1: 170539399-170539399 on Build GRCh38
rs772771053 Chr.1: 170539236-170539236 on Build GRCh38
rs939138134 Chr.1: 170539158-170539158 on Build GRCh38
rs747376663 Chr.1: 170539404-170539404 on Build GRCh38
rs752751787 Chr.1: 170539251-170539251 on Build GRCh38
rs754082743 Chr.1: 170539582-170539582 on Build GRCh38
rs780531933 Chr.1: 170539360-170539361 on Build GRCh38
rs963729755 Chr.1: 170539523-170539523 on Build GRCh38
rs766244250 Chr.1: 170539238-170539238 on Build GRCh38
rs148682649 Chr.1: 170539420-170539420 on Build GRCh38
rs1051880720 Chr.1: 170539211-170539211 on Build GRCh38
rs768414016 Chr.1: 170539509-170539509 on Build GRCh38
rs780562193 Chr.1: 170539398-170539398 on Build GRCh38
rs750567904 Chr.1: 170539536-170539536 on Build GRCh38
rs769272155 Chr.1: 170539403-170539403 on Build GRCh38
rs146131654 Chr.1: 170539239-170539239 on Build GRCh38
rs139675053 Chr.1: 170539356-170539356 on Build GRCh38
rs144383104 Chr.1: 170539231-170539231 on Build GRCh38
rs190850168 Chr.1: 170539304-170539304 on Build GRCh38
rs577697246 Chr.1: 170539496-170539496 on Build GRCh38
rs143258916 Chr.1: 170539293-170539293 on Build GRCh38
rs964155812 Chr.1: 170539556-170539556 on Build GRCh38
rs747179838 Chr.1: 170539256-170539256 on Build GRCh38
rs767686838 Chr.1: 170539430-170539430 on Build GRCh38
rs756799492 Chr.1: 170539493-170539493 on Build GRCh38
rs758414105 Chr.1: 170539369-170539369 on Build GRCh38
rs371601165 Chr.1: 170539584-170539584 on Build GRCh38
rs770355472 Chr.1: 170539227-170539227 on Build GRCh38
rs781535204 Chr.1: 170539261-170539261 on Build GRCh38
rs761243601 Chr.1: 170539230-170539230 on Build GRCh38
rs760452446 Chr.1: 170539237-170539237 on Build GRCh38
rs374051444 Chr.1: 170539217-170539217 on Build GRCh38
rs753804167 Chr.1: 170539168-170539168 on Build GRCh38
rs755004175 Chr.1: 170539170-170539170 on Build GRCh38
rs201116057 Chr.1: 170539172-170539172 on Build GRCh38
rs1047652359 Chr.1: 170539440-170539440 on Build GRCh38
rs915036837 Chr.1: 170539110-170539110 on Build GRCh38
rs1042276551 Chr.1: 170539139-170539139 on Build GRCh38

View Full Product Details