Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100573408-100573904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX31
Entrez Gene ID
169166
Gene Name
sorting nexin 31
Gene Aliases
-
UniGene
Hs.567758
Gene Chromosome Location
Chr.8: 100572882-100663410 on Build GRCh38
Transcripts
NM_152628.3 XM_005250815.3 XM_011516899.1 XM_017013153.1 XM_017013154.1 XM_017013155.1 XM_017013156.1 XM_017013157.1 XM_017013158.1 XM_017013159.1 XM_017013160.1 XM_017013161.1 XM_017013162.1 XM_017013163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3778784 c.1312G>A p.E438K Chr.8: 100573876-100573876 on Build GRCh38
1453645 c.1296delT p.F432fs*4 Chr.8: 100573892-100573892 on Build GRCh38
1579139 c.1289G>T p.C430F Chr.8: 100573899-100573899 on Build GRCh38
4877594 c.1290C>T p.C430C Chr.8: 100573898-100573898 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs891690876 Chr.8: 100573745-100573745 on Build GRCh38
rs1010583023 Chr.8: 100573771-100573771 on Build GRCh38
rs891505254 Chr.8: 100573736-100573736 on Build GRCh38
rs779507196 Chr.8: 100573865-100573865 on Build GRCh38
rs927121113 Chr.8: 100573648-100573648 on Build GRCh38
rs955746171 Chr.8: 100573505-100573505 on Build GRCh38
rs772127873 Chr.8: 100573436-100573436 on Build GRCh38
rs187251977 Chr.8: 100573445-100573445 on Build GRCh38
rs747205140 Chr.8: 100573613-100573613 on Build GRCh38
rs911841899 Chr.8: 100573667-100573668 on Build GRCh38
rs535086731 Chr.8: 100573465-100573465 on Build GRCh38
rs919928413 Chr.8: 100573623-100573623 on Build GRCh38
rs1028457305 Chr.8: 100573495-100573495 on Build GRCh38
rs372142006 Chr.8: 100573842-100573842 on Build GRCh38
rs764029476 Chr.8: 100573826-100573826 on Build GRCh38
rs1034987643 Chr.8: 100573766-100573766 on Build GRCh38
rs1038866170 Chr.8: 100573672-100573672 on Build GRCh38
rs77900518 Chr.8: 100573678-100573678 on Build GRCh38
rs757968591 Chr.8: 100573857-100573857 on Build GRCh38
rs999929025 Chr.8: 100573454-100573454 on Build GRCh38
rs904190703 Chr.8: 100573808-100573808 on Build GRCh38
rs576354840 Chr.8: 100573688-100573688 on Build GRCh38
rs574034373 Chr.8: 100573754-100573754 on Build GRCh38
rs962931485 Chr.8: 100573774-100573774 on Build GRCh38
rs1044914293 Chr.8: 100573631-100573631 on Build GRCh38
rs1028782269 Chr.8: 100573820-100573820 on Build GRCh38
rs995938892 Chr.8: 100573811-100573811 on Build GRCh38
rs761541878 Chr.8: 100573624-100573624 on Build GRCh38
rs907477526 Chr.8: 100573745-100573746 on Build GRCh38
rs143275419 Chr.8: 100573515-100573515 on Build GRCh38
rs549668432 Chr.8: 100573703-100573704 on Build GRCh38
rs935803924 Chr.8: 100573706-100573706 on Build GRCh38
rs773805331 Chr.8: 100573880-100573883 on Build GRCh38
rs1054229799 Chr.8: 100573711-100573711 on Build GRCh38
rs986727521 Chr.8: 100573664-100573664 on Build GRCh38
rs776433966 Chr.8: 100573621-100573621 on Build GRCh38
rs1024570841 Chr.8: 100573558-100573558 on Build GRCh38
rs763040079 Chr.8: 100573742-100573742 on Build GRCh38
rs368683915 Chr.8: 100573835-100573835 on Build GRCh38
rs748703713 Chr.8: 100573866-100573866 on Build GRCh38
rs565654018 Chr.8: 100573556-100573556 on Build GRCh38
rs75299360 Chr.8: 100573668-100573668 on Build GRCh38
rs971633431 Chr.8: 100573611-100573611 on Build GRCh38
rs555711742 Chr.8: 100573743-100573743 on Build GRCh38
rs555880180 Chr.8: 100573777-100573777 on Build GRCh38
rs542733084 Chr.8: 100573617-100573617 on Build GRCh38
rs929994289 Chr.8: 100573649-100573649 on Build GRCh38
rs753527944 Chr.8: 100573828-100573828 on Build GRCh38
rs6997235 Chr.8: 100573588-100573588 on Build GRCh38
rs549312186 Chr.8: 100573437-100573437 on Build GRCh38
rs912849307 Chr.8: 100573614-100573614 on Build GRCh38
rs765152923 Chr.8: 100573628-100573628 on Build GRCh38
rs543732726 Chr.8: 100573718-100573718 on Build GRCh38
rs745518848 Chr.8: 100573850-100573850 on Build GRCh38
rs944621487 Chr.8: 100573668-100573668 on Build GRCh38
rs767875315 Chr.8: 100573872-100573874 on Build GRCh38

View Full Product Details