Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100919156-100919661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAZ
Entrez Gene ID
7534
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta
Gene Aliases
14-3-3-zeta, HEL-S-3, HEL-S-93, HEL4, KCIP-1, YWHAD
UniGene
Hs.492407
Gene Chromosome Location
Chr.8: 100918576-100954068 on Build GRCh38
Transcripts
NM_001135699.1 NM_001135700.1 NM_001135701.1 NM_001135702.1 NM_003406.3 NM_145690.2 XM_005251061.3 XM_005251063.3 XM_017013810.1 XM_017013811.1

SNP Detail

SNP ID Chromosome Location
rs779319598 Chr.8: 100919374-100919374 on Build GRCh38
rs1044368704 Chr.8: 100919502-100919502 on Build GRCh38
rs992064921 Chr.8: 100919474-100919474 on Build GRCh38
rs140382471 Chr.8: 100919530-100919530 on Build GRCh38
rs1005710212 Chr.8: 100919238-100919238 on Build GRCh38
rs941409449 Chr.8: 100919535-100919535 on Build GRCh38
rs564686645 Chr.8: 100919203-100919203 on Build GRCh38
rs770949836 Chr.8: 100919457-100919458 on Build GRCh38
rs56284211 Chr.8: 100919373-100919373 on Build GRCh38
rs115295165 Chr.8: 100919332-100919332 on Build GRCh38
rs562576549 Chr.8: 100919447-100919447 on Build GRCh38
rs923733216 Chr.8: 100919450-100919450 on Build GRCh38
rs1017056646 Chr.8: 100919252-100919252 on Build GRCh38
rs183506990 Chr.8: 100919561-100919561 on Build GRCh38
rs985513748 Chr.8: 100919426-100919426 on Build GRCh38
rs187173723 Chr.8: 100919309-100919309 on Build GRCh38
rs754929245 Chr.8: 100919464-100919464 on Build GRCh38
rs749382387 Chr.8: 100919421-100919421 on Build GRCh38
rs1018067657 Chr.8: 100919435-100919435 on Build GRCh38
rs552290985 Chr.8: 100919449-100919450 on Build GRCh38
rs188798657 Chr.8: 100919633-100919633 on Build GRCh38
rs548418297 Chr.8: 100919495-100919495 on Build GRCh38
rs577607650 Chr.8: 100919239-100919242 on Build GRCh38
rs897295590 Chr.8: 100919553-100919553 on Build GRCh38
rs751076085 Chr.8: 100919190-100919190 on Build GRCh38
rs11551356 Chr.8: 100919206-100919206 on Build GRCh38
rs950197339 Chr.8: 100919204-100919204 on Build GRCh38
rs917807812 Chr.8: 100919486-100919486 on Build GRCh38
rs542590378 Chr.8: 100919393-100919393 on Build GRCh38
rs756248525 Chr.8: 100919260-100919260 on Build GRCh38
rs116414929 Chr.8: 100919199-100919199 on Build GRCh38
rs765881259 Chr.8: 100919189-100919189 on Build GRCh38
rs191755037 Chr.8: 100919448-100919448 on Build GRCh38
rs1038511510 Chr.8: 100919545-100919545 on Build GRCh38
rs950107878 Chr.8: 100919487-100919487 on Build GRCh38
rs752347610 Chr.8: 100919275-100919275 on Build GRCh38

View Full Product Details