Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 102551534-102552034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ODF1
Entrez Gene ID
4956
Gene Name
outer dense fiber of sperm tails 1
Gene Aliases
CT133, HSPB10, ODF, ODF2, ODF27, ODFP, ODFPG, ODFPGA, ODFPGB, RT7, SODF
UniGene
Hs.159274
Gene Chromosome Location
Chr.8: 102551620-102561017 on Build GRCh38
Transcripts
NM_024410.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1094422 c.182G>A p.R61Q Chr.8: 102551909-102551909 on Build GRCh38
5613004 c.39G>C p.R13S Chr.8: 102551766-102551766 on Build GRCh38
273034 c.112G>A p.D38N Chr.8: 102551839-102551839 on Build GRCh38
75691 c.14G>C p.S5T Chr.8: 102551741-102551741 on Build GRCh38
5457365 c.87C>T p.D29D Chr.8: 102551814-102551814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148423452 Chr.8: 102551886-102551886 on Build GRCh38
rs1055902067 Chr.8: 102551620-102551620 on Build GRCh38
rs752251572 Chr.8: 102551731-102551731 on Build GRCh38
rs146870404 Chr.8: 102551993-102551993 on Build GRCh38
rs3808357 Chr.8: 102551679-102551679 on Build GRCh38
rs760076149 Chr.8: 102551963-102551963 on Build GRCh38
rs200964058 Chr.8: 102551834-102551834 on Build GRCh38
rs573895260 Chr.8: 102551771-102551771 on Build GRCh38
rs917622336 Chr.8: 102551874-102551874 on Build GRCh38
rs150929068 Chr.8: 102551922-102551922 on Build GRCh38
rs373220032 Chr.8: 102551750-102551750 on Build GRCh38
rs144503717 Chr.8: 102551838-102551838 on Build GRCh38
rs752938995 Chr.8: 102551975-102551975 on Build GRCh38
rs781658558 Chr.8: 102551856-102551856 on Build GRCh38
rs549572663 Chr.8: 102551848-102551848 on Build GRCh38
rs780468049 Chr.8: 102551773-102551773 on Build GRCh38
rs989769146 Chr.8: 102551824-102551824 on Build GRCh38
rs142535568 Chr.8: 102551909-102551909 on Build GRCh38
rs756233236 Chr.8: 102551862-102551862 on Build GRCh38
rs1018012859 Chr.8: 102551790-102551790 on Build GRCh38
rs780785186 Chr.8: 102551918-102551918 on Build GRCh38
rs748427672 Chr.8: 102551859-102551859 on Build GRCh38
rs938734369 Chr.8: 102551610-102551610 on Build GRCh38
rs878878717 Chr.8: 102551850-102551850 on Build GRCh38
rs555242050 Chr.8: 102551757-102551757 on Build GRCh38
rs770707514 Chr.8: 102551815-102551815 on Build GRCh38
rs139792741 Chr.8: 102551970-102551970 on Build GRCh38
rs143906399 Chr.8: 102551978-102551978 on Build GRCh38
rs777846782 Chr.8: 102551985-102551985 on Build GRCh38
rs575317947 Chr.8: 102551685-102551685 on Build GRCh38
rs749325863 Chr.8: 102551877-102551877 on Build GRCh38
rs963799490 Chr.8: 102551792-102551792 on Build GRCh38
rs760197119 Chr.8: 102551829-102551829 on Build GRCh38
rs774200345 Chr.8: 102551823-102551823 on Build GRCh38
rs767036956 Chr.8: 102551937-102551937 on Build GRCh38
rs990502083 Chr.8: 102551666-102551666 on Build GRCh38
rs777107469 Chr.8: 102551752-102551752 on Build GRCh38
rs150498672 Chr.8: 102551678-102551678 on Build GRCh38
rs941997256 Chr.8: 102551646-102551646 on Build GRCh38
rs141788431 Chr.8: 102551575-102551575 on Build GRCh38
rs763645116 Chr.8: 102551923-102551923 on Build GRCh38
rs369351331 Chr.8: 102551814-102551814 on Build GRCh38
rs374235407 Chr.8: 102551958-102551958 on Build GRCh38
rs201140535 Chr.8: 102551826-102551826 on Build GRCh38
rs367874547 Chr.8: 102551884-102551884 on Build GRCh38
rs751297816 Chr.8: 102551694-102551694 on Build GRCh38
rs761202706 Chr.8: 102551935-102551935 on Build GRCh38
rs201634418 Chr.8: 102551915-102551915 on Build GRCh38
rs776260918 Chr.8: 102551925-102551925 on Build GRCh38
rs941838249 Chr.8: 102551713-102551713 on Build GRCh38
rs941754461 Chr.8: 102551950-102551950 on Build GRCh38
rs375133237 Chr.8: 102551782-102551782 on Build GRCh38
rs1037609003 Chr.8: 102551720-102551720 on Build GRCh38
rs938843387 Chr.8: 102551837-102551837 on Build GRCh38
rs781546214 Chr.8: 102551807-102551807 on Build GRCh38
rs931908517 Chr.8: 102551971-102551971 on Build GRCh38
rs577757103 Chr.8: 102551827-102551827 on Build GRCh38
rs530595332 Chr.8: 102551846-102551846 on Build GRCh38
rs774239198 Chr.8: 102551900-102551900 on Build GRCh38
rs769951903 Chr.8: 102551812-102551812 on Build GRCh38
rs894574542 Chr.8: 102551625-102551625 on Build GRCh38
rs140507903 Chr.8: 102551778-102551778 on Build GRCh38
rs777913768 Chr.8: 102551873-102551873 on Build GRCh38
rs756426479 Chr.8: 102551976-102551976 on Build GRCh38
rs759006752 Chr.8: 102551839-102551839 on Build GRCh38
rs761973754 Chr.8: 102551677-102551677 on Build GRCh38
rs117958007 Chr.8: 102551828-102551828 on Build GRCh38
rs113537224 Chr.8: 102551723-102551723 on Build GRCh38
rs759194568 Chr.8: 102551717-102551717 on Build GRCh38
rs142790437 Chr.8: 102551811-102551811 on Build GRCh38
rs755306472 Chr.8: 102551801-102551801 on Build GRCh38
rs771876215 Chr.8: 102551914-102551914 on Build GRCh38
rs937690602 Chr.8: 102551623-102551623 on Build GRCh38
rs766977668 Chr.8: 102551842-102551842 on Build GRCh38

View Full Product Details