Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 112223944-112224482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD3
Entrez Gene ID
114788
Gene Name
CUB and Sushi multiple domains 3
Gene Aliases
-
UniGene
Hs.91381
Gene Chromosome Location
Chr.8: 112222928-113437499 on Build GRCh38
Transcripts
NM_052900.2 NM_198123.1 NM_198124.1 XM_011516815.2 XM_011516816.2 XM_017013008.1 XM_017013009.1 XM_017013010.1 XM_017013011.1 XM_017013012.1

SNP Detail

SNP ID Chromosome Location
rs1001355314 Chr.8: 112224212-112224212 on Build GRCh38
rs898162039 Chr.8: 112224066-112224066 on Build GRCh38
rs759631973 Chr.8: 112224389-112224389 on Build GRCh38
rs187117037 Chr.8: 112224046-112224046 on Build GRCh38
rs956887198 Chr.8: 112224331-112224331 on Build GRCh38
rs1008015700 Chr.8: 112224432-112224432 on Build GRCh38
rs755796727 Chr.8: 112223968-112223968 on Build GRCh38
rs1037962862 Chr.8: 112224351-112224351 on Build GRCh38
rs779929802 Chr.8: 112223990-112223990 on Build GRCh38
rs968758172 Chr.8: 112224264-112224264 on Build GRCh38
rs920089575 Chr.8: 112224415-112224415 on Build GRCh38
rs761138653 Chr.8: 112224156-112224156 on Build GRCh38
rs182827902 Chr.8: 112224014-112224014 on Build GRCh38
rs530601319 Chr.8: 112224013-112224013 on Build GRCh38
rs1007963282 Chr.8: 112224186-112224186 on Build GRCh38
rs550288174 Chr.8: 112224018-112224018 on Build GRCh38
rs1028086412 Chr.8: 112224161-112224161 on Build GRCh38
rs7001344 Chr.8: 112224200-112224200 on Build GRCh38
rs988767528 Chr.8: 112224251-112224251 on Build GRCh38
rs938794605 Chr.8: 112224318-112224318 on Build GRCh38
rs938868879 Chr.8: 112224015-112224015 on Build GRCh38
rs186219990 Chr.8: 112223976-112223976 on Build GRCh38
rs956770692 Chr.8: 112224012-112224012 on Build GRCh38
rs968109188 Chr.8: 112223970-112223970 on Build GRCh38
rs142558618 Chr.8: 112224409-112224409 on Build GRCh38
rs995546780 Chr.8: 112224158-112224158 on Build GRCh38
rs1004558773 Chr.8: 112224137-112224137 on Build GRCh38
rs563397723 Chr.8: 112224021-112224021 on Build GRCh38
rs988714473 Chr.8: 112223973-112223973 on Build GRCh38
rs537243951 Chr.8: 112224421-112224421 on Build GRCh38
rs770517588 Chr.8: 112224204-112224205 on Build GRCh38
rs975670708 Chr.8: 112224412-112224413 on Build GRCh38
rs766664318 Chr.8: 112224178-112224178 on Build GRCh38
rs914614054 Chr.8: 112223979-112223979 on Build GRCh38
rs867894198 Chr.8: 112224152-112224152 on Build GRCh38
rs1031088640 Chr.8: 112224262-112224262 on Build GRCh38
rs980484509 Chr.8: 112224286-112224286 on Build GRCh38
rs1021923086 Chr.8: 112224258-112224258 on Build GRCh38
rs899383058 Chr.8: 112224359-112224359 on Build GRCh38
rs886064678 Chr.8: 112224095-112224095 on Build GRCh38
rs561732118 Chr.8: 112224309-112224309 on Build GRCh38
rs148862906 Chr.8: 112224408-112224408 on Build GRCh38
rs867467501 Chr.8: 112224032-112224032 on Build GRCh38
rs979572078 Chr.8: 112223984-112223984 on Build GRCh38
rs940821913 Chr.8: 112224346-112224346 on Build GRCh38
rs572787409 Chr.8: 112224023-112224023 on Build GRCh38
rs190692163 Chr.8: 112224005-112224005 on Build GRCh38
rs992862759 Chr.8: 112224324-112224324 on Build GRCh38
rs535356840 Chr.8: 112224269-112224269 on Build GRCh38
rs930378567 Chr.8: 112224055-112224055 on Build GRCh38
rs566055851 Chr.8: 112224211-112224211 on Build GRCh38
rs907628704 Chr.8: 112224452-112224452 on Build GRCh38
rs940979381 Chr.8: 112224025-112224025 on Build GRCh38
rs955850609 Chr.8: 112224247-112224247 on Build GRCh38
rs908088027 Chr.8: 112224332-112224332 on Build GRCh38

View Full Product Details