Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 124312391-124312938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM65
Entrez Gene ID
157378
Gene Name
transmembrane protein 65
Gene Aliases
-
UniGene
Hs.187646
Gene Chromosome Location
Chr.8: 124306189-124372699 on Build GRCh38
Transcripts
NM_194291.2 XM_011516847.2 XM_017013104.1

SNP Detail

SNP ID Chromosome Location
rs1019661424 Chr.8: 124312635-124312635 on Build GRCh38
rs922071554 Chr.8: 124312438-124312438 on Build GRCh38
rs142132293 Chr.8: 124312701-124312701 on Build GRCh38
rs192060286 Chr.8: 124312419-124312419 on Build GRCh38
rs201630586 Chr.8: 124312448-124312448 on Build GRCh38
rs949294744 Chr.8: 124312851-124312851 on Build GRCh38
rs760141844 Chr.8: 124312650-124312650 on Build GRCh38
rs536756691 Chr.8: 124312604-124312604 on Build GRCh38
rs979996980 Chr.8: 124312746-124312746 on Build GRCh38
rs891400206 Chr.8: 124312671-124312671 on Build GRCh38
rs917880522 Chr.8: 124312520-124312523 on Build GRCh38
rs979463421 Chr.8: 124312461-124312461 on Build GRCh38
rs140109621 Chr.8: 124312614-124312614 on Build GRCh38
rs192579749 Chr.8: 124312815-124312815 on Build GRCh38
rs953571538 Chr.8: 124312704-124312704 on Build GRCh38
rs1044927150 Chr.8: 124312543-124312543 on Build GRCh38
rs995272426 Chr.8: 124312570-124312570 on Build GRCh38
rs777508559 Chr.8: 124312644-124312645 on Build GRCh38
rs570029361 Chr.8: 124312644-124312644 on Build GRCh38
rs925425072 Chr.8: 124312767-124312767 on Build GRCh38
rs949370974 Chr.8: 124312855-124312855 on Build GRCh38
rs961684399 Chr.8: 124312684-124312684 on Build GRCh38
rs976225505 Chr.8: 124312425-124312425 on Build GRCh38
rs990590898 Chr.8: 124312500-124312500 on Build GRCh38
rs917846003 Chr.8: 124312508-124312508 on Build GRCh38
rs1027045721 Chr.8: 124312775-124312775 on Build GRCh38
rs772505804 Chr.8: 124312682-124312682 on Build GRCh38
rs988160903 Chr.8: 124312836-124312836 on Build GRCh38
rs67451110 Chr.8: 124312545-124312550 on Build GRCh38
rs953478703 Chr.8: 124312453-124312453 on Build GRCh38
rs1008892314 Chr.8: 124312681-124312681 on Build GRCh38
rs997029026 Chr.8: 124312588-124312588 on Build GRCh38
rs900227775 Chr.8: 124312587-124312587 on Build GRCh38
rs200312672 Chr.8: 124312552-124312552 on Build GRCh38
rs936469708 Chr.8: 124312540-124312540 on Build GRCh38
rs761190614 Chr.8: 124312847-124312847 on Build GRCh38
rs1042080861 Chr.8: 124312534-124312534 on Build GRCh38
rs186947147 Chr.8: 124312585-124312585 on Build GRCh38
rs974005865 Chr.8: 124312751-124312751 on Build GRCh38
rs1044978176 Chr.8: 124312887-124312887 on Build GRCh38
rs923840677 Chr.8: 124312899-124312899 on Build GRCh38
rs925341371 Chr.8: 124312477-124312481 on Build GRCh38
rs5894703 Chr.8: 124312552-124312557 on Build GRCh38
rs990645006 Chr.8: 124312832-124312832 on Build GRCh38
rs978407793 Chr.8: 124312889-124312889 on Build GRCh38
rs182980005 Chr.8: 124312520-124312520 on Build GRCh38
rs902176705 Chr.8: 124312538-124312538 on Build GRCh38
rs774029267 Chr.8: 124312757-124312759 on Build GRCh38

View Full Product Details