Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142659890-142660387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JRK
Entrez Gene ID
8629
Gene Name
Jrk helix-turn-helix protein
Gene Aliases
JH8, jerky
UniGene
Hs.535903
Gene Chromosome Location
Chr.8: 142651501-142670132 on Build GRCh38
Transcripts
NM_001077527.2 NM_001279352.1 NM_003724.3 XM_006716677.3 XM_006716678.3 XM_011517354.1 XM_011517355.1

SNP Detail

SNP ID Chromosome Location
rs587771614 Chr.8: 142660184-142660184 on Build GRCh38
rs782227134 Chr.8: 142660179-142660179 on Build GRCh38
rs116638341 Chr.8: 142660117-142660117 on Build GRCh38
rs1053337714 Chr.8: 142660351-142660351 on Build GRCh38
rs74911193 Chr.8: 142660200-142660200 on Build GRCh38
rs904677419 Chr.8: 142660051-142660051 on Build GRCh38
rs952512329 Chr.8: 142660161-142660161 on Build GRCh38
rs979228534 Chr.8: 142660227-142660227 on Build GRCh38
rs151294612 Chr.8: 142660131-142660131 on Build GRCh38
rs587665149 Chr.8: 142659920-142659920 on Build GRCh38
rs944918265 Chr.8: 142660367-142660367 on Build GRCh38
rs934810694 Chr.8: 142660302-142660302 on Build GRCh38
rs1053284753 Chr.8: 142660078-142660078 on Build GRCh38
rs1043235197 Chr.8: 142660047-142660047 on Build GRCh38
rs183248872 Chr.8: 142660189-142660189 on Build GRCh38
rs775311482 Chr.8: 142660268-142660268 on Build GRCh38
rs4736361 Chr.8: 142659963-142659963 on Build GRCh38
rs926151006 Chr.8: 142660233-142660233 on Build GRCh38
rs781969895 Chr.8: 142660108-142660108 on Build GRCh38
rs587654347 Chr.8: 142660359-142660359 on Build GRCh38
rs192295406 Chr.8: 142659934-142659934 on Build GRCh38
rs948851029 Chr.8: 142660224-142660224 on Build GRCh38
rs999077547 Chr.8: 142660076-142660076 on Build GRCh38
rs908456725 Chr.8: 142660188-142660188 on Build GRCh38
rs587628086 Chr.8: 142660077-142660077 on Build GRCh38
rs587730660 Chr.8: 142660007-142660007 on Build GRCh38
rs1007888609 Chr.8: 142660093-142660093 on Build GRCh38
rs915990028 Chr.8: 142660012-142660012 on Build GRCh38
rs587750786 Chr.8: 142660203-142660203 on Build GRCh38
rs781822255 Chr.8: 142660036-142660036 on Build GRCh38
rs892153884 Chr.8: 142660085-142660085 on Build GRCh38
rs73714644 Chr.8: 142660119-142660119 on Build GRCh38
rs4736362 Chr.8: 142659969-142659969 on Build GRCh38
rs782342293 Chr.8: 142660044-142660044 on Build GRCh38
rs369870576 Chr.8: 142660004-142660004 on Build GRCh38
rs142421805 Chr.8: 142660129-142660129 on Build GRCh38

View Full Product Details