Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142846308-142846813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GML
Entrez Gene ID
2765
Gene Name
glycosylphosphatidylinositol anchored molecule like
Gene Aliases
LY6DL
UniGene
Hs.661218
Gene Chromosome Location
Chr.8: 142834801-142846846 on Build GRCh38
Transcripts
NM_002066.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1455281 c.372C>A p.P124P Chr.8: 142846585-142846585 on Build GRCh38
1455282 c.376G>T p.E126* Chr.8: 142846589-142846589 on Build GRCh38
4542252 c.314G>T p.C105F Chr.8: 142846527-142846527 on Build GRCh38
4546626 c.403G>A p.G135R Chr.8: 142846616-142846616 on Build GRCh38
749826 c.341G>T p.G114V Chr.8: 142846554-142846554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369120924 Chr.8: 142846733-142846733 on Build GRCh38
rs779158824 Chr.8: 142846366-142846366 on Build GRCh38
rs762611954 Chr.8: 142846652-142846655 on Build GRCh38
rs747875803 Chr.8: 142846481-142846481 on Build GRCh38
rs867707390 Chr.8: 142846478-142846478 on Build GRCh38
rs931827429 Chr.8: 142846708-142846708 on Build GRCh38
rs759090122 Chr.8: 142846493-142846493 on Build GRCh38
rs372983219 Chr.8: 142846529-142846529 on Build GRCh38
rs763374571 Chr.8: 142846553-142846553 on Build GRCh38
rs375056481 Chr.8: 142846406-142846406 on Build GRCh38
rs867879835 Chr.8: 142846758-142846758 on Build GRCh38
rs763854288 Chr.8: 142846742-142846742 on Build GRCh38
rs368774634 Chr.8: 142846644-142846644 on Build GRCh38
rs552824030 Chr.8: 142846382-142846384 on Build GRCh38
rs372538137 Chr.8: 142846532-142846532 on Build GRCh38
rs747330918 Chr.8: 142846419-142846419 on Build GRCh38
rs142273167 Chr.8: 142846502-142846502 on Build GRCh38
rs769605108 Chr.8: 142846703-142846703 on Build GRCh38
rs865957199 Chr.8: 142846757-142846757 on Build GRCh38
rs371765242 Chr.8: 142846735-142846735 on Build GRCh38
rs542278471 Chr.8: 142846445-142846445 on Build GRCh38
rs746958641 Chr.8: 142846679-142846679 on Build GRCh38
rs774772973 Chr.8: 142846562-142846562 on Build GRCh38
rs764687178 Chr.8: 142846557-142846557 on Build GRCh38
rs768006984 Chr.8: 142846729-142846729 on Build GRCh38
rs771487905 Chr.8: 142846436-142846438 on Build GRCh38
rs185910108 Chr.8: 142846724-142846724 on Build GRCh38
rs368431597 Chr.8: 142846509-142846509 on Build GRCh38
rs778134361 Chr.8: 142846393-142846393 on Build GRCh38
rs781312085 Chr.8: 142846434-142846434 on Build GRCh38
rs377485226 Chr.8: 142846374-142846374 on Build GRCh38
rs769257205 Chr.8: 142846496-142846496 on Build GRCh38
rs556795765 Chr.8: 142846338-142846338 on Build GRCh38
rs757207035 Chr.8: 142846680-142846680 on Build GRCh38
rs531308322 Chr.8: 142846512-142846512 on Build GRCh38
rs141438947 Chr.8: 142846404-142846404 on Build GRCh38
rs780863749 Chr.8: 142846681-142846681 on Build GRCh38
rs868125646 Chr.8: 142846436-142846436 on Build GRCh38
rs375010101 Chr.8: 142846345-142846345 on Build GRCh38
rs780265738 Chr.8: 142846585-142846585 on Build GRCh38
rs756321470 Chr.8: 142846453-142846453 on Build GRCh38
rs199919156 Chr.8: 142846377-142846377 on Build GRCh38
rs772854866 Chr.8: 142846754-142846754 on Build GRCh38
rs28642293 Chr.8: 142846384-142846384 on Build GRCh38
rs770646887 Chr.8: 142846381-142846381 on Build GRCh38
rs923231210 Chr.8: 142846707-142846707 on Build GRCh38
rs762352406 Chr.8: 142846574-142846574 on Build GRCh38
rs777600172 Chr.8: 142846658-142846658 on Build GRCh38
rs755194594 Chr.8: 142846739-142846739 on Build GRCh38
rs983583157 Chr.8: 142846688-142846688 on Build GRCh38
rs748950914 Chr.8: 142846716-142846716 on Build GRCh38
rs374503896 Chr.8: 142846634-142846634 on Build GRCh38
rs1015301083 Chr.8: 142846378-142846378 on Build GRCh38
rs371313765 Chr.8: 142846693-142846693 on Build GRCh38
rs969542167 Chr.8: 142846410-142846410 on Build GRCh38
rs752519280 Chr.8: 142846642-142846642 on Build GRCh38
rs753099946 Chr.8: 142846393-142846393 on Build GRCh38
rs778734434 Chr.8: 142846458-142846458 on Build GRCh38
rs538583064 Chr.8: 142846573-142846573 on Build GRCh38
rs777102614 Chr.8: 142846479-142846479 on Build GRCh38
rs868806239 Chr.8: 142846638-142846638 on Build GRCh38
rs532242687 Chr.8: 142846713-142846713 on Build GRCh38
rs772336593 Chr.8: 142846385-142846386 on Build GRCh38
rs745574768 Chr.8: 142846545-142846545 on Build GRCh38
rs750486186 Chr.8: 142846630-142846630 on Build GRCh38
rs905407772 Chr.8: 142846782-142846782 on Build GRCh38
rs981381577 Chr.8: 142846414-142846414 on Build GRCh38
rs181298814 Chr.8: 142846586-142846586 on Build GRCh38
rs200183265 Chr.8: 142846731-142846731 on Build GRCh38
rs776002476 Chr.8: 142846551-142846551 on Build GRCh38
rs769514354 Chr.8: 142846549-142846549 on Build GRCh38
rs148835436 Chr.8: 142846632-142846632 on Build GRCh38
rs775150246 Chr.8: 142846577-142846578 on Build GRCh38
rs887895624 Chr.8: 142846335-142846335 on Build GRCh38
rs150154295 Chr.8: 142846730-142846730 on Build GRCh38
rs968350566 Chr.8: 142846435-142846435 on Build GRCh38
rs763569289 Chr.8: 142846397-142846397 on Build GRCh38
rs765766032 Chr.8: 142846386-142846386 on Build GRCh38
rs527963435 Chr.8: 142846421-142846421 on Build GRCh38
rs770394915 Chr.8: 142846517-142846517 on Build GRCh38
rs541422332 Chr.8: 142846776-142846776 on Build GRCh38
rs1057204938 Chr.8: 142846783-142846783 on Build GRCh38
rs762281783 Chr.8: 142846726-142846726 on Build GRCh38
rs923030153 Chr.8: 142846588-142846588 on Build GRCh38
rs2097573 Chr.8: 142846569-142846569 on Build GRCh38
rs201791514 Chr.8: 142846736-142846736 on Build GRCh38
rs774770679 Chr.8: 142846725-142846725 on Build GRCh38
rs539785327 Chr.8: 142846407-142846407 on Build GRCh38

View Full Product Details