Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143269282-143269791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLI4
Entrez Gene ID
2738
Gene Name
GLI family zinc finger 4
Gene Aliases
HKR4, ZNF928
UniGene
Hs.400533
Gene Chromosome Location
Chr.8: 143267437-143276931 on Build GRCh38
Transcripts
NM_138465.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7427068 c.54T>G p.S18R Chr.8: 143269450-143269450 on Build GRCh38
4687169 c.65C>T p.P22L Chr.8: 143269461-143269461 on Build GRCh38
1096911 c.6A>G p.A2A Chr.8: 143269402-143269402 on Build GRCh38
1096914 c.113G>T p.G38V Chr.8: 143269509-143269509 on Build GRCh38
4923654 c.49G>T p.V17F Chr.8: 143269445-143269445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200429907 Chr.8: 143269532-143269532 on Build GRCh38
rs919698218 Chr.8: 143269306-143269306 on Build GRCh38
rs753198667 Chr.8: 143269428-143269428 on Build GRCh38
rs577456591 Chr.8: 143269627-143269627 on Build GRCh38
rs747024865 Chr.8: 143269438-143269438 on Build GRCh38
rs774195489 Chr.8: 143269408-143269408 on Build GRCh38
rs201915516 Chr.8: 143269565-143269565 on Build GRCh38
rs369475004 Chr.8: 143269384-143269384 on Build GRCh38
rs1046604014 Chr.8: 143269749-143269749 on Build GRCh38
rs915553168 Chr.8: 143269628-143269628 on Build GRCh38
rs924203902 Chr.8: 143269758-143269758 on Build GRCh38
rs1053406213 Chr.8: 143269476-143269476 on Build GRCh38
rs755750676 Chr.8: 143269353-143269353 on Build GRCh38
rs931234062 Chr.8: 143269319-143269319 on Build GRCh38
rs554258851 Chr.8: 143269586-143269586 on Build GRCh38
rs376494816 Chr.8: 143269371-143269371 on Build GRCh38
rs371542284 Chr.8: 143269362-143269362 on Build GRCh38
rs919668548 Chr.8: 143269671-143269671 on Build GRCh38
rs1053269565 Chr.8: 143269711-143269711 on Build GRCh38
rs115428991 Chr.8: 143269440-143269440 on Build GRCh38
rs895381916 Chr.8: 143269390-143269390 on Build GRCh38
rs150869412 Chr.8: 143269723-143269723 on Build GRCh38
rs750743876 Chr.8: 143269623-143269623 on Build GRCh38
rs748101436 Chr.8: 143269463-143269463 on Build GRCh38
rs1009126746 Chr.8: 143269757-143269757 on Build GRCh38
rs755262919 Chr.8: 143269420-143269420 on Build GRCh38
rs750125243 Chr.8: 143269350-143269350 on Build GRCh38
rs141216873 Chr.8: 143269542-143269542 on Build GRCh38
rs748294672 Chr.8: 143269369-143269369 on Build GRCh38
rs201963714 Chr.8: 143269536-143269536 on Build GRCh38
rs769427628 Chr.8: 143269561-143269561 on Build GRCh38
rs890727442 Chr.8: 143269744-143269744 on Build GRCh38
rs763368669 Chr.8: 143269572-143269572 on Build GRCh38
rs551875744 Chr.8: 143269634-143269634 on Build GRCh38
rs764750875 Chr.8: 143269443-143269444 on Build GRCh38
rs757453066 Chr.8: 143269521-143269521 on Build GRCh38
rs775438304 Chr.8: 143269387-143269387 on Build GRCh38
rs772707089 Chr.8: 143269412-143269412 on Build GRCh38
rs930937432 Chr.8: 143269312-143269312 on Build GRCh38
rs550786510 Chr.8: 143269366-143269366 on Build GRCh38
rs528064303 Chr.8: 143269692-143269692 on Build GRCh38
rs766995041 Chr.8: 143269410-143269410 on Build GRCh38
rs771931653 Chr.8: 143269481-143269481 on Build GRCh38
rs572013200 Chr.8: 143269437-143269437 on Build GRCh38
rs546606247 Chr.8: 143269694-143269694 on Build GRCh38
rs1039449318 Chr.8: 143269760-143269760 on Build GRCh38
rs770480409 Chr.8: 143269544-143269544 on Build GRCh38
rs760326524 Chr.8: 143269413-143269413 on Build GRCh38
rs542050652 Chr.8: 143269602-143269602 on Build GRCh38
rs772725486 Chr.8: 143269483-143269483 on Build GRCh38
rs184754009 Chr.8: 143269541-143269541 on Build GRCh38
rs925691123 Chr.8: 143269693-143269693 on Build GRCh38
rs751718721 Chr.8: 143269511-143269511 on Build GRCh38
rs1056781479 Chr.8: 143269404-143269404 on Build GRCh38
rs759111416 Chr.8: 143269560-143269560 on Build GRCh38
rs148852053 Chr.8: 143269414-143269414 on Build GRCh38
rs754138770 Chr.8: 143269360-143269360 on Build GRCh38
rs781397475 Chr.8: 143269529-143269529 on Build GRCh38
rs761788760 Chr.8: 143269409-143269409 on Build GRCh38
rs765582894 Chr.8: 143269499-143269499 on Build GRCh38
rs919747237 Chr.8: 143269311-143269311 on Build GRCh38
rs76377145 Chr.8: 143269765-143269765 on Build GRCh38
rs764644820 Chr.8: 143269509-143269509 on Build GRCh38
rs577328798 Chr.8: 143269535-143269535 on Build GRCh38
rs373384193 Chr.8: 143269538-143269538 on Build GRCh38
rs376013527 Chr.8: 143269489-143269489 on Build GRCh38
rs6558341 Chr.8: 143269462-143269462 on Build GRCh38
rs780236838 Chr.8: 143269523-143269526 on Build GRCh38
rs563707532 Chr.8: 143269689-143269689 on Build GRCh38
rs760132326 Chr.8: 143269485-143269485 on Build GRCh38
rs774876940 Chr.8: 143269544-143269544 on Build GRCh38
rs749040399 Chr.8: 143269398-143269398 on Build GRCh38
rs530477571 Chr.8: 143269606-143269606 on Build GRCh38
rs758887197 Chr.8: 143269688-143269688 on Build GRCh38
rs758839949 Chr.8: 143269507-143269507 on Build GRCh38
rs776308872 Chr.8: 143269487-143269487 on Build GRCh38
rs972598759 Chr.8: 143269633-143269633 on Build GRCh38
rs373617213 Chr.8: 143269486-143269486 on Build GRCh38
rs530689968 Chr.8: 143269355-143269355 on Build GRCh38
rs145492318 Chr.8: 143269401-143269401 on Build GRCh38
rs747298924 Chr.8: 143269376-143269376 on Build GRCh38
rs781522097 Chr.8: 143269447-143269447 on Build GRCh38
rs563916761 Chr.8: 143269605-143269605 on Build GRCh38
rs370732649 Chr.8: 143269543-143269543 on Build GRCh38
rs776197637 Chr.8: 143269546-143269546 on Build GRCh38
rs753035853 Chr.8: 143269500-143269500 on Build GRCh38
rs746072098 Chr.8: 143269453-143269453 on Build GRCh38
rs200614249 Chr.8: 143269425-143269425 on Build GRCh38
rs763399463 Chr.8: 143269419-143269420 on Build GRCh38
rs768383740 Chr.8: 143269461-143269461 on Build GRCh38

View Full Product Details