Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 25676825-25677350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUSC1
Entrez Gene ID
286319
Gene Name
tumor suppressor candidate 1
Gene Aliases
TSG-9, TSG9
UniGene
Hs.26268
Gene Chromosome Location
Chr.9: 25676389-25678858 on Build GRCh38
Transcripts
NM_001004125.2

SNP Detail

SNP ID Chromosome Location
rs756261559 Chr.9: 25677237-25677237 on Build GRCh38
rs148285595 Chr.9: 25677245-25677245 on Build GRCh38
rs35009436 Chr.9: 25677187-25677188 on Build GRCh38
rs904676426 Chr.9: 25677303-25677303 on Build GRCh38
rs769960733 Chr.9: 25677009-25677009 on Build GRCh38
rs1033461241 Chr.9: 25676972-25676972 on Build GRCh38
rs972581953 Chr.9: 25677185-25677186 on Build GRCh38
rs566848199 Chr.9: 25677301-25677301 on Build GRCh38
rs1033379478 Chr.9: 25677042-25677042 on Build GRCh38
rs1026482680 Chr.9: 25677017-25677017 on Build GRCh38
rs1024806111 Chr.9: 25676928-25676928 on Build GRCh38
rs554811472 Chr.9: 25677290-25677290 on Build GRCh38
rs114948668 Chr.9: 25677046-25677046 on Build GRCh38
rs943988957 Chr.9: 25677234-25677234 on Build GRCh38
rs759363871 Chr.9: 25676986-25676986 on Build GRCh38
rs898954971 Chr.9: 25677315-25677315 on Build GRCh38
rs74174596 Chr.9: 25677312-25677312 on Build GRCh38
rs190513567 Chr.9: 25677235-25677235 on Build GRCh38
rs530810299 Chr.9: 25676968-25676968 on Build GRCh38
rs12348 Chr.9: 25677217-25677217 on Build GRCh38
rs991856502 Chr.9: 25676855-25676856 on Build GRCh38
rs1001817360 Chr.9: 25677307-25677307 on Build GRCh38
rs549039364 Chr.9: 25676969-25676969 on Build GRCh38
rs912119701 Chr.9: 25677078-25677078 on Build GRCh38
rs879173210 Chr.9: 25677302-25677303 on Build GRCh38
rs919816379 Chr.9: 25676906-25676906 on Build GRCh38
rs373259364 Chr.9: 25676941-25676941 on Build GRCh38
rs553523693 Chr.9: 25676936-25676936 on Build GRCh38
rs7044566 Chr.9: 25676915-25676915 on Build GRCh38
rs931276321 Chr.9: 25677214-25677214 on Build GRCh38
rs1041125778 Chr.9: 25676967-25676967 on Build GRCh38
rs79831554 Chr.9: 25677313-25677313 on Build GRCh38
rs538613749 Chr.9: 25677207-25677207 on Build GRCh38
rs7028310 Chr.9: 25676957-25676957 on Build GRCh38
rs1128957 Chr.9: 25677259-25677259 on Build GRCh38
rs185786031 Chr.9: 25676879-25676879 on Build GRCh38
rs10812299 Chr.9: 25676897-25676897 on Build GRCh38
rs879176158 Chr.9: 25677301-25677302 on Build GRCh38
rs936540243 Chr.9: 25676880-25676880 on Build GRCh38
rs375220619 Chr.9: 25677302-25677302 on Build GRCh38
rs1036918579 Chr.9: 25676903-25676903 on Build GRCh38
rs1009435975 Chr.9: 25676921-25676921 on Build GRCh38
rs911241489 Chr.9: 25677223-25677223 on Build GRCh38
rs143160482 Chr.9: 25677132-25677132 on Build GRCh38
rs1002186121 Chr.9: 25677038-25677038 on Build GRCh38
rs762390443 Chr.9: 25677265-25677265 on Build GRCh38
rs889773606 Chr.9: 25676956-25676956 on Build GRCh38
rs557559665 Chr.9: 25677138-25677138 on Build GRCh38
rs78145805 Chr.9: 25677302-25677302 on Build GRCh38
rs947987868 Chr.9: 25677210-25677210 on Build GRCh38
rs987730304 Chr.9: 25677047-25677047 on Build GRCh38
rs149036155 Chr.9: 25677128-25677128 on Build GRCh38
rs936075269 Chr.9: 25677166-25677166 on Build GRCh38
rs775593285 Chr.9: 25677155-25677155 on Build GRCh38
rs963445937 Chr.9: 25676984-25676984 on Build GRCh38
rs571428995 Chr.9: 25677119-25677119 on Build GRCh38

View Full Product Details