Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113163414-113163957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC31A2
Entrez Gene ID
1318
Gene Name
solute carrier family 31 member 2
Gene Aliases
COPT2, CTR2, hCTR2
UniGene
Hs.24030
Gene Chromosome Location
Chr.9: 113150958-113164142 on Build GRCh38
Transcripts
NM_001860.2 XM_011518216.1 XM_017014267.1

SNP Detail

SNP ID Chromosome Location
rs1003377255 Chr.9: 113163877-113163877 on Build GRCh38
rs538971568 Chr.9: 113163895-113163895 on Build GRCh38
rs976249427 Chr.9: 113163824-113163824 on Build GRCh38
rs374119464 Chr.9: 113163764-113163764 on Build GRCh38
rs993822904 Chr.9: 113163921-113163921 on Build GRCh38
rs972619787 Chr.9: 113163465-113163466 on Build GRCh38
rs935354094 Chr.9: 113163663-113163663 on Build GRCh38
rs1007933175 Chr.9: 113163741-113163741 on Build GRCh38
rs73655811 Chr.9: 113163762-113163762 on Build GRCh38
rs900929591 Chr.9: 113163497-113163497 on Build GRCh38
rs755159733 Chr.9: 113163775-113163775 on Build GRCh38
rs999234875 Chr.9: 113163499-113163499 on Build GRCh38
rs1128776 Chr.9: 113163673-113163673 on Build GRCh38
rs398087524 Chr.9: 113163919-113163919 on Build GRCh38
rs372157339 Chr.9: 113163863-113163863 on Build GRCh38
rs759215522 Chr.9: 113163442-113163442 on Build GRCh38
rs989905670 Chr.9: 113163826-113163826 on Build GRCh38
rs763887008 Chr.9: 113163769-113163769 on Build GRCh38
rs11318484 Chr.9: 113163709-113163709 on Build GRCh38
rs774792184 Chr.9: 113163582-113163582 on Build GRCh38
rs905105809 Chr.9: 113163664-113163664 on Build GRCh38
rs71491074 Chr.9: 113163672-113163673 on Build GRCh38
rs896814816 Chr.9: 113163758-113163758 on Build GRCh38
rs924050425 Chr.9: 113163811-113163811 on Build GRCh38
rs1139952 Chr.9: 113163744-113163744 on Build GRCh38
rs1023997661 Chr.9: 113163628-113163628 on Build GRCh38
rs919868075 Chr.9: 113163468-113163469 on Build GRCh38
rs1052643000 Chr.9: 113163854-113163854 on Build GRCh38
rs530573480 Chr.9: 113163780-113163780 on Build GRCh38
rs542450886 Chr.9: 113163668-113163668 on Build GRCh38
rs746930860 Chr.9: 113163669-113163669 on Build GRCh38
rs536317644 Chr.9: 113163850-113163850 on Build GRCh38
rs910795068 Chr.9: 113163457-113163457 on Build GRCh38
rs1023892927 Chr.9: 113163776-113163776 on Build GRCh38
rs952753769 Chr.9: 113163768-113163768 on Build GRCh38
rs3840998 Chr.9: 113163918-113163918 on Build GRCh38
rs376164622 Chr.9: 113163837-113163837 on Build GRCh38
rs993279222 Chr.9: 113163924-113163924 on Build GRCh38
rs946412684 Chr.9: 113163869-113163869 on Build GRCh38
rs762427301 Chr.9: 113163650-113163650 on Build GRCh38
rs972098735 Chr.9: 113163743-113163743 on Build GRCh38
rs199716271 Chr.9: 113163921-113163921 on Build GRCh38
rs368706181 Chr.9: 113163825-113163825 on Build GRCh38
rs952174048 Chr.9: 113163753-113163753 on Build GRCh38
rs760713516 Chr.9: 113163730-113163730 on Build GRCh38
rs561094457 Chr.9: 113163441-113163441 on Build GRCh38
rs994263280 Chr.9: 113163761-113163761 on Build GRCh38
rs952559120 Chr.9: 113163469-113163472 on Build GRCh38
rs35913692 Chr.9: 113163668-113163669 on Build GRCh38
rs1044727171 Chr.9: 113163874-113163874 on Build GRCh38
rs530115075 Chr.9: 113163599-113163599 on Build GRCh38
rs916642859 Chr.9: 113163522-113163522 on Build GRCh38
rs1028180378 Chr.9: 113163925-113163925 on Build GRCh38
rs553624363 Chr.9: 113163862-113163862 on Build GRCh38
rs1010557448 Chr.9: 113163624-113163624 on Build GRCh38
rs200054929 Chr.9: 113163476-113163477 on Build GRCh38
rs747788471 Chr.9: 113163746-113163755 on Build GRCh38
rs567403925 Chr.9: 113163843-113163843 on Build GRCh38
rs73655810 Chr.9: 113163715-113163715 on Build GRCh38
rs1040754382 Chr.9: 113163908-113163908 on Build GRCh38
rs865859106 Chr.9: 113163885-113163885 on Build GRCh38
rs201909352 Chr.9: 113163879-113163880 on Build GRCh38
rs397821170 Chr.9: 113163920-113163920 on Build GRCh38
rs923936117 Chr.9: 113163652-113163652 on Build GRCh38
rs377362700 Chr.9: 113163669-113163670 on Build GRCh38
rs905127571 Chr.9: 113163875-113163875 on Build GRCh38
rs554111575 Chr.9: 113163529-113163529 on Build GRCh38
rs944881044 Chr.9: 113163458-113163458 on Build GRCh38
rs10603 Chr.9: 113163923-113163923 on Build GRCh38
rs12982 Chr.9: 113163718-113163718 on Build GRCh38
rs982185990 Chr.9: 113163791-113163791 on Build GRCh38
rs915466534 Chr.9: 113163835-113163835 on Build GRCh38
rs1040563784 Chr.9: 113163470-113163470 on Build GRCh38
rs935078877 Chr.9: 113163853-113163853 on Build GRCh38
rs944907868 Chr.9: 113163905-113163906 on Build GRCh38

View Full Product Details