Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 116699588-116700090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTN2
Entrez Gene ID
23245
Gene Name
astrotactin 2
Gene Aliases
bA67K19.1
UniGene
Hs.601562
Gene Chromosome Location
Chr.9: 116425225-117415039 on Build GRCh38
Transcripts
NM_014010.4 NM_001099679.1 NM_012210.3 XM_005251813.3 XM_011518398.2 XM_017014486.1

Gene Symbol
TRIM32
Entrez Gene ID
22954
Gene Name
tripartite motif containing 32
Gene Aliases
BBS11, HT2A, LGMD2H, TATIP
UniGene
Hs.591910
Gene Chromosome Location
Chr.9: 116687302-116701300 on Build GRCh38
Transcripts
NM_014010.4 NM_001099679.1 NM_012210.3 XM_005251813.3 XM_011518398.2 XM_017014486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
165130 c.1926C>T p.I642I Chr.9: 116699668-116699668 on Build GRCh38
5210384 c.1856C>T p.P619L Chr.9: 116699598-116699598 on Build GRCh38
1459631 c.1923G>C p.K641N Chr.9: 116699665-116699665 on Build GRCh38
3664043 c.1924A>G p.I642V Chr.9: 116699666-116699666 on Build GRCh38
3779589 c.1853G>A p.C618Y Chr.9: 116699595-116699595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763124613 Chr.9: 116699890-116699890 on Build GRCh38
rs955524991 Chr.9: 116699756-116699756 on Build GRCh38
rs1005610120 Chr.9: 116699611-116699611 on Build GRCh38
rs533209692 Chr.9: 116699844-116699844 on Build GRCh38
rs555611773 Chr.9: 116699899-116699899 on Build GRCh38
rs1001894063 Chr.9: 116699888-116699888 on Build GRCh38
rs539458612 Chr.9: 116699880-116699880 on Build GRCh38
rs200638331 Chr.9: 116699707-116699707 on Build GRCh38
rs532084545 Chr.9: 116699733-116699733 on Build GRCh38
rs776734216 Chr.9: 116699625-116699625 on Build GRCh38
rs752475495 Chr.9: 116699613-116699614 on Build GRCh38
rs1009722752 Chr.9: 116700030-116700030 on Build GRCh38
rs1029412675 Chr.9: 116699915-116699915 on Build GRCh38
rs752319293 Chr.9: 116699722-116699722 on Build GRCh38
rs761193342 Chr.9: 116699613-116699613 on Build GRCh38
rs549577425 Chr.9: 116699857-116699857 on Build GRCh38
rs142781513 Chr.9: 116699705-116699705 on Build GRCh38
rs941048867 Chr.9: 116699889-116699889 on Build GRCh38
rs1054772558 Chr.9: 116699892-116699892 on Build GRCh38
rs534626362 Chr.9: 116700053-116700053 on Build GRCh38
rs911872116 Chr.9: 116699816-116699816 on Build GRCh38
rs994976935 Chr.9: 116699647-116699647 on Build GRCh38
rs917114280 Chr.9: 116699865-116699865 on Build GRCh38
rs3019 Chr.9: 116699833-116699833 on Build GRCh38
rs886063381 Chr.9: 116699987-116699987 on Build GRCh38
rs567913198 Chr.9: 116699910-116699910 on Build GRCh38
rs527671692 Chr.9: 116699799-116699799 on Build GRCh38
rs748896830 Chr.9: 116699748-116699748 on Build GRCh38
rs569502547 Chr.9: 116699874-116699874 on Build GRCh38
rs373660071 Chr.9: 116699836-116699836 on Build GRCh38
rs1026437899 Chr.9: 116699955-116699955 on Build GRCh38
rs950918681 Chr.9: 116699973-116699973 on Build GRCh38
rs535411189 Chr.9: 116700013-116700014 on Build GRCh38
rs1009549456 Chr.9: 116699993-116699993 on Build GRCh38
rs750626801 Chr.9: 116699681-116699681 on Build GRCh38
rs996640260 Chr.9: 116699913-116699913 on Build GRCh38
rs542506728 Chr.9: 116699743-116699743 on Build GRCh38
rs899549661 Chr.9: 116699699-116699699 on Build GRCh38
rs763172140 Chr.9: 116699696-116699696 on Build GRCh38
rs868402796 Chr.9: 116699856-116699856 on Build GRCh38
rs769987584 Chr.9: 116699732-116699732 on Build GRCh38
rs886063380 Chr.9: 116699816-116699816 on Build GRCh38
rs765555919 Chr.9: 116699673-116699673 on Build GRCh38
rs1021226731 Chr.9: 116700031-116700031 on Build GRCh38
rs748468497 Chr.9: 116699709-116699709 on Build GRCh38
rs149956877 Chr.9: 116699881-116699881 on Build GRCh38
rs557238372 Chr.9: 116699839-116699839 on Build GRCh38
rs571526516 Chr.9: 116699788-116699788 on Build GRCh38
rs968100197 Chr.9: 116699800-116699800 on Build GRCh38
rs770577221 Chr.9: 116699749-116699749 on Build GRCh38
rs370691123 Chr.9: 116699872-116699872 on Build GRCh38
rs377368040 Chr.9: 116699868-116699868 on Build GRCh38
rs143186354 Chr.9: 116699635-116699635 on Build GRCh38
rs1046346181 Chr.9: 116699879-116699879 on Build GRCh38
rs777277975 Chr.9: 116699739-116699739 on Build GRCh38
rs974784095 Chr.9: 116700035-116700035 on Build GRCh38
rs769128859 Chr.9: 116699618-116699618 on Build GRCh38
rs761878106 Chr.9: 116699634-116699634 on Build GRCh38
rs755010346 Chr.9: 116699715-116699715 on Build GRCh38
rs770788386 Chr.9: 116699933-116699933 on Build GRCh38
rs138735929 Chr.9: 116699665-116699665 on Build GRCh38
rs781242319 Chr.9: 116699720-116699720 on Build GRCh38

View Full Product Details