Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 21657316-21657806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL34
Entrez Gene ID
257240
Gene Name
kelch like family member 34
Gene Aliases
-
UniGene
Hs.448572
Gene Chromosome Location
Chr.X: 21655349-21658387 on Build GRCh38
Transcripts
NM_153270.2

SNP Detail

SNP ID Chromosome Location
rs753996140 Chr.X: 21657518-21657518 on Build GRCh38
rs964337415 Chr.X: 21657645-21657645 on Build GRCh38
rs1019590812 Chr.X: 21657725-21657725 on Build GRCh38
rs371348001 Chr.X: 21657592-21657592 on Build GRCh38
rs865930398 Chr.X: 21657374-21657374 on Build GRCh38
rs751970705 Chr.X: 21657723-21657723 on Build GRCh38
rs772871282 Chr.X: 21657438-21657438 on Build GRCh38
rs750897735 Chr.X: 21657525-21657525 on Build GRCh38
rs756199541 Chr.X: 21657339-21657339 on Build GRCh38
rs776401139 Chr.X: 21657403-21657403 on Build GRCh38
rs746678164 Chr.X: 21657406-21657406 on Build GRCh38
rs374517026 Chr.X: 21657344-21657344 on Build GRCh38
rs754611732 Chr.X: 21657728-21657728 on Build GRCh38
rs761882723 Chr.X: 21657699-21657699 on Build GRCh38
rs764464742 Chr.X: 21657692-21657692 on Build GRCh38
rs764233521 Chr.X: 21657509-21657509 on Build GRCh38
rs1050310700 Chr.X: 21657679-21657679 on Build GRCh38
rs762874032 Chr.X: 21657446-21657446 on Build GRCh38
rs754252619 Chr.X: 21657348-21657348 on Build GRCh38
rs778834130 Chr.X: 21657751-21657751 on Build GRCh38
rs746623316 Chr.X: 21657539-21657539 on Build GRCh38
rs780545583 Chr.X: 21657556-21657556 on Build GRCh38
rs780966856 Chr.X: 21657731-21657731 on Build GRCh38
rs952808878 Chr.X: 21657772-21657772 on Build GRCh38
rs761695657 Chr.X: 21657418-21657418 on Build GRCh38
rs975354120 Chr.X: 21657664-21657664 on Build GRCh38
rs1016584711 Chr.X: 21657614-21657614 on Build GRCh38
rs761962628 Chr.X: 21657784-21657784 on Build GRCh38
rs775800069 Chr.X: 21657472-21657472 on Build GRCh38
rs1011142790 Chr.X: 21657615-21657615 on Build GRCh38
rs774322119 Chr.X: 21657657-21657657 on Build GRCh38
rs752297477 Chr.X: 21657733-21657733 on Build GRCh38
rs773138820 Chr.X: 21657565-21657565 on Build GRCh38
rs765244460 Chr.X: 21657704-21657704 on Build GRCh38
rs763594004 Chr.X: 21657399-21657399 on Build GRCh38
rs79990091 Chr.X: 21657637-21657637 on Build GRCh38
rs771035738 Chr.X: 21657749-21657749 on Build GRCh38
rs758371599 Chr.X: 21657716-21657716 on Build GRCh38
rs777172787 Chr.X: 21657744-21657744 on Build GRCh38
rs769819112 Chr.X: 21657431-21657431 on Build GRCh38
rs745719849 Chr.X: 21657753-21657753 on Build GRCh38
rs761004702 Chr.X: 21657504-21657504 on Build GRCh38
rs776567163 Chr.X: 21657695-21657695 on Build GRCh38
rs766346715 Chr.X: 21657758-21657758 on Build GRCh38
rs867906838 Chr.X: 21657413-21657413 on Build GRCh38
rs750891129 Chr.X: 21657707-21657707 on Build GRCh38
rs1024520163 Chr.X: 21657390-21657390 on Build GRCh38
rs367621290 Chr.X: 21657370-21657370 on Build GRCh38
rs749199097 Chr.X: 21657624-21657624 on Build GRCh38
rs760953354 Chr.X: 21657666-21657666 on Build GRCh38
rs780419501 Chr.X: 21657532-21657532 on Build GRCh38
rs766284824 Chr.X: 21657449-21657449 on Build GRCh38
rs770804766 Chr.X: 21657629-21657629 on Build GRCh38
rs747754470 Chr.X: 21657561-21657561 on Build GRCh38
rs766697400 Chr.X: 21657717-21657717 on Build GRCh38
rs1025723195 Chr.X: 21657767-21657767 on Build GRCh38
rs750266650 Chr.X: 21657562-21657562 on Build GRCh38
rs746297690 Chr.X: 21657397-21657397 on Build GRCh38
rs776851093 Chr.X: 21657773-21657773 on Build GRCh38
rs750879357 Chr.X: 21657575-21657575 on Build GRCh38
rs768059361 Chr.X: 21657343-21657349 on Build GRCh38
rs755595762 Chr.X: 21657738-21657738 on Build GRCh38
rs754666766 Chr.X: 21657555-21657555 on Build GRCh38
rs147510822 Chr.X: 21657519-21657519 on Build GRCh38
rs148155634 Chr.X: 21657526-21657526 on Build GRCh38
rs927641108 Chr.X: 21657430-21657430 on Build GRCh38
rs768104028 Chr.X: 21657412-21657412 on Build GRCh38
rs984336600 Chr.X: 21657343-21657343 on Build GRCh38
rs757820494 Chr.X: 21657350-21657350 on Build GRCh38
rs1003304266 Chr.X: 21657477-21657477 on Build GRCh38

View Full Product Details