Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 30665419-30665918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GK
Entrez Gene ID
2710
Gene Name
glycerol kinase
Gene Aliases
GK1, GKD
UniGene
Hs.1466
Gene Chromosome Location
Chr.X: 30653348-30731462 on Build GRCh38
Transcripts
NM_000167.5 NM_001128127.2 NM_001205019.1 NM_203391.3 XM_005274488.4 XM_006724483.2 XM_006724484.2 XM_006724485.2 XM_006724486.3 XM_011545491.2 XM_011545492.1 XM_011545494.2 XM_017029409.1 XM_017029410.1 XM_017029411.1 XM_017029412.1

SNP Detail

SNP ID Chromosome Location
rs917642534 Chr.X: 30665655-30665655 on Build GRCh38
rs889750596 Chr.X: 30665816-30665816 on Build GRCh38
rs1000788569 Chr.X: 30665791-30665791 on Build GRCh38
rs772232381 Chr.X: 30665605-30665605 on Build GRCh38
rs12011728 Chr.X: 30665833-30665833 on Build GRCh38
rs1045142795 Chr.X: 30665735-30665735 on Build GRCh38
rs775803183 Chr.X: 30665609-30665609 on Build GRCh38
rs769703845 Chr.X: 30665567-30665567 on Build GRCh38
rs187813593 Chr.X: 30665620-30665620 on Build GRCh38
rs942705534 Chr.X: 30665826-30665826 on Build GRCh38
rs778339477 Chr.X: 30665684-30665684 on Build GRCh38
rs748419072 Chr.X: 30665635-30665635 on Build GRCh38
rs750496587 Chr.X: 30665827-30665827 on Build GRCh38
rs183749105 Chr.X: 30665871-30665871 on Build GRCh38
rs59708571 Chr.X: 30665613-30665613 on Build GRCh38
rs192627100 Chr.X: 30665681-30665681 on Build GRCh38
rs748088787 Chr.X: 30665555-30665555 on Build GRCh38
rs992442239 Chr.X: 30665508-30665508 on Build GRCh38
rs904836887 Chr.X: 30665749-30665749 on Build GRCh38
rs923192563 Chr.X: 30665683-30665683 on Build GRCh38
rs1050376728 Chr.X: 30665804-30665804 on Build GRCh38
rs780609835 Chr.X: 30665479-30665479 on Build GRCh38
rs767784694 Chr.X: 30665612-30665612 on Build GRCh38
rs892342811 Chr.X: 30665875-30665877 on Build GRCh38
rs1009796739 Chr.X: 30665888-30665888 on Build GRCh38
rs772946236 Chr.X: 30665596-30665596 on Build GRCh38
rs969090124 Chr.X: 30665578-30665578 on Build GRCh38
rs1023695225 Chr.X: 30665554-30665554 on Build GRCh38
rs762777173 Chr.X: 30665598-30665598 on Build GRCh38
rs762470061 Chr.X: 30665634-30665634 on Build GRCh38

View Full Product Details