Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40600497-40601038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6AP2
Entrez Gene ID
10159
Gene Name
ATPase H+ transporting accessory protein 2
Gene Aliases
APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, HT028, M8-9, MRXE, MRXSH, MSTP009, PRR, RENR, XMRE, XPDS
UniGene
Hs.495960
Gene Chromosome Location
Chr.X: 40580964-40606637 on Build GRCh38
Transcripts
NM_005765.2

SNP Detail

SNP ID Chromosome Location
rs796692227 Chr.X: 40600943-40600944 on Build GRCh38
rs5963810 Chr.X: 40600597-40600597 on Build GRCh38
rs920691383 Chr.X: 40600547-40600547 on Build GRCh38
rs796052284 Chr.X: 40600801-40600801 on Build GRCh38
rs756957661 Chr.X: 40600834-40600834 on Build GRCh38
rs915127567 Chr.X: 40600584-40600584 on Build GRCh38
rs754941083 Chr.X: 40601007-40601007 on Build GRCh38
rs930737503 Chr.X: 40600567-40600567 on Build GRCh38
rs752631976 Chr.X: 40600756-40600756 on Build GRCh38
rs1040292551 Chr.X: 40600757-40600757 on Build GRCh38
rs748585332 Chr.X: 40600920-40600920 on Build GRCh38
rs386416932 Chr.X: 40600932-40600933 on Build GRCh38
rs369610808 Chr.X: 40600736-40600736 on Build GRCh38
rs748473953 Chr.X: 40600930-40600931 on Build GRCh38
rs759415915 Chr.X: 40600725-40600725 on Build GRCh38
rs200731446 Chr.X: 40600931-40600931 on Build GRCh38
rs775975435 Chr.X: 40600932-40600932 on Build GRCh38
rs751460365 Chr.X: 40600922-40600922 on Build GRCh38
rs766677772 Chr.X: 40600711-40600711 on Build GRCh38
rs897518327 Chr.X: 40600773-40600773 on Build GRCh38
rs910420816 Chr.X: 40600606-40600606 on Build GRCh38
rs62586810 Chr.X: 40600947-40600947 on Build GRCh38
rs7691 Chr.X: 40600788-40600788 on Build GRCh38
rs770281803 Chr.X: 40600998-40600998 on Build GRCh38
rs985938151 Chr.X: 40600638-40600638 on Build GRCh38
rs751286464 Chr.X: 40600714-40600714 on Build GRCh38
rs149902247 Chr.X: 40600845-40600845 on Build GRCh38
rs111931105 Chr.X: 40600883-40600883 on Build GRCh38
rs763965127 Chr.X: 40600717-40600718 on Build GRCh38
rs988495837 Chr.X: 40600581-40600583 on Build GRCh38
rs753465548 Chr.X: 40600807-40600807 on Build GRCh38
rs1031658893 Chr.X: 40600534-40600534 on Build GRCh38
rs763065804 Chr.X: 40600927-40600927 on Build GRCh38
rs868510664 Chr.X: 40600976-40600976 on Build GRCh38
rs201270582 Chr.X: 40600895-40600895 on Build GRCh38
rs773258258 Chr.X: 40600924-40600924 on Build GRCh38
rs755257243 Chr.X: 40600929-40600930 on Build GRCh38
rs113375106 Chr.X: 40600882-40600882 on Build GRCh38
rs114591549 Chr.X: 40600964-40600964 on Build GRCh38
rs768963528 Chr.X: 40600944-40600945 on Build GRCh38
rs777353753 Chr.X: 40600789-40600789 on Build GRCh38
rs10590549 Chr.X: 40600931-40600932 on Build GRCh38
rs769027148 Chr.X: 40600880-40600880 on Build GRCh38
rs767246367 Chr.X: 40600743-40600743 on Build GRCh38
rs928953322 Chr.X: 40600842-40600842 on Build GRCh38
rs1049781297 Chr.X: 40600954-40600954 on Build GRCh38
rs781435666 Chr.X: 40600914-40600914 on Build GRCh38
rs866122254 Chr.X: 40600945-40600945 on Build GRCh38
rs747308629 Chr.X: 40600848-40600848 on Build GRCh38
rs72623242 Chr.X: 40600932-40600932 on Build GRCh38

View Full Product Details