Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 43949490-43949985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDP
Entrez Gene ID
4693
Gene Name
NDP, norrin cystine knot growth factor
Gene Aliases
EVR2, FEVR, ND
UniGene
Hs.522615
Gene Chromosome Location
Chr.X: 43948776-43973675 on Build GRCh38
Transcripts
NM_000266.3

SNP Detail

SNP ID Chromosome Location
rs756474198 Chr.X: 43949816-43949816 on Build GRCh38
rs137852220 Chr.X: 43949839-43949839 on Build GRCh38
rs956085715 Chr.X: 43949654-43949654 on Build GRCh38
rs104894867 Chr.X: 43949932-43949932 on Build GRCh38
rs104894878 Chr.X: 43949840-43949840 on Build GRCh38
rs772386222 Chr.X: 43949927-43949927 on Build GRCh38
rs968449222 Chr.X: 43949749-43949749 on Build GRCh38
rs28933684 Chr.X: 43949831-43949831 on Build GRCh38
rs113574479 Chr.X: 43949750-43949750 on Build GRCh38
rs779079304 Chr.X: 43949858-43949858 on Build GRCh38
rs794727478 Chr.X: 43949836-43949836 on Build GRCh38
rs749818420 Chr.X: 43949829-43949829 on Build GRCh38
rs104894875 Chr.X: 43949888-43949888 on Build GRCh38
rs990859747 Chr.X: 43949652-43949652 on Build GRCh38
rs761675721 Chr.X: 43949961-43949961 on Build GRCh38
rs979786330 Chr.X: 43949774-43949774 on Build GRCh38
rs1048545964 Chr.X: 43949863-43949863 on Build GRCh38
rs1030222239 Chr.X: 43949577-43949577 on Build GRCh38
rs776631081 Chr.X: 43949948-43949948 on Build GRCh38
rs104894883 Chr.X: 43949899-43949899 on Build GRCh38
rs1057518836 Chr.X: 43949887-43949887 on Build GRCh38
rs988622347 Chr.X: 43949702-43949702 on Build GRCh38
rs1057518793 Chr.X: 43949933-43949933 on Build GRCh38
rs768396026 Chr.X: 43949946-43949946 on Build GRCh38
rs931311293 Chr.X: 43949634-43949634 on Build GRCh38
rs997377300 Chr.X: 43949538-43949538 on Build GRCh38
rs140653237 Chr.X: 43949789-43949789 on Build GRCh38
rs372300274 Chr.X: 43949796-43949796 on Build GRCh38
rs761377592 Chr.X: 43949763-43949763 on Build GRCh38
rs78267814 Chr.X: 43949882-43949882 on Build GRCh38
rs727504030 Chr.X: 43949935-43949935 on Build GRCh38
rs104894873 Chr.X: 43949817-43949817 on Build GRCh38
rs104894871 Chr.X: 43949914-43949914 on Build GRCh38
rs104894877 Chr.X: 43949913-43949913 on Build GRCh38
rs104894876 Chr.X: 43949873-43949873 on Build GRCh38
rs886043023 Chr.X: 43949876-43949876 on Build GRCh38
rs757619775 Chr.X: 43949571-43949571 on Build GRCh38
rs369137059 Chr.X: 43949601-43949601 on Build GRCh38
rs900452652 Chr.X: 43949533-43949533 on Build GRCh38
rs749936422 Chr.X: 43949787-43949787 on Build GRCh38
rs760673487 Chr.X: 43949714-43949714 on Build GRCh38
rs888568881 Chr.X: 43949921-43949921 on Build GRCh38
rs778584609 Chr.X: 43949814-43949814 on Build GRCh38
rs767917288 Chr.X: 43949708-43949708 on Build GRCh38
rs916638099 Chr.X: 43949685-43949685 on Build GRCh38
rs745967643 Chr.X: 43949892-43949892 on Build GRCh38
rs752083237 Chr.X: 43949534-43949534 on Build GRCh38
rs73475744 Chr.X: 43949785-43949785 on Build GRCh38
rs771129712 Chr.X: 43949847-43949847 on Build GRCh38

View Full Product Details