Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47223466-47223964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK16
Entrez Gene ID
5127
Gene Name
cyclin dependent kinase 16
Gene Aliases
PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1
UniGene
Hs.496068
Gene Chromosome Location
Chr.X: 47217881-47229997 on Build GRCh38
Transcripts
NM_001170460.1 NM_006201.4 NM_033018.3 XM_011543920.2 XM_011543921.1 XM_011543922.1 XM_011543923.1 XM_011543924.1 XM_011543925.2 XM_011543926.1 XM_011543927.2 XM_011543928.1 XM_017029569.1 XM_017029570.1 XM_017029571.1 XM_017029572.1 XM_017029573.1

SNP Detail

SNP ID Chromosome Location
rs939000663 Chr.X: 47223636-47223636 on Build GRCh38
rs892211170 Chr.X: 47223670-47223670 on Build GRCh38
rs773527419 Chr.X: 47223684-47223684 on Build GRCh38
rs1019267098 Chr.X: 47223793-47223793 on Build GRCh38
rs901783744 Chr.X: 47223824-47223824 on Build GRCh38
rs752502574 Chr.X: 47223726-47223726 on Build GRCh38
rs763421284 Chr.X: 47223807-47223807 on Build GRCh38
rs1006772902 Chr.X: 47223508-47223508 on Build GRCh38
rs777629546 Chr.X: 47223631-47223631 on Build GRCh38
rs951369106 Chr.X: 47223872-47223872 on Build GRCh38
rs770246662 Chr.X: 47223782-47223782 on Build GRCh38
rs777717613 Chr.X: 47223727-47223727 on Build GRCh38
rs745553260 Chr.X: 47223646-47223646 on Build GRCh38
rs371576867 Chr.X: 47223736-47223736 on Build GRCh38
rs778322170 Chr.X: 47223751-47223751 on Build GRCh38
rs910017016 Chr.X: 47223565-47223565 on Build GRCh38
rs764418822 Chr.X: 47223619-47223619 on Build GRCh38
rs767712961 Chr.X: 47223722-47223722 on Build GRCh38
rs748834033 Chr.X: 47223632-47223632 on Build GRCh38
rs755643650 Chr.X: 47223870-47223870 on Build GRCh38
rs773807417 Chr.X: 47223525-47223525 on Build GRCh38
rs777199690 Chr.X: 47223674-47223674 on Build GRCh38
rs764501733 Chr.X: 47223554-47223554 on Build GRCh38
rs779776447 Chr.X: 47223902-47223904 on Build GRCh38
rs142251178 Chr.X: 47223706-47223706 on Build GRCh38
rs1007362279 Chr.X: 47223758-47223758 on Build GRCh38
rs180754755 Chr.X: 47223582-47223582 on Build GRCh38
rs747908155 Chr.X: 47223616-47223616 on Build GRCh38
rs377023226 Chr.X: 47223511-47223511 on Build GRCh38
rs187232602 Chr.X: 47223613-47223613 on Build GRCh38
rs765856405 Chr.X: 47223678-47223678 on Build GRCh38
rs186090617 Chr.X: 47223917-47223917 on Build GRCh38
rs763127145 Chr.X: 47223701-47223701 on Build GRCh38
rs183454877 Chr.X: 47223916-47223916 on Build GRCh38
rs774072481 Chr.X: 47223641-47223641 on Build GRCh38
rs766878365 Chr.X: 47223809-47223809 on Build GRCh38
rs759556075 Chr.X: 47223494-47223494 on Build GRCh38
rs539067436 Chr.X: 47223847-47223847 on Build GRCh38
rs762446019 Chr.X: 47223566-47223566 on Build GRCh38
rs754825902 Chr.X: 47223609-47223609 on Build GRCh38
rs867927569 Chr.X: 47223594-47223594 on Build GRCh38
rs761130276 Chr.X: 47223536-47223536 on Build GRCh38
rs969516113 Chr.X: 47223923-47223924 on Build GRCh38
rs757194532 Chr.X: 47223738-47223738 on Build GRCh38
rs750593584 Chr.X: 47223583-47223583 on Build GRCh38
rs755082329 Chr.X: 47223664-47223666 on Build GRCh38
rs369198498 Chr.X: 47223515-47223515 on Build GRCh38
rs781739652 Chr.X: 47223813-47223813 on Build GRCh38
rs762259531 Chr.X: 47223677-47223677 on Build GRCh38
rs998592637 Chr.X: 47223834-47223834 on Build GRCh38
rs752824712 Chr.X: 47223584-47223584 on Build GRCh38
rs770305098 Chr.X: 47223637-47223637 on Build GRCh38
rs771814612 Chr.X: 47223673-47223673 on Build GRCh38
rs766632657 Chr.X: 47223596-47223596 on Build GRCh38
rs778920672 Chr.X: 47223728-47223729 on Build GRCh38
rs933795059 Chr.X: 47223939-47223939 on Build GRCh38
rs140496175 Chr.X: 47223724-47223724 on Build GRCh38
rs771912390 Chr.X: 47223767-47223767 on Build GRCh38
rs994038845 Chr.X: 47223541-47223541 on Build GRCh38
rs200177369 Chr.X: 47223735-47223735 on Build GRCh38
rs755234077 Chr.X: 47223715-47223715 on Build GRCh38
rs866692551 Chr.X: 47223799-47223799 on Build GRCh38
rs748530996 Chr.X: 47223780-47223780 on Build GRCh38
rs980991361 Chr.X: 47223928-47223928 on Build GRCh38
rs751693661 Chr.X: 47223604-47223604 on Build GRCh38
rs2273272 Chr.X: 47223787-47223787 on Build GRCh38
rs151248551 Chr.X: 47223709-47223709 on Build GRCh38
rs748364754 Chr.X: 47223938-47223938 on Build GRCh38

View Full Product Details