Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47836740-47837236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF81
Entrez Gene ID
347344
Gene Name
zinc finger protein 81
Gene Aliases
HFZ20, MRX45, dJ54B20.6
UniGene
Hs.114246
Gene Chromosome Location
Chr.X: 47836864-47925626 on Build GRCh38
Transcripts
NM_007137.3 XM_005272600.3 XM_011543900.2 XM_017029486.1

SNP Detail

SNP ID Chromosome Location
rs980758677 Chr.X: 47837068-47837068 on Build GRCh38
rs1207492 Chr.X: 47837154-47837154 on Build GRCh38
rs782627675 Chr.X: 47837150-47837150 on Build GRCh38
rs1025189200 Chr.X: 47837055-47837055 on Build GRCh38
rs1050147738 Chr.X: 47836885-47836885 on Build GRCh38
rs1033710788 Chr.X: 47837112-47837112 on Build GRCh38
rs142198340 Chr.X: 47836840-47836840 on Build GRCh38
rs1016881047 Chr.X: 47836938-47836938 on Build GRCh38
rs782692640 Chr.X: 47836947-47836947 on Build GRCh38
rs898076909 Chr.X: 47837029-47837029 on Build GRCh38
rs782413396 Chr.X: 47837203-47837203 on Build GRCh38
rs782215507 Chr.X: 47836827-47836827 on Build GRCh38
rs782488145 Chr.X: 47836869-47836869 on Build GRCh38
rs994116394 Chr.X: 47837050-47837050 on Build GRCh38
rs932984303 Chr.X: 47836874-47836874 on Build GRCh38
rs372088446 Chr.X: 47836846-47836846 on Build GRCh38
rs980789631 Chr.X: 47836794-47836794 on Build GRCh38
rs1006378433 Chr.X: 47836923-47836923 on Build GRCh38
rs945994378 Chr.X: 47836842-47836842 on Build GRCh38
rs782471165 Chr.X: 47837179-47837179 on Build GRCh38
rs926724003 Chr.X: 47836816-47836816 on Build GRCh38
rs970674795 Chr.X: 47837064-47837064 on Build GRCh38
rs990417229 Chr.X: 47836833-47836833 on Build GRCh38
rs901472469 Chr.X: 47836856-47836856 on Build GRCh38
rs782533266 Chr.X: 47836803-47836803 on Build GRCh38
rs937153552 Chr.X: 47836832-47836832 on Build GRCh38
rs782765630 Chr.X: 47836798-47836798 on Build GRCh38
rs888893612 Chr.X: 47836901-47836901 on Build GRCh38
rs782039134 Chr.X: 47836818-47836818 on Build GRCh38
rs782485252 Chr.X: 47836782-47836782 on Build GRCh38

View Full Product Details