Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48686639-48687038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WAS
Entrez Gene ID
7454
Gene Name
Wiskott-Aldrich syndrome
Gene Aliases
IMD2, SCNX, THC, THC1, WASP, WASPA
UniGene
Hs.2157
Gene Chromosome Location
Chr.X: 48683753-48691427 on Build GRCh38
Transcripts
NM_000377.2 XM_011543977.2 XM_017029786.1

SNP Detail

SNP ID Chromosome Location
rs782693720 Chr.X: 48686740-48686740 on Build GRCh38
rs376545922 Chr.X: 48686815-48686815 on Build GRCh38
rs928562174 Chr.X: 48686890-48686890 on Build GRCh38
rs781860824 Chr.X: 48686797-48686797 on Build GRCh38
rs781954340 Chr.X: 48686710-48686710 on Build GRCh38
rs782415042 Chr.X: 48686927-48686927 on Build GRCh38
rs180938515 Chr.X: 48686995-48686995 on Build GRCh38
rs782259006 Chr.X: 48687003-48687003 on Build GRCh38
rs782489564 Chr.X: 48686777-48686777 on Build GRCh38
rs1017613764 Chr.X: 48686692-48686692 on Build GRCh38
rs371005311 Chr.X: 48686839-48686839 on Build GRCh38
rs377126493 Chr.X: 48686731-48686731 on Build GRCh38
rs782638115 Chr.X: 48686907-48686907 on Build GRCh38
rs782401032 Chr.X: 48686972-48686972 on Build GRCh38
rs132630270 Chr.X: 48686928-48686928 on Build GRCh38
rs371056209 Chr.X: 48686966-48686966 on Build GRCh38
rs370188924 Chr.X: 48686735-48686735 on Build GRCh38
rs782517747 Chr.X: 48686798-48686798 on Build GRCh38
rs782301435 Chr.X: 48686827-48686827 on Build GRCh38
rs374458439 Chr.X: 48686901-48686901 on Build GRCh38
rs938467045 Chr.X: 48686916-48686916 on Build GRCh38
rs782745511 Chr.X: 48686796-48686796 on Build GRCh38
rs963437222 Chr.X: 48686726-48686726 on Build GRCh38
rs782061167 Chr.X: 48686958-48686958 on Build GRCh38
rs782584950 Chr.X: 48686853-48686853 on Build GRCh38
rs782563873 Chr.X: 48686772-48686773 on Build GRCh38
rs782286374 Chr.X: 48686945-48686945 on Build GRCh38
rs947162120 Chr.X: 48686947-48686947 on Build GRCh38
rs782224455 Chr.X: 48686859-48686859 on Build GRCh38
rs781792192 Chr.X: 48686688-48686688 on Build GRCh38
rs200261212 Chr.X: 48686814-48686814 on Build GRCh38
rs374035804 Chr.X: 48687007-48687007 on Build GRCh38
rs368379103 Chr.X: 48686917-48686917 on Build GRCh38
rs781799705 Chr.X: 48686744-48686744 on Build GRCh38

View Full Product Details