Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107067403-107067929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM41
Entrez Gene ID
55285
Gene Name
RNA binding motif protein 41
Gene Aliases
-
UniGene
Hs.139053
Gene Chromosome Location
Chr.X: 107061885-107118827 on Build GRCh38
Transcripts
NM_001171080.1 NM_001324242.1 NM_001324243.1 NM_001324244.1 NM_018301.4 NR_136728.1 NR_136729.1 NR_136730.1 NR_136731.1 NR_136732.1 XM_011530980.2 XM_011530981.2

SNP Detail

SNP ID Chromosome Location
rs961608045 Chr.X: 107067628-107067628 on Build GRCh38
rs924188441 Chr.X: 107067883-107067883 on Build GRCh38
rs530938924 Chr.X: 107067768-107067768 on Build GRCh38
rs779150169 Chr.X: 107067879-107067881 on Build GRCh38
rs773559637 Chr.X: 107067519-107067519 on Build GRCh38
rs766967021 Chr.X: 107067678-107067678 on Build GRCh38
rs902767078 Chr.X: 107067539-107067539 on Build GRCh38
rs763221762 Chr.X: 107067517-107067518 on Build GRCh38
rs746251164 Chr.X: 107067472-107067472 on Build GRCh38
rs891475556 Chr.X: 107067898-107067898 on Build GRCh38
rs201179450 Chr.X: 107067591-107067591 on Build GRCh38
rs780411576 Chr.X: 107067483-107067483 on Build GRCh38
rs976720064 Chr.X: 107067659-107067659 on Build GRCh38
rs763525417 Chr.X: 107067596-107067596 on Build GRCh38
rs772741968 Chr.X: 107067680-107067680 on Build GRCh38
rs188307943 Chr.X: 107067748-107067748 on Build GRCh38
rs146046356 Chr.X: 107067513-107067513 on Build GRCh38
rs774217965 Chr.X: 107067581-107067581 on Build GRCh38
rs764575589 Chr.X: 107067615-107067616 on Build GRCh38
rs201043594 Chr.X: 107067598-107067598 on Build GRCh38
rs985612751 Chr.X: 107067443-107067443 on Build GRCh38
rs747725710 Chr.X: 107067562-107067562 on Build GRCh38
rs35091797 Chr.X: 107067640-107067641 on Build GRCh38
rs759201937 Chr.X: 107067692-107067692 on Build GRCh38
rs772335736 Chr.X: 107067515-107067515 on Build GRCh38
rs915434596 Chr.X: 107067459-107067459 on Build GRCh38
rs1008420811 Chr.X: 107067829-107067829 on Build GRCh38
rs764073827 Chr.X: 107067579-107067579 on Build GRCh38
rs1031776971 Chr.X: 107067489-107067489 on Build GRCh38
rs139896702 Chr.X: 107067661-107067661 on Build GRCh38
rs932226013 Chr.X: 107067800-107067800 on Build GRCh38
rs760216147 Chr.X: 107067693-107067693 on Build GRCh38
rs375893620 Chr.X: 107067633-107067633 on Build GRCh38
rs369133655 Chr.X: 107067742-107067742 on Build GRCh38
rs375097107 Chr.X: 107067586-107067586 on Build GRCh38
rs890610534 Chr.X: 107067806-107067806 on Build GRCh38
rs769652000 Chr.X: 107067548-107067548 on Build GRCh38
rs190229792 Chr.X: 107067595-107067595 on Build GRCh38
rs749711213 Chr.X: 107067495-107067495 on Build GRCh38
rs766235074 Chr.X: 107067599-107067599 on Build GRCh38
rs779461197 Chr.X: 107067604-107067604 on Build GRCh38
rs769883873 Chr.X: 107067891-107067891 on Build GRCh38
rs1037918766 Chr.X: 107067753-107067753 on Build GRCh38
rs143393406 Chr.X: 107067666-107067666 on Build GRCh38
rs779204935 Chr.X: 107067470-107067470 on Build GRCh38
rs934320252 Chr.X: 107067553-107067553 on Build GRCh38
rs771610732 Chr.X: 107067565-107067565 on Build GRCh38
rs747235400 Chr.X: 107067535-107067535 on Build GRCh38
rs758482338 Chr.X: 107067616-107067616 on Build GRCh38
rs1052402586 Chr.X: 107067801-107067801 on Build GRCh38
rs183776232 Chr.X: 107067745-107067745 on Build GRCh38
rs776769775 Chr.X: 107067642-107067642 on Build GRCh38
rs939628239 Chr.X: 107067890-107067890 on Build GRCh38
rs747177878 Chr.X: 107067626-107067626 on Build GRCh38
rs762700068 Chr.X: 107067566-107067566 on Build GRCh38
rs775388523 Chr.X: 107067559-107067559 on Build GRCh38
rs17850127 Chr.X: 107067643-107067643 on Build GRCh38
rs985581690 Chr.X: 107067701-107067701 on Build GRCh38
rs761307924 Chr.X: 107067582-107067582 on Build GRCh38
rs748235606 Chr.X: 107067514-107067514 on Build GRCh38
rs757705767 Chr.X: 107067437-107067437 on Build GRCh38
rs200106292 Chr.X: 107067597-107067597 on Build GRCh38
rs766029438 Chr.X: 107067740-107067740 on Build GRCh38
rs371858336 Chr.X: 107067504-107067504 on Build GRCh38
rs778010677 Chr.X: 107067619-107067619 on Build GRCh38
rs145969952 Chr.X: 107067605-107067605 on Build GRCh38

View Full Product Details