Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111298781-111299329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCX
Entrez Gene ID
1641
Gene Name
doublecortin
Gene Aliases
DBCN, DC, LISX, SCLH, XLIS
UniGene
Hs.34780
Gene Chromosome Location
Chr.X: 111293779-111412232 on Build GRCh38
Transcripts
NM_000555.3 NM_001195553.1 NM_178151.2 NM_178152.2 NM_178153.2 XM_011530878.2 XM_011530879.2 XM_011530880.2 XM_017029312.1

SNP Detail

SNP ID Chromosome Location
rs1031881202 Chr.X: 111298874-111298874 on Build GRCh38
rs769947646 Chr.X: 111298951-111298952 on Build GRCh38
rs921678386 Chr.X: 111298820-111298820 on Build GRCh38
rs764951400 Chr.X: 111298875-111298875 on Build GRCh38
rs1012663705 Chr.X: 111298984-111298984 on Build GRCh38
rs751945938 Chr.X: 111299267-111299267 on Build GRCh38
rs990263829 Chr.X: 111299229-111299229 on Build GRCh38
rs924092476 Chr.X: 111298918-111298918 on Build GRCh38
rs189925378 Chr.X: 111299086-111299086 on Build GRCh38
rs1051451688 Chr.X: 111299296-111299297 on Build GRCh38
rs559997251 Chr.X: 111298950-111298951 on Build GRCh38
rs762504911 Chr.X: 111299138-111299138 on Build GRCh38
rs1049054937 Chr.X: 111299005-111299005 on Build GRCh38
rs1004791631 Chr.X: 111299040-111299040 on Build GRCh38
rs1029170008 Chr.X: 111299217-111299217 on Build GRCh38
rs1008466633 Chr.X: 111298858-111298858 on Build GRCh38
rs989801126 Chr.X: 111298839-111298839 on Build GRCh38
rs374103647 Chr.X: 111298950-111298950 on Build GRCh38
rs758875806 Chr.X: 111299223-111299223 on Build GRCh38
rs1056962679 Chr.X: 111298972-111298972 on Build GRCh38
rs764676736 Chr.X: 111299148-111299148 on Build GRCh38
rs193201301 Chr.X: 111299224-111299224 on Build GRCh38
rs980099381 Chr.X: 111299042-111299042 on Build GRCh38
rs1037518425 Chr.X: 111299258-111299258 on Build GRCh38
rs1047701914 Chr.X: 111299225-111299225 on Build GRCh38
rs776021971 Chr.X: 111298952-111298953 on Build GRCh38
rs907781752 Chr.X: 111299237-111299237 on Build GRCh38
rs185796489 Chr.X: 111298823-111298823 on Build GRCh38
rs965887816 Chr.X: 111299105-111299105 on Build GRCh38
rs895197596 Chr.X: 111298981-111298981 on Build GRCh38
rs16986475 Chr.X: 111299039-111299039 on Build GRCh38
rs776472621 Chr.X: 111298807-111298807 on Build GRCh38
rs995905933 Chr.X: 111299292-111299292 on Build GRCh38
rs1014790397 Chr.X: 111299049-111299049 on Build GRCh38
rs1042568880 Chr.X: 111298884-111298884 on Build GRCh38
rs970141920 Chr.X: 111299038-111299038 on Build GRCh38
rs941872623 Chr.X: 111299244-111299244 on Build GRCh38
rs760911437 Chr.X: 111298882-111298882 on Build GRCh38
rs367594952 Chr.X: 111299281-111299281 on Build GRCh38
rs915483325 Chr.X: 111298860-111298860 on Build GRCh38

View Full Product Details