Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 129788345-129788820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SASH3
Entrez Gene ID
54440
Gene Name
SAM and SH3 domain containing 3
Gene Aliases
753P9, CXorf9, HACS2, SH3D6C, SLY
UniGene
Hs.61469
Gene Chromosome Location
Chr.X: 129779916-129795201 on Build GRCh38
Transcripts
NM_018990.3 XM_006724763.1

SNP Detail

SNP ID Chromosome Location
rs200345952 Chr.X: 129788493-129788493 on Build GRCh38
rs750490550 Chr.X: 129788409-129788409 on Build GRCh38
rs975693954 Chr.X: 129788790-129788790 on Build GRCh38
rs954314144 Chr.X: 129788787-129788787 on Build GRCh38
rs185925691 Chr.X: 129788675-129788675 on Build GRCh38
rs932067026 Chr.X: 129788676-129788676 on Build GRCh38
rs372399973 Chr.X: 129788587-129788587 on Build GRCh38
rs370279222 Chr.X: 129788412-129788412 on Build GRCh38
rs909404825 Chr.X: 129788763-129788763 on Build GRCh38
rs762296859 Chr.X: 129788568-129788568 on Build GRCh38
rs994308779 Chr.X: 129788376-129788376 on Build GRCh38
rs941326748 Chr.X: 129788784-129788784 on Build GRCh38
rs976573349 Chr.X: 129788645-129788645 on Build GRCh38
rs763333259 Chr.X: 129788553-129788553 on Build GRCh38
rs373745394 Chr.X: 129788442-129788442 on Build GRCh38
rs751989627 Chr.X: 129788383-129788383 on Build GRCh38
rs760907326 Chr.X: 129788607-129788607 on Build GRCh38
rs750507113 Chr.X: 129788388-129788403 on Build GRCh38
rs757507115 Chr.X: 129788411-129788411 on Build GRCh38
rs767739916 Chr.X: 129788404-129788404 on Build GRCh38
rs757355731 Chr.X: 129788399-129788399 on Build GRCh38
rs769234147 Chr.X: 129788783-129788783 on Build GRCh38
rs745351861 Chr.X: 129788762-129788762 on Build GRCh38
rs758210629 Chr.X: 129788627-129788627 on Build GRCh38
rs1031792637 Chr.X: 129788752-129788752 on Build GRCh38
rs367931250 Chr.X: 129788470-129788470 on Build GRCh38
rs1001334320 Chr.X: 129788416-129788416 on Build GRCh38
rs181791974 Chr.X: 129788378-129788378 on Build GRCh38
rs143665198 Chr.X: 129788511-129788511 on Build GRCh38
rs2231129 Chr.X: 129788616-129788616 on Build GRCh38
rs746831042 Chr.X: 129788490-129788490 on Build GRCh38
rs907168028 Chr.X: 129788662-129788662 on Build GRCh38
rs777743704 Chr.X: 129788660-129788660 on Build GRCh38
rs777820817 Chr.X: 129788628-129788628 on Build GRCh38
rs146852075 Chr.X: 129788554-129788554 on Build GRCh38
rs376975688 Chr.X: 129788379-129788379 on Build GRCh38
rs774521851 Chr.X: 129788566-129788566 on Build GRCh38
rs745337836 Chr.X: 129788522-129788522 on Build GRCh38
rs375620085 Chr.X: 129788510-129788510 on Build GRCh38
rs775165842 Chr.X: 129788549-129788549 on Build GRCh38
rs758391634 Chr.X: 129788448-129788448 on Build GRCh38
rs777364769 Chr.X: 129788468-129788468 on Build GRCh38
rs753956959 Chr.X: 129788626-129788626 on Build GRCh38
rs751190880 Chr.X: 129788670-129788670 on Build GRCh38
rs780774140 Chr.X: 129788716-129788716 on Build GRCh38
rs990083435 Chr.X: 129788601-129788601 on Build GRCh38
rs779253669 Chr.X: 129788529-129788529 on Build GRCh38
rs372485823 Chr.X: 129788585-129788585 on Build GRCh38

View Full Product Details