Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 192808949-192809452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS2
Entrez Gene ID
5997
Gene Name
regulator of G-protein signaling 2
Gene Aliases
G0S8
UniGene
Hs.78944
Gene Chromosome Location
Chr.1: 192809039-192812277 on Build GRCh38
Transcripts
NM_002923.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4995339 c.68G>A p.G23D Chr.1: 192809139-192809139 on Build GRCh38
3934344 c.4C>A p.Q2K Chr.1: 192809075-192809075 on Build GRCh38
6326048 c.25G>T p.V9F Chr.1: 192809096-192809096 on Build GRCh38
6060254 c.6A>T p.Q2H Chr.1: 192809077-192809077 on Build GRCh38
4840703 c.93A>C p.E31D Chr.1: 192809164-192809164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773800785 Chr.1: 192809161-192809161 on Build GRCh38
rs760249368 Chr.1: 192809057-192809057 on Build GRCh38
rs980675191 Chr.1: 192809212-192809212 on Build GRCh38
rs866356485 Chr.1: 192809369-192809369 on Build GRCh38
rs1026377769 Chr.1: 192809151-192809151 on Build GRCh38
rs763303561 Chr.1: 192809065-192809065 on Build GRCh38
rs769502722 Chr.1: 192809077-192809077 on Build GRCh38
rs1039116247 Chr.1: 192809211-192809211 on Build GRCh38
rs563683359 Chr.1: 192809073-192809073 on Build GRCh38
rs953539347 Chr.1: 192808984-192808984 on Build GRCh38
rs756254339 Chr.1: 192809086-192809086 on Build GRCh38
rs536686953 Chr.1: 192809012-192809012 on Build GRCh38
rs185750160 Chr.1: 192809133-192809133 on Build GRCh38
rs769235048 Chr.1: 192809117-192809117 on Build GRCh38
rs867658748 Chr.1: 192809220-192809220 on Build GRCh38
rs558037302 Chr.1: 192809007-192809007 on Build GRCh38
rs1040633610 Chr.1: 192809292-192809292 on Build GRCh38
rs745440695 Chr.1: 192809116-192809116 on Build GRCh38
rs12130714 Chr.1: 192809029-192809029 on Build GRCh38
rs74466425 Chr.1: 192809125-192809125 on Build GRCh38
rs1026935218 Chr.1: 192809311-192809311 on Build GRCh38
rs148489044 Chr.1: 192809139-192809139 on Build GRCh38
rs376661452 Chr.1: 192809120-192809120 on Build GRCh38
rs757270622 Chr.1: 192809042-192809042 on Build GRCh38
rs541275903 Chr.1: 192809272-192809272 on Build GRCh38
rs780440673 Chr.1: 192809089-192809089 on Build GRCh38
rs778221511 Chr.1: 192809041-192809041 on Build GRCh38
rs912151054 Chr.1: 192809420-192809420 on Build GRCh38
rs148929202 Chr.1: 192809071-192809071 on Build GRCh38
rs993680770 Chr.1: 192809310-192809310 on Build GRCh38
rs765465115 Chr.1: 192809206-192809206 on Build GRCh38
rs774626899 Chr.1: 192809059-192809066 on Build GRCh38
rs762233679 Chr.1: 192809152-192809152 on Build GRCh38
rs1051777810 Chr.1: 192809063-192809063 on Build GRCh38
rs371843799 Chr.1: 192809059-192809059 on Build GRCh38
rs369970749 Chr.1: 192809205-192809205 on Build GRCh38
rs890546712 Chr.1: 192809018-192809018 on Build GRCh38
rs369259338 Chr.1: 192809100-192809100 on Build GRCh38
rs1022535708 Chr.1: 192809049-192809049 on Build GRCh38
rs184293069 Chr.1: 192809027-192809027 on Build GRCh38
rs750495025 Chr.1: 192809070-192809070 on Build GRCh38
rs866708006 Chr.1: 192809419-192809419 on Build GRCh38
rs951949473 Chr.1: 192808994-192808994 on Build GRCh38
rs781487798 Chr.1: 192809047-192809047 on Build GRCh38
rs754132319 Chr.1: 192809200-192809200 on Build GRCh38
rs927759277 Chr.1: 192809212-192809212 on Build GRCh38
rs892495487 Chr.1: 192809002-192809002 on Build GRCh38
rs886353545 Chr.1: 192809247-192809247 on Build GRCh38
rs372230356 Chr.1: 192809105-192809105 on Build GRCh38
rs987714152 Chr.1: 192809404-192809404 on Build GRCh38
rs754050437 Chr.1: 192809093-192809093 on Build GRCh38
rs767970789 Chr.1: 192809157-192809157 on Build GRCh38
rs541337584 Chr.1: 192809048-192809048 on Build GRCh38
rs142499684 Chr.1: 192809082-192809082 on Build GRCh38
rs770282525 Chr.1: 192809058-192809058 on Build GRCh38
rs189578388 Chr.1: 192809080-192809080 on Build GRCh38
rs367901296 Chr.1: 192809072-192809072 on Build GRCh38
rs74716343 Chr.1: 192809231-192809231 on Build GRCh38
rs774619759 Chr.1: 192809142-192809142 on Build GRCh38
rs534938259 Chr.1: 192808982-192808982 on Build GRCh38
rs1032736092 Chr.1: 192809122-192809122 on Build GRCh38
rs984654492 Chr.1: 192808999-192808999 on Build GRCh38
rs1009581056 Chr.1: 192809006-192809006 on Build GRCh38
rs762913377 Chr.1: 192809101-192809101 on Build GRCh38
rs533676666 Chr.1: 192809060-192809060 on Build GRCh38
rs1045796970 Chr.1: 192809259-192809259 on Build GRCh38
rs141030117 Chr.1: 192809076-192809076 on Build GRCh38
rs564483322 Chr.1: 192809055-192809055 on Build GRCh38
rs987859903 Chr.1: 192809203-192809203 on Build GRCh38
rs766529182 Chr.1: 192809199-192809199 on Build GRCh38
rs970046081 Chr.1: 192809409-192809409 on Build GRCh38
rs1032681987 Chr.1: 192809276-192809276 on Build GRCh38
rs1019097728 Chr.1: 192809131-192809131 on Build GRCh38
rs902114592 Chr.1: 192809297-192809297 on Build GRCh38
rs372424395 Chr.1: 192809025-192809025 on Build GRCh38
rs374205631 Chr.1: 192809031-192809031 on Build GRCh38
rs944806999 Chr.1: 192809019-192809019 on Build GRCh38
rs781181895 Chr.1: 192809106-192809106 on Build GRCh38
rs957124649 Chr.1: 192809299-192809299 on Build GRCh38
rs117986927 Chr.1: 192809215-192809215 on Build GRCh38
rs367837079 Chr.1: 192809040-192809040 on Build GRCh38
rs879230476 Chr.1: 192808990-192808990 on Build GRCh38
rs562689527 Chr.1: 192809296-192809297 on Build GRCh38
rs781632024 Chr.1: 192809389-192809389 on Build GRCh38
rs760844773 Chr.1: 192809179-192809179 on Build GRCh38
rs759349861 Chr.1: 192809064-192809064 on Build GRCh38
rs370673230 Chr.1: 192809035-192809035 on Build GRCh38
rs748585377 Chr.1: 192809021-192809021 on Build GRCh38
rs753890153 Chr.1: 192809111-192809111 on Build GRCh38
rs888801862 Chr.1: 192809290-192809290 on Build GRCh38
rs143288784 Chr.1: 192809069-192809069 on Build GRCh38
rs778871706 Chr.1: 192809251-192809251 on Build GRCh38
rs201310006 Chr.1: 192809273-192809273 on Build GRCh38
rs75930797 Chr.1: 192809034-192809034 on Build GRCh38
rs767540087 Chr.1: 192809032-192809032 on Build GRCh38
rs751951851 Chr.1: 192809067-192809067 on Build GRCh38
rs377647978 Chr.1: 192809219-192809219 on Build GRCh38
rs747921480 Chr.1: 192809051-192809051 on Build GRCh38
rs760672186 Chr.1: 192809075-192809075 on Build GRCh38
rs1007784918 Chr.1: 192809291-192809291 on Build GRCh38
rs757061626 Chr.1: 192809213-192809213 on Build GRCh38
rs905498660 Chr.1: 192809264-192809264 on Build GRCh38
rs545452217 Chr.1: 192809112-192809112 on Build GRCh38
rs755372508 Chr.1: 192809037-192809037 on Build GRCh38
rs200916949 Chr.1: 192809038-192809038 on Build GRCh38
rs577574239 Chr.1: 192809010-192809010 on Build GRCh38
rs188922079 Chr.1: 192809210-192809210 on Build GRCh38
rs768501145 Chr.1: 192809030-192809030 on Build GRCh38
rs368290103 Chr.1: 192809050-192809050 on Build GRCh38
rs762277643 Chr.1: 192809083-192809083 on Build GRCh38
rs755650674 Chr.1: 192809228-192809228 on Build GRCh38
rs193051407 Chr.1: 192809084-192809084 on Build GRCh38
rs145125159 Chr.1: 192809078-192809078 on Build GRCh38
rs966247612 Chr.1: 192809132-192809132 on Build GRCh38
rs772206894 Chr.1: 192809053-192809053 on Build GRCh38
rs377637167 Chr.1: 192809066-192809066 on Build GRCh38
rs558306841 Chr.1: 192809401-192809401 on Build GRCh38

View Full Product Details