Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193059325-193059815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TROVE2
Entrez Gene ID
6738
Gene Name
TROVE domain family member 2
Gene Aliases
RO60, RORNP, SSA2
UniGene
Hs.288178
Gene Chromosome Location
Chr.1: 193059422-193091777 on Build GRCh38
Transcripts
NM_001173524.1 NR_033393.1 XM_006711495.3 XM_006711496.3 XM_006711497.3 XM_011509922.2 XM_017002180.1 XM_017002181.1 XM_017002182.1 XM_017002183.1 NM_001199261.1 NM_001199262.1 NM_001199263.1 NM_015984.3 NR_037607.1 XM_005245244.4 XM_005245246.4 XM_005245247.4 XM_005245248.4 XM_005245249.4 XM_006711366.3 XM_006711367.3 XM_006711368.3 XM_006711369.2 XM_006711370.3 XM_006711371.3 XM_011509604.1 XM_011509607.2 XM_011509608.2 XM_011509609.2 XM_017001429.1 XM_017001430.1 XM_017001431.1 XM_017001432.1 XM_017001433.1 XM_017001434.1 XM_017001435.1 XM_017001436.1 XM_017001437.1 XM_017001438.1 XM_017001439.1 XM_017001440.1 XM_017001441.1 XM_017001442.1 XM_017001443.1 XM_017001444.1

Gene Symbol
UCHL5
Entrez Gene ID
51377
Gene Name
ubiquitin C-terminal hydrolase L5
Gene Aliases
CGI-70, INO80R, UCH-L5, UCH37
UniGene
Hs.145469
Gene Chromosome Location
Chr.1: 193012366-193060140 on Build GRCh38
Transcripts
NM_001173524.1 NR_033393.1 XM_006711495.3 XM_006711496.3 XM_006711497.3 XM_011509922.2 XM_017002180.1 XM_017002181.1 XM_017002182.1 XM_017002183.1 NM_001199261.1 NM_001199262.1 NM_001199263.1 NM_015984.3 NR_037607.1 XM_005245244.4 XM_005245246.4 XM_005245247.4 XM_005245248.4 XM_005245249.4 XM_006711366.3 XM_006711367.3 XM_006711368.3 XM_006711369.2 XM_006711370.3 XM_006711371.3 XM_011509604.1 XM_011509607.2 XM_011509608.2 XM_011509609.2 XM_017001429.1 XM_017001430.1 XM_017001431.1 XM_017001432.1 XM_017001433.1 XM_017001434.1 XM_017001435.1 XM_017001436.1 XM_017001437.1 XM_017001438.1 XM_017001439.1 XM_017001440.1 XM_017001441.1 XM_017001442.1 XM_017001443.1 XM_017001444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6897192 c.460C>T p.R154C Chr.1: 193059422-193059422 on Build GRCh38
4447375 c.375C>T p.A125A Chr.1: 193059507-193059507 on Build GRCh38
4447376 c.367-1G>A p.? Chr.1: 193059516-193059516 on Build GRCh38
7109713 c.369C>T p.A123A Chr.1: 193059513-193059513 on Build GRCh38
6875243 c.469+1G>A p.? Chr.1: 193059412-193059412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915051827 Chr.1: 193059450-193059450 on Build GRCh38
rs755000610 Chr.1: 193059597-193059597 on Build GRCh38
rs746782874 Chr.1: 193059346-193059346 on Build GRCh38
rs928451556 Chr.1: 193059502-193059502 on Build GRCh38
rs958795489 Chr.1: 193059413-193059413 on Build GRCh38
rs755016729 Chr.1: 193059422-193059422 on Build GRCh38
rs771708450 Chr.1: 193059479-193059479 on Build GRCh38
rs756179067 Chr.1: 193059465-193059465 on Build GRCh38
rs574106821 Chr.1: 193059659-193059659 on Build GRCh38
rs761236410 Chr.1: 193059417-193059417 on Build GRCh38
rs755906052 Chr.1: 193059531-193059531 on Build GRCh38
rs776497792 Chr.1: 193059667-193059667 on Build GRCh38
rs566136646 Chr.1: 193059435-193059435 on Build GRCh38
rs752691558 Chr.1: 193059518-193059518 on Build GRCh38
rs938614756 Chr.1: 193059516-193059516 on Build GRCh38
rs750258277 Chr.1: 193059526-193059526 on Build GRCh38
rs775773753 Chr.1: 193059579-193059579 on Build GRCh38
rs759393706 Chr.1: 193059770-193059770 on Build GRCh38
rs776699194 Chr.1: 193059628-193059628 on Build GRCh38
rs748755345 Chr.1: 193059470-193059470 on Build GRCh38
rs558260630 Chr.1: 193059583-193059583 on Build GRCh38
rs747643089 Chr.1: 193059544-193059544 on Build GRCh38
rs1043203307 Chr.1: 193059364-193059364 on Build GRCh38
rs773989576 Chr.1: 193059407-193059407 on Build GRCh38
rs1024380496 Chr.1: 193059668-193059673 on Build GRCh38
rs773624204 Chr.1: 193059480-193059480 on Build GRCh38
rs777153396 Chr.1: 193059494-193059494 on Build GRCh38
rs1036452061 Chr.1: 193059775-193059775 on Build GRCh38
rs557556271 Chr.1: 193059638-193059638 on Build GRCh38
rs746210569 Chr.1: 193059653-193059653 on Build GRCh38
rs781177060 Chr.1: 193059449-193059449 on Build GRCh38
rs953386694 Chr.1: 193059462-193059462 on Build GRCh38
rs756996124 Chr.1: 193059691-193059691 on Build GRCh38
rs771722738 Chr.1: 193059546-193059546 on Build GRCh38
rs773262274 Chr.1: 193059763-193059763 on Build GRCh38
rs560261095 Chr.1: 193059455-193059455 on Build GRCh38
rs774552709 Chr.1: 193059664-193059664 on Build GRCh38
rs763338014 Chr.1: 193059522-193059522 on Build GRCh38
rs776074192 Chr.1: 193059658-193059658 on Build GRCh38
rs772343277 Chr.1: 193059760-193059760 on Build GRCh38
rs778551602 Chr.1: 193059539-193059539 on Build GRCh38
rs756394454 Chr.1: 193059720-193059720 on Build GRCh38
rs758305862 Chr.1: 193059679-193059679 on Build GRCh38
rs751792697 Chr.1: 193059584-193059584 on Build GRCh38
rs765406131 Chr.1: 193059429-193059429 on Build GRCh38
rs374438890 Chr.1: 193059418-193059418 on Build GRCh38
rs763452972 Chr.1: 193059722-193059722 on Build GRCh38
rs190007176 Chr.1: 193059438-193059438 on Build GRCh38
rs977443312 Chr.1: 193059426-193059426 on Build GRCh38
rs898079732 Chr.1: 193059568-193059568 on Build GRCh38
rs530753177 Chr.1: 193059371-193059371 on Build GRCh38
rs199691954 Chr.1: 193059474-193059474 on Build GRCh38
rs763767679 Chr.1: 193059530-193059530 on Build GRCh38
rs773835261 Chr.1: 193059655-193059656 on Build GRCh38
rs368044332 Chr.1: 193059637-193059637 on Build GRCh38
rs369216608 Chr.1: 193059547-193059547 on Build GRCh38
rs773534587 Chr.1: 193059416-193059416 on Build GRCh38
rs747836649 Chr.1: 193059477-193059477 on Build GRCh38
rs779984713 Chr.1: 193059467-193059467 on Build GRCh38
rs748818112 Chr.1: 193059391-193059391 on Build GRCh38
rs776785054 Chr.1: 193059453-193059454 on Build GRCh38
rs774932777 Chr.1: 193059390-193059390 on Build GRCh38
rs747403377 Chr.1: 193059548-193059548 on Build GRCh38
rs777424002 Chr.1: 193059622-193059622 on Build GRCh38
rs760131149 Chr.1: 193059563-193059563 on Build GRCh38
rs759183061 Chr.1: 193059669-193059669 on Build GRCh38
rs762160317 Chr.1: 193059665-193059665 on Build GRCh38
rs776841435 Chr.1: 193059553-193059553 on Build GRCh38
rs769281190 Chr.1: 193059751-193059751 on Build GRCh38
rs763637741 Chr.1: 193059686-193059686 on Build GRCh38
rs771274393 Chr.1: 193059549-193059549 on Build GRCh38
rs931783115 Chr.1: 193059469-193059469 on Build GRCh38
rs1015625242 Chr.1: 193059358-193059358 on Build GRCh38
rs767916160 Chr.1: 193059410-193059410 on Build GRCh38
rs770453297 Chr.1: 193059579-193059579 on Build GRCh38
rs752580164 Chr.1: 193059678-193059678 on Build GRCh38
rs765186607 Chr.1: 193059672-193059672 on Build GRCh38
rs567314976 Chr.1: 193059395-193059395 on Build GRCh38
rs765410075 Chr.1: 193059508-193059508 on Build GRCh38
rs760410119 Chr.1: 193059587-193059587 on Build GRCh38
rs746248721 Chr.1: 193059713-193059713 on Build GRCh38
rs781303295 Chr.1: 193059373-193059373 on Build GRCh38
rs3834070 Chr.1: 193059734-193059736 on Build GRCh38
rs778657251 Chr.1: 193059609-193059609 on Build GRCh38
rs745852506 Chr.1: 193059379-193059379 on Build GRCh38
rs764401466 Chr.1: 193059523-193059523 on Build GRCh38
rs1027745813 Chr.1: 193059400-193059400 on Build GRCh38
rs148813425 Chr.1: 193059496-193059496 on Build GRCh38
rs369154096 Chr.1: 193059421-193059421 on Build GRCh38
rs780653948 Chr.1: 193059703-193059703 on Build GRCh38
rs762947133 Chr.1: 193059786-193059786 on Build GRCh38
rs1011209470 Chr.1: 193059360-193059360 on Build GRCh38
rs868182289 Chr.1: 193059434-193059434 on Build GRCh38
rs769826855 Chr.1: 193059380-193059380 on Build GRCh38
rs761916189 Chr.1: 193059535-193059536 on Build GRCh38
rs762105906 Chr.1: 193059408-193059408 on Build GRCh38
rs754838839 Chr.1: 193059538-193059538 on Build GRCh38
rs953630764 Chr.1: 193059627-193059627 on Build GRCh38
rs959864589 Chr.1: 193059747-193059747 on Build GRCh38
rs750305351 Chr.1: 193059693-193059693 on Build GRCh38
rs757860045 Chr.1: 193059617-193059617 on Build GRCh38
rs768675750 Chr.1: 193059372-193059372 on Build GRCh38
rs778262800 Chr.1: 193059476-193059476 on Build GRCh38
rs962478263 Chr.1: 193059639-193059639 on Build GRCh38
rs752204405 Chr.1: 193059613-193059613 on Build GRCh38
rs17435945 Chr.1: 193059732-193059734 on Build GRCh38
rs760568377 Chr.1: 193059487-193059487 on Build GRCh38
rs371900922 Chr.1: 193059758-193059758 on Build GRCh38
rs995071591 Chr.1: 193059570-193059570 on Build GRCh38
rs543312220 Chr.1: 193059750-193059750 on Build GRCh38
rs550735071 Chr.1: 193059372-193059372 on Build GRCh38
rs947062013 Chr.1: 193059555-193059555 on Build GRCh38
rs781670691 Chr.1: 193059640-193059640 on Build GRCh38
rs775063451 Chr.1: 193059754-193059755 on Build GRCh38
rs764008097 Chr.1: 193059787-193059787 on Build GRCh38
rs1008549433 Chr.1: 193059352-193059352 on Build GRCh38
rs374760111 Chr.1: 193059562-193059562 on Build GRCh38
rs962140158 Chr.1: 193059491-193059491 on Build GRCh38
rs765062215 Chr.1: 193059782-193059782 on Build GRCh38
rs763070484 Chr.1: 193059581-193059581 on Build GRCh38
rs2887639 Chr.1: 193059625-193059625 on Build GRCh38
rs775232023 Chr.1: 193059784-193059784 on Build GRCh38
rs768809409 Chr.1: 193059660-193059660 on Build GRCh38
rs770202512 Chr.1: 193059567-193059567 on Build GRCh38
rs774628651 Chr.1: 193059755-193059755 on Build GRCh38
rs780730938 Chr.1: 193059740-193059740 on Build GRCh38
rs1054039178 Chr.1: 193059448-193059448 on Build GRCh38
rs770350655 Chr.1: 193059657-193059657 on Build GRCh38
rs745658262 Chr.1: 193059460-193059460 on Build GRCh38
rs928671146 Chr.1: 193059743-193059743 on Build GRCh38
rs779889336 Chr.1: 193059533-193059533 on Build GRCh38
rs574981667 Chr.1: 193059591-193059591 on Build GRCh38
rs753569159 Chr.1: 193059592-193059592 on Build GRCh38
rs902958268 Chr.1: 193059580-193059580 on Build GRCh38
rs771343075 Chr.1: 193059490-193059490 on Build GRCh38

View Full Product Details