Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202193668-202194157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGR6
Entrez Gene ID
59352
Gene Name
leucine rich repeat containing G protein-coupled receptor 6
Gene Aliases
GPCR, VTS20631
UniGene
Hs.497402
Gene Chromosome Location
Chr.1: 202193990-202319761 on Build GRCh38
Transcripts
NM_001017403.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5718876 c.92G>A p.G31E Chr.1: 202194081-202194081 on Build GRCh38
3481652 c.82C>T p.P28S Chr.1: 202194071-202194071 on Build GRCh38
5038581 c.89C>G p.P30R Chr.1: 202194078-202194078 on Build GRCh38
6222991 c.109G>C p.A37P Chr.1: 202194098-202194098 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs571155507 Chr.1: 202194079-202194079 on Build GRCh38
rs528901762 Chr.1: 202193695-202193695 on Build GRCh38
rs1015545086 Chr.1: 202193985-202193985 on Build GRCh38
rs780714353 Chr.1: 202194071-202194071 on Build GRCh38
rs763826160 Chr.1: 202193948-202193948 on Build GRCh38
rs761492020 Chr.1: 202194041-202194041 on Build GRCh38
rs1042641769 Chr.1: 202193856-202193856 on Build GRCh38
rs777775153 Chr.1: 202194097-202194097 on Build GRCh38
rs76654397 Chr.1: 202194133-202194133 on Build GRCh38
rs932520708 Chr.1: 202193844-202193852 on Build GRCh38
rs779402458 Chr.1: 202194078-202194078 on Build GRCh38
rs973491443 Chr.1: 202194015-202194015 on Build GRCh38
rs150296775 Chr.1: 202194067-202194067 on Build GRCh38
rs991066985 Chr.1: 202193711-202193711 on Build GRCh38
rs907967507 Chr.1: 202193829-202193830 on Build GRCh38
rs755420828 Chr.1: 202194074-202194074 on Build GRCh38
rs1025200608 Chr.1: 202193965-202193965 on Build GRCh38
rs942131931 Chr.1: 202193832-202193832 on Build GRCh38
rs963485863 Chr.1: 202194012-202194012 on Build GRCh38
rs762854585 Chr.1: 202194119-202194119 on Build GRCh38
rs368357450 Chr.1: 202194136-202194136 on Build GRCh38
rs770024037 Chr.1: 202194093-202194094 on Build GRCh38
rs775426943 Chr.1: 202194115-202194115 on Build GRCh38
rs890939593 Chr.1: 202193850-202193850 on Build GRCh38
rs1025066252 Chr.1: 202193754-202193754 on Build GRCh38
rs959612406 Chr.1: 202193706-202193706 on Build GRCh38
rs537785733 Chr.1: 202193959-202193959 on Build GRCh38
rs970924892 Chr.1: 202193756-202193756 on Build GRCh38
rs961982069 Chr.1: 202194086-202194086 on Build GRCh38
rs1001062905 Chr.1: 202193887-202193887 on Build GRCh38
rs754361019 Chr.1: 202194073-202194073 on Build GRCh38
rs943898327 Chr.1: 202193855-202193855 on Build GRCh38
rs973425226 Chr.1: 202193836-202193836 on Build GRCh38
rs766999265 Chr.1: 202194055-202194055 on Build GRCh38
rs548676284 Chr.1: 202193909-202193909 on Build GRCh38
rs12748690 Chr.1: 202193720-202193720 on Build GRCh38
rs770874974 Chr.1: 202194114-202194114 on Build GRCh38
rs774753564 Chr.1: 202193974-202193974 on Build GRCh38
rs2361431 Chr.1: 202193935-202193935 on Build GRCh38
rs12752893 Chr.1: 202193984-202193984 on Build GRCh38
rs747127917 Chr.1: 202194099-202194099 on Build GRCh38
rs912470052 Chr.1: 202194082-202194082 on Build GRCh38
rs983920570 Chr.1: 202193769-202193769 on Build GRCh38
rs971276548 Chr.1: 202193967-202193967 on Build GRCh38
rs774124605 Chr.1: 202194139-202194139 on Build GRCh38
rs902243375 Chr.1: 202193868-202193868 on Build GRCh38
rs1012416262 Chr.1: 202193951-202193951 on Build GRCh38
rs1049675665 Chr.1: 202193846-202193847 on Build GRCh38
rs751261112 Chr.1: 202194032-202194032 on Build GRCh38
rs921959924 Chr.1: 202193838-202193838 on Build GRCh38
rs772335116 Chr.1: 202194096-202194096 on Build GRCh38
rs1053377183 Chr.1: 202193929-202193929 on Build GRCh38

View Full Product Details