Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206767590-206768116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL10
Entrez Gene ID
3586
Gene Name
interleukin 10
Gene Aliases
CSIF, GVHDS, IL-10, IL10A, TGIF
UniGene
Hs.193717
Gene Chromosome Location
Chr.1: 206767603-206772494 on Build GRCh38
Transcripts
NM_000572.2

SNP Detail

SNP ID Chromosome Location
rs41432052 Chr.1: 206767663-206767663 on Build GRCh38
rs888877216 Chr.1: 206768026-206768026 on Build GRCh38
rs892735266 Chr.1: 206768064-206768064 on Build GRCh38
rs560237614 Chr.1: 206767937-206767937 on Build GRCh38
rs950652961 Chr.1: 206767960-206767960 on Build GRCh38
rs577489766 Chr.1: 206767763-206767763 on Build GRCh38
rs1027786456 Chr.1: 206768001-206768001 on Build GRCh38
rs563939146 Chr.1: 206767755-206767755 on Build GRCh38
rs904195200 Chr.1: 206767716-206767716 on Build GRCh38
rs45549141 Chr.1: 206767835-206767835 on Build GRCh38
rs756406101 Chr.1: 206768035-206768035 on Build GRCh38
rs570361233 Chr.1: 206768025-206768025 on Build GRCh38
rs34162991 Chr.1: 206767747-206767747 on Build GRCh38
rs1018640953 Chr.1: 206768080-206768080 on Build GRCh38
rs943116671 Chr.1: 206767701-206767701 on Build GRCh38
rs6687786 Chr.1: 206767875-206767875 on Build GRCh38
rs548565659 Chr.1: 206767879-206767879 on Build GRCh38
rs919144833 Chr.1: 206767656-206767656 on Build GRCh38
rs892710292 Chr.1: 206767856-206767856 on Build GRCh38
rs764050049 Chr.1: 206768005-206768005 on Build GRCh38
rs550777040 Chr.1: 206768022-206768022 on Build GRCh38
rs562134038 Chr.1: 206767891-206767891 on Build GRCh38
rs1018588769 Chr.1: 206767724-206767724 on Build GRCh38
rs937668341 Chr.1: 206767973-206767973 on Build GRCh38
rs977721049 Chr.1: 206767771-206767771 on Build GRCh38
rs987768546 Chr.1: 206767976-206767976 on Build GRCh38
rs550854859 Chr.1: 206768019-206768019 on Build GRCh38
rs935821876 Chr.1: 206767824-206767824 on Build GRCh38
rs561904879 Chr.1: 206767890-206767890 on Build GRCh38
rs3024499 Chr.1: 206767684-206767684 on Build GRCh38
rs34987208 Chr.1: 206767878-206767879 on Build GRCh38
rs915327103 Chr.1: 206767868-206767868 on Build GRCh38
rs560076279 Chr.1: 206767876-206767876 on Build GRCh38
rs776158751 Chr.1: 206767825-206767825 on Build GRCh38
rs528755040 Chr.1: 206767878-206767878 on Build GRCh38
rs911739023 Chr.1: 206767977-206767977 on Build GRCh38
rs966083083 Chr.1: 206768091-206768091 on Build GRCh38
rs898026394 Chr.1: 206767915-206767915 on Build GRCh38
rs924494313 Chr.1: 206767797-206767797 on Build GRCh38
rs989614985 Chr.1: 206767853-206767853 on Build GRCh38
rs1010177399 Chr.1: 206767872-206767872 on Build GRCh38
rs531127355 Chr.1: 206767918-206767918 on Build GRCh38
rs575320961 Chr.1: 206767666-206767666 on Build GRCh38
rs911652710 Chr.1: 206767699-206767699 on Build GRCh38
rs1038771347 Chr.1: 206767704-206767704 on Build GRCh38
rs542356582 Chr.1: 206767752-206767752 on Build GRCh38
rs926332207 Chr.1: 206767959-206767959 on Build GRCh38
rs1052774436 Chr.1: 206768036-206768036 on Build GRCh38
rs5743629 Chr.1: 206768021-206768021 on Build GRCh38
rs879722125 Chr.1: 206767751-206767751 on Build GRCh38
rs974491434 Chr.1: 206768010-206768010 on Build GRCh38
rs945667057 Chr.1: 206768088-206768088 on Build GRCh38
rs1054484414 Chr.1: 206767839-206767839 on Build GRCh38
rs374338753 Chr.1: 206767896-206767896 on Build GRCh38
rs935613934 Chr.1: 206767837-206767837 on Build GRCh38

View Full Product Details