Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 218345094-218345599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFB2
Entrez Gene ID
7042
Gene Name
transforming growth factor beta 2
Gene Aliases
LDS4, TGF-beta2
UniGene
Hs.133379
Gene Chromosome Location
Chr.1: 218345334-218444619 on Build GRCh38
Transcripts
NM_001135599.2 NM_003238.3 NR_046268.1

Gene Symbol
TGFB2-AS1
Entrez Gene ID
728463
Gene Name
TGFB2 antisense RNA 1 (head to head)
Gene Aliases
-
UniGene
Hs.568469
Gene Chromosome Location
Chr.1: 218344196-218345678 on Build GRCh38
Transcripts
NM_001135599.2 NM_003238.3 NR_046268.1

SNP Detail

SNP ID Chromosome Location
rs182668739 Chr.1: 218345280-218345280 on Build GRCh38
rs527548756 Chr.1: 218345557-218345557 on Build GRCh38
rs1035061456 Chr.1: 218345266-218345266 on Build GRCh38
rs553448992 Chr.1: 218345334-218345334 on Build GRCh38
rs890469471 Chr.1: 218345163-218345163 on Build GRCh38
rs568654101 Chr.1: 218345226-218345226 on Build GRCh38
rs529994753 Chr.1: 218345248-218345248 on Build GRCh38
rs886045959 Chr.1: 218345549-218345549 on Build GRCh38
rs1002933360 Chr.1: 218345453-218345453 on Build GRCh38
rs930783155 Chr.1: 218345505-218345505 on Build GRCh38
rs538038366 Chr.1: 218345183-218345183 on Build GRCh38
rs1035765337 Chr.1: 218345459-218345459 on Build GRCh38
rs1048322487 Chr.1: 218345521-218345521 on Build GRCh38
rs932394021 Chr.1: 218345342-218345342 on Build GRCh38
rs886045958 Chr.1: 218345431-218345431 on Build GRCh38
rs1039081763 Chr.1: 218345480-218345480 on Build GRCh38
rs986748665 Chr.1: 218345432-218345432 on Build GRCh38
rs7550232 Chr.1: 218345173-218345173 on Build GRCh38
rs562062506 Chr.1: 218345482-218345482 on Build GRCh38
rs886045957 Chr.1: 218345435-218345436 on Build GRCh38
rs933965340 Chr.1: 218345143-218345143 on Build GRCh38
rs555819740 Chr.1: 218345434-218345434 on Build GRCh38
rs886615793 Chr.1: 218345538-218345538 on Build GRCh38
rs576544841 Chr.1: 218345363-218345363 on Build GRCh38
rs540633823 Chr.1: 218345562-218345562 on Build GRCh38
rs979775498 Chr.1: 218345531-218345531 on Build GRCh38
rs1023096585 Chr.1: 218345438-218345438 on Build GRCh38
rs1052161642 Chr.1: 218345161-218345161 on Build GRCh38
rs899665931 Chr.1: 218345325-218345325 on Build GRCh38
rs79389033 Chr.1: 218345519-218345519 on Build GRCh38
rs943404961 Chr.1: 218345170-218345170 on Build GRCh38
rs1047926171 Chr.1: 218345237-218345237 on Build GRCh38
rs967807603 Chr.1: 218345346-218345346 on Build GRCh38
rs955435489 Chr.1: 218345414-218345414 on Build GRCh38
rs11466364 Chr.1: 218345577-218345577 on Build GRCh38
rs10482718 Chr.1: 218345400-218345400 on Build GRCh38
rs796274385 Chr.1: 218345425-218345432 on Build GRCh38
rs532676710 Chr.1: 218345578-218345578 on Build GRCh38
rs959042613 Chr.1: 218345274-218345274 on Build GRCh38
rs1051300736 Chr.1: 218345353-218345353 on Build GRCh38
rs923848715 Chr.1: 218345128-218345128 on Build GRCh38
rs946666002 Chr.1: 218345286-218345286 on Build GRCh38
rs1021988847 Chr.1: 218345309-218345309 on Build GRCh38
rs762270178 Chr.1: 218345200-218345200 on Build GRCh38
rs1003814377 Chr.1: 218345386-218345386 on Build GRCh38
rs11466363 Chr.1: 218345472-218345472 on Build GRCh38
rs978329367 Chr.1: 218345360-218345360 on Build GRCh38
rs996167129 Chr.1: 218345431-218345432 on Build GRCh38
rs544635781 Chr.1: 218345273-218345273 on Build GRCh38
rs11466361 Chr.1: 218345147-218345147 on Build GRCh38
rs113028862 Chr.1: 218345470-218345470 on Build GRCh38
rs967958942 Chr.1: 218345522-218345522 on Build GRCh38
rs765653988 Chr.1: 218345238-218345238 on Build GRCh38
rs920668489 Chr.1: 218345483-218345483 on Build GRCh38
rs753951462 Chr.1: 218345185-218345185 on Build GRCh38
rs956132795 Chr.1: 218345468-218345468 on Build GRCh38
rs143910852 Chr.1: 218345202-218345202 on Build GRCh38
rs756102671 Chr.1: 218345267-218345267 on Build GRCh38
rs575868653 Chr.1: 218345441-218345441 on Build GRCh38

View Full Product Details