Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228061166-228061671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNT3A
Entrez Gene ID
89780
Gene Name
Wnt family member 3A
Gene Aliases
-
UniGene
Hs.336930
Gene Chromosome Location
Chr.1: 228007022-228067113 on Build GRCh38
Transcripts
NM_033131.3 XM_011544319.2

SNP Detail

SNP ID Chromosome Location
rs999516763 Chr.1: 228061216-228061216 on Build GRCh38
rs74143637 Chr.1: 228061483-228061483 on Build GRCh38
rs949625479 Chr.1: 228061428-228061428 on Build GRCh38
rs115388753 Chr.1: 228061627-228061627 on Build GRCh38
rs1013639334 Chr.1: 228061191-228061191 on Build GRCh38
rs1008177599 Chr.1: 228061588-228061588 on Build GRCh38
rs900492555 Chr.1: 228061486-228061486 on Build GRCh38
rs1053345856 Chr.1: 228061603-228061603 on Build GRCh38
rs1046457334 Chr.1: 228061444-228061444 on Build GRCh38
rs1028277158 Chr.1: 228061233-228061233 on Build GRCh38
rs938953857 Chr.1: 228061397-228061397 on Build GRCh38
rs184957961 Chr.1: 228061577-228061577 on Build GRCh38
rs776862123 Chr.1: 228061214-228061214 on Build GRCh38
rs916716461 Chr.1: 228061423-228061423 on Build GRCh38
rs563216428 Chr.1: 228061305-228061305 on Build GRCh38
rs377073555 Chr.1: 228061593-228061593 on Build GRCh38
rs1011452876 Chr.1: 228061545-228061545 on Build GRCh38
rs1011736258 Chr.1: 228061636-228061636 on Build GRCh38
rs893806201 Chr.1: 228061513-228061513 on Build GRCh38
rs776111147 Chr.1: 228061457-228061457 on Build GRCh38
rs377410017 Chr.1: 228061292-228061292 on Build GRCh38
rs979748174 Chr.1: 228061347-228061347 on Build GRCh38
rs1008041807 Chr.1: 228061276-228061276 on Build GRCh38
rs895319718 Chr.1: 228061188-228061188 on Build GRCh38
rs942137811 Chr.1: 228061185-228061185 on Build GRCh38
rs529770420 Chr.1: 228061401-228061401 on Build GRCh38
rs1053691238 Chr.1: 228061500-228061500 on Build GRCh38
rs991489885 Chr.1: 228061410-228061410 on Build GRCh38
rs566710933 Chr.1: 228061475-228061475 on Build GRCh38
rs757016002 Chr.1: 228061578-228061578 on Build GRCh38
rs575184643 Chr.1: 228061521-228061521 on Build GRCh38
rs531344906 Chr.1: 228061203-228061203 on Build GRCh38
rs928853562 Chr.1: 228061371-228061371 on Build GRCh38
rs902635010 Chr.1: 228061479-228061479 on Build GRCh38
rs767215885 Chr.1: 228061509-228061510 on Build GRCh38
rs760672042 Chr.1: 228061328-228061328 on Build GRCh38
rs908893826 Chr.1: 228061367-228061367 on Build GRCh38
rs562612669 Chr.1: 228061358-228061358 on Build GRCh38
rs950348068 Chr.1: 228061334-228061334 on Build GRCh38
rs74143636 Chr.1: 228061285-228061285 on Build GRCh38
rs997378070 Chr.1: 228061649-228061649 on Build GRCh38
rs1017403318 Chr.1: 228061302-228061302 on Build GRCh38
rs1017977329 Chr.1: 228061565-228061565 on Build GRCh38
rs746835243 Chr.1: 228061311-228061311 on Build GRCh38
rs983137366 Chr.1: 228061344-228061344 on Build GRCh38
rs942089497 Chr.1: 228061474-228061474 on Build GRCh38
rs181720093 Chr.1: 228061325-228061325 on Build GRCh38
rs551590725 Chr.1: 228061407-228061407 on Build GRCh38
rs184591774 Chr.1: 228061272-228061272 on Build GRCh38

View Full Product Details