Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32462857-32463359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
UniGene
Hs.585464
Gene Chromosome Location
Chr.10: 32446082-32882864 on Build GRCh38
Transcripts
NM_001026383.2 NM_001321115.1 NM_145023.5 NR_109826.1 NR_109827.1 XM_006717505.1 XM_006717506.1 XM_011519663.1 XM_011519664.1 XM_011519666.1 XM_011519667.1 XM_011519668.1 XM_011519669.1 XM_011519670.1 XM_011519671.1 XM_011519672.1 XM_011519673.1 XM_011519674.1 XM_011519675.1 XM_011519676.1 XM_011519677.1 XM_011519678.1 XM_011519679.1 XM_011519680.2 XM_011519684.2 XM_011519686.2 XM_011519688.2 XM_017016638.1 XM_017016639.1 XM_017016640.1 XM_017016643.1 XM_017016644.1 XM_017016645.1 XM_017016647.1 XM_017016650.1 XM_017016651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
159807 c.499C>G p.Q167E Chr.10: 32463038-32463038 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs187275637 Chr.10: 32462912-32462912 on Build GRCh38
rs962073034 Chr.10: 32462958-32462958 on Build GRCh38
rs199697638 Chr.10: 32463060-32463060 on Build GRCh38
rs945080744 Chr.10: 32463109-32463109 on Build GRCh38
rs560656168 Chr.10: 32463042-32463042 on Build GRCh38
rs975790655 Chr.10: 32463271-32463271 on Build GRCh38
rs2505380 Chr.10: 32462973-32462973 on Build GRCh38
rs561867074 Chr.10: 32463120-32463120 on Build GRCh38
rs779925017 Chr.10: 32463034-32463034 on Build GRCh38
rs566441984 Chr.10: 32462910-32462910 on Build GRCh38
rs372793242 Chr.10: 32463061-32463061 on Build GRCh38
rs552825444 Chr.10: 32463256-32463256 on Build GRCh38
rs766393125 Chr.10: 32463003-32463010 on Build GRCh38
rs575114825 Chr.10: 32462956-32462956 on Build GRCh38
rs763385542 Chr.10: 32463055-32463055 on Build GRCh38
rs1054919412 Chr.10: 32463151-32463151 on Build GRCh38
rs748945459 Chr.10: 32462998-32462998 on Build GRCh38
rs761981084 Chr.10: 32463048-32463048 on Build GRCh38
rs958803797 Chr.10: 32462928-32462928 on Build GRCh38
rs78864560 Chr.10: 32463037-32463037 on Build GRCh38
rs910771839 Chr.10: 32463285-32463285 on Build GRCh38
rs777473184 Chr.10: 32462984-32462984 on Build GRCh38
rs1022116580 Chr.10: 32463194-32463194 on Build GRCh38
rs758929012 Chr.10: 32462967-32462967 on Build GRCh38
rs184037309 Chr.10: 32463299-32463299 on Build GRCh38
rs77855068 Chr.10: 32463033-32463033 on Build GRCh38
rs907163449 Chr.10: 32463067-32463067 on Build GRCh38
rs193065073 Chr.10: 32463000-32463000 on Build GRCh38
rs765348327 Chr.10: 32462960-32462960 on Build GRCh38
rs966067741 Chr.10: 32463264-32463264 on Build GRCh38
rs866733906 Chr.10: 32463214-32463214 on Build GRCh38
rs539408682 Chr.10: 32462983-32462983 on Build GRCh38
rs2460701 Chr.10: 32463257-32463257 on Build GRCh38
rs1035320997 Chr.10: 32463104-32463104 on Build GRCh38
rs986322839 Chr.10: 32463025-32463025 on Build GRCh38
rs141119129 Chr.10: 32462889-32462889 on Build GRCh38
rs540353936 Chr.10: 32463080-32463080 on Build GRCh38
rs755369244 Chr.10: 32462982-32462982 on Build GRCh38
rs146678326 Chr.10: 32463135-32463135 on Build GRCh38
rs146477841 Chr.10: 32462888-32462888 on Build GRCh38
rs1033460330 Chr.10: 32462925-32462925 on Build GRCh38
rs769816144 Chr.10: 32463049-32463049 on Build GRCh38
rs1005758908 Chr.10: 32463183-32463183 on Build GRCh38
rs923648722 Chr.10: 32462894-32462894 on Build GRCh38
rs746814587 Chr.10: 32463035-32463035 on Build GRCh38
rs888289366 Chr.10: 32463155-32463155 on Build GRCh38
rs1005005948 Chr.10: 32463001-32463002 on Build GRCh38
rs200536902 Chr.10: 32463054-32463054 on Build GRCh38
rs750890635 Chr.10: 32462965-32462965 on Build GRCh38
rs553589872 Chr.10: 32462944-32462944 on Build GRCh38
rs1006299855 Chr.10: 32462950-32462950 on Build GRCh38
rs140092049 Chr.10: 32463184-32463184 on Build GRCh38
rs77999276 Chr.10: 32462903-32462903 on Build GRCh38
rs962412202 Chr.10: 32463211-32463211 on Build GRCh38
rs1002056762 Chr.10: 32463145-32463145 on Build GRCh38
rs776889762 Chr.10: 32463040-32463040 on Build GRCh38
rs745746480 Chr.10: 32463031-32463031 on Build GRCh38
rs551081702 Chr.10: 32463276-32463276 on Build GRCh38
rs566711025 Chr.10: 32463238-32463238 on Build GRCh38
rs913600747 Chr.10: 32463106-32463106 on Build GRCh38
rs201456400 Chr.10: 32463009-32463009 on Build GRCh38
rs759926713 Chr.10: 32463072-32463072 on Build GRCh38

View Full Product Details