Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70422011-70422518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4EBP2
Entrez Gene ID
1979
Gene Name
eukaryotic translation initiation factor 4E binding protein 2
Gene Aliases
4EBP2, PHASII
UniGene
Hs.621200
Gene Chromosome Location
Chr.10: 70404105-70428618 on Build GRCh38
Transcripts
NM_004096.4

SNP Detail

SNP ID Chromosome Location
rs916925364 Chr.10: 70422080-70422080 on Build GRCh38
rs971531816 Chr.10: 70422094-70422094 on Build GRCh38
rs538636167 Chr.10: 70422228-70422228 on Build GRCh38
rs1005751572 Chr.10: 70422049-70422049 on Build GRCh38
rs1024363684 Chr.10: 70422353-70422353 on Build GRCh38
rs369796060 Chr.10: 70422317-70422317 on Build GRCh38
rs957154267 Chr.10: 70422411-70422411 on Build GRCh38
rs188646299 Chr.10: 70422278-70422278 on Build GRCh38
rs150577060 Chr.10: 70422044-70422044 on Build GRCh38
rs1040069027 Chr.10: 70422229-70422229 on Build GRCh38
rs781509968 Chr.10: 70422367-70422367 on Build GRCh38
rs989909860 Chr.10: 70422418-70422418 on Build GRCh38
rs560502743 Chr.10: 70422226-70422226 on Build GRCh38
rs977212791 Chr.10: 70422111-70422111 on Build GRCh38
rs540369035 Chr.10: 70422456-70422456 on Build GRCh38
rs1031419390 Chr.10: 70422401-70422401 on Build GRCh38
rs1017179417 Chr.10: 70422054-70422054 on Build GRCh38
rs570904830 Chr.10: 70422142-70422142 on Build GRCh38
rs991120196 Chr.10: 70422079-70422079 on Build GRCh38
rs934039333 Chr.10: 70422254-70422254 on Build GRCh38
rs7093504 Chr.10: 70422046-70422046 on Build GRCh38
rs552619743 Chr.10: 70422102-70422102 on Build GRCh38
rs576904101 Chr.10: 70422320-70422320 on Build GRCh38
rs910464639 Chr.10: 70422213-70422213 on Build GRCh38
rs901190016 Chr.10: 70422254-70422254 on Build GRCh38
rs116482385 Chr.10: 70422203-70422203 on Build GRCh38
rs1043100 Chr.10: 70422091-70422091 on Build GRCh38
rs532703087 Chr.10: 70422230-70422230 on Build GRCh38
rs573462638 Chr.10: 70422442-70422442 on Build GRCh38
rs1013193319 Chr.10: 70422321-70422321 on Build GRCh38
rs527528089 Chr.10: 70422070-70422070 on Build GRCh38
rs943264414 Chr.10: 70422470-70422470 on Build GRCh38
rs45450199 Chr.10: 70422354-70422354 on Build GRCh38
rs894691251 Chr.10: 70422308-70422308 on Build GRCh38
rs763051514 Chr.10: 70422120-70422120 on Build GRCh38
rs534451528 Chr.10: 70422193-70422194 on Build GRCh38
rs139575224 Chr.10: 70422266-70422266 on Build GRCh38
rs117563909 Chr.10: 70422396-70422396 on Build GRCh38
rs546105882 Chr.10: 70422296-70422296 on Build GRCh38
rs4746035 Chr.10: 70422274-70422274 on Build GRCh38

View Full Product Details