Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73053059-73053573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P4HA1
Entrez Gene ID
5033
Gene Name
prolyl 4-hydroxylase subunit alpha 1
Gene Aliases
P4HA
UniGene
Hs.500047
Gene Chromosome Location
Chr.10: 73007217-73096974 on Build GRCh38
Transcripts
NM_000917.3 NM_001017962.2 NM_001142595.1 NM_001142596.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6020842 c.541T>C p.Y181H Chr.10: 73053513-73053513 on Build GRCh38
6020841 c.541T>C p.Y181H Chr.10: 73053513-73053513 on Build GRCh38
1260776 c.591C>T p.G197G Chr.10: 73053463-73053463 on Build GRCh38
5759704 c.539A>G p.Y180C Chr.10: 73053515-73053515 on Build GRCh38
5759705 c.539A>G p.Y180C Chr.10: 73053515-73053515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566474557 Chr.10: 73053449-73053449 on Build GRCh38
rs1024039064 Chr.10: 73053462-73053462 on Build GRCh38
rs555414636 Chr.10: 73053202-73053204 on Build GRCh38
rs201536150 Chr.10: 73053384-73053384 on Build GRCh38
rs759911959 Chr.10: 73053348-73053348 on Build GRCh38
rs781075726 Chr.10: 73053438-73053438 on Build GRCh38
rs750115106 Chr.10: 73053523-73053523 on Build GRCh38
rs891227904 Chr.10: 73053266-73053266 on Build GRCh38
rs763014418 Chr.10: 73053309-73053309 on Build GRCh38
rs754351127 Chr.10: 73053516-73053516 on Build GRCh38
rs775773147 Chr.10: 73053495-73053495 on Build GRCh38
rs570926997 Chr.10: 73053085-73053085 on Build GRCh38
rs35382319 Chr.10: 73053430-73053430 on Build GRCh38
rs202243570 Chr.10: 73053505-73053505 on Build GRCh38
rs757835999 Chr.10: 73053124-73053124 on Build GRCh38
rs532473620 Chr.10: 73053255-73053255 on Build GRCh38
rs951980788 Chr.10: 73053192-73053192 on Build GRCh38
rs759684737 Chr.10: 73053490-73053490 on Build GRCh38
rs764773456 Chr.10: 73053518-73053518 on Build GRCh38
rs1009116453 Chr.10: 73053316-73053316 on Build GRCh38
rs201200914 Chr.10: 73053463-73053463 on Build GRCh38
rs150873070 Chr.10: 73053262-73053262 on Build GRCh38
rs918191045 Chr.10: 73053527-73053527 on Build GRCh38
rs753250145 Chr.10: 73053354-73053354 on Build GRCh38
rs753442855 Chr.10: 73053381-73053381 on Build GRCh38
rs142587629 Chr.10: 73053500-73053500 on Build GRCh38
rs557049252 Chr.10: 73053215-73053215 on Build GRCh38
rs771181284 Chr.10: 73053338-73053338 on Build GRCh38
rs756388181 Chr.10: 73053235-73053235 on Build GRCh38
rs770948550 Chr.10: 73053469-73053469 on Build GRCh38
rs533814117 Chr.10: 73053543-73053543 on Build GRCh38
rs1000115429 Chr.10: 73053376-73053376 on Build GRCh38
rs200155008 Chr.10: 73053412-73053412 on Build GRCh38
rs905300232 Chr.10: 73053128-73053128 on Build GRCh38
rs1015276662 Chr.10: 73053303-73053303 on Build GRCh38
rs764561642 Chr.10: 73053506-73053506 on Build GRCh38
rs199883693 Chr.10: 73053342-73053342 on Build GRCh38
rs756810248 Chr.10: 73053382-73053382 on Build GRCh38
rs772496105 Chr.10: 73053491-73053491 on Build GRCh38
rs979158950 Chr.10: 73053440-73053440 on Build GRCh38
rs201215553 Chr.10: 73053352-73053352 on Build GRCh38
rs778735708 Chr.10: 73053390-73053390 on Build GRCh38
rs774533074 Chr.10: 73053347-73053347 on Build GRCh38
rs538318651 Chr.10: 73053155-73053155 on Build GRCh38
rs546678701 Chr.10: 73053095-73053095 on Build GRCh38
rs941631976 Chr.10: 73053206-73053206 on Build GRCh38
rs1039063029 Chr.10: 73053216-73053216 on Build GRCh38
rs748359213 Chr.10: 73053287-73053288 on Build GRCh38
rs200447386 Chr.10: 73053432-73053432 on Build GRCh38
rs568319415 Chr.10: 73053373-73053373 on Build GRCh38
rs569368332 Chr.10: 73053293-73053293 on Build GRCh38
rs200760249 Chr.10: 73053413-73053413 on Build GRCh38
rs774583834 Chr.10: 73053483-73053483 on Build GRCh38
rs746404043 Chr.10: 73053178-73053178 on Build GRCh38
rs761026912 Chr.10: 73053372-73053372 on Build GRCh38
rs200285096 Chr.10: 73053419-73053419 on Build GRCh38
rs767730928 Chr.10: 73053351-73053351 on Build GRCh38

View Full Product Details