Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 74119843-74120366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3M1
Entrez Gene ID
26985
Gene Name
adaptor related protein complex 3 mu 1 subunit
Gene Aliases
-
UniGene
Hs.500104
Gene Chromosome Location
Chr.10: 74120257-74151085 on Build GRCh38
Transcripts
NM_001320263.1 NM_001320264.1 NM_001320265.1 NM_012095.5 NM_207012.3 NR_135191.1 NM_003373.3 NM_014000.2

Gene Symbol
VCL
Entrez Gene ID
7414
Gene Name
vinculin
Gene Aliases
CMD1W, CMH15, HEL114, MV, MVCL
UniGene
Hs.643896
Gene Chromosome Location
Chr.10: 73998114-74120160 on Build GRCh38
Transcripts
NM_001320263.1 NM_001320264.1 NM_001320265.1 NM_012095.5 NM_207012.3 NR_135191.1 NM_003373.3 NM_014000.2

SNP Detail

SNP ID Chromosome Location
rs370456912 Chr.10: 74120011-74120012 on Build GRCh38
rs937984852 Chr.10: 74119888-74119888 on Build GRCh38
rs540882056 Chr.10: 74120090-74120090 on Build GRCh38
rs984674608 Chr.10: 74119877-74119877 on Build GRCh38
rs957266115 Chr.10: 74120064-74120064 on Build GRCh38
rs892639264 Chr.10: 74119981-74119981 on Build GRCh38
rs577038840 Chr.10: 74119963-74119963 on Build GRCh38
rs372750308 Chr.10: 74119920-74119920 on Build GRCh38
rs1001907440 Chr.10: 74120021-74120021 on Build GRCh38
rs941614996 Chr.10: 74120264-74120265 on Build GRCh38
rs559715454 Chr.10: 74120117-74120117 on Build GRCh38
rs151279619 Chr.10: 74119981-74119982 on Build GRCh38
rs886047230 Chr.10: 74119930-74119930 on Build GRCh38
rs3829205 Chr.10: 74120018-74120018 on Build GRCh38
rs928016175 Chr.10: 74120183-74120184 on Build GRCh38
rs984669667 Chr.10: 74120115-74120115 on Build GRCh38
rs554923257 Chr.10: 74120022-74120022 on Build GRCh38
rs922611475 Chr.10: 74120263-74120263 on Build GRCh38
rs560246125 Chr.10: 74120247-74120247 on Build GRCh38
rs897416572 Chr.10: 74120318-74120319 on Build GRCh38
rs1030245107 Chr.10: 74120034-74120034 on Build GRCh38
rs922738089 Chr.10: 74119961-74119961 on Build GRCh38
rs377566645 Chr.10: 74119879-74119880 on Build GRCh38
rs201189785 Chr.10: 74120023-74120024 on Build GRCh38
rs778868100 Chr.10: 74120187-74120187 on Build GRCh38
rs140046305 Chr.10: 74119872-74119873 on Build GRCh38
rs909922125 Chr.10: 74120253-74120253 on Build GRCh38
rs541995253 Chr.10: 74120118-74120118 on Build GRCh38
rs962005859 Chr.10: 74120019-74120019 on Build GRCh38
rs35726593 Chr.10: 74119985-74119986 on Build GRCh38
rs1801382 Chr.10: 74119900-74119900 on Build GRCh38
rs200756813 Chr.10: 74120018-74120019 on Build GRCh38
rs8255 Chr.10: 74119992-74119992 on Build GRCh38
rs190121653 Chr.10: 74120203-74120203 on Build GRCh38
rs971665708 Chr.10: 74120155-74120155 on Build GRCh38
rs886047229 Chr.10: 74119877-74119877 on Build GRCh38
rs575785254 Chr.10: 74119962-74119962 on Build GRCh38
rs529702817 Chr.10: 74120060-74120060 on Build GRCh38
rs975409835 Chr.10: 74120224-74120224 on Build GRCh38
rs1049614 Chr.10: 74120057-74120057 on Build GRCh38
rs544380855 Chr.10: 74120305-74120305 on Build GRCh38
rs3037361 Chr.10: 74120031-74120032 on Build GRCh38
rs867628561 Chr.10: 74120036-74120036 on Build GRCh38
rs780201805 Chr.10: 74119886-74119886 on Build GRCh38
rs1032684844 Chr.10: 74120059-74120059 on Build GRCh38
rs886047228 Chr.10: 74119881-74119882 on Build GRCh38
rs1055067108 Chr.10: 74119907-74119907 on Build GRCh38
rs951915285 Chr.10: 74120086-74120086 on Build GRCh38
rs886047231 Chr.10: 74120010-74120011 on Build GRCh38
rs1020096525 Chr.10: 74120154-74120155 on Build GRCh38
rs1044043729 Chr.10: 74120011-74120011 on Build GRCh38
rs990749663 Chr.10: 74120229-74120229 on Build GRCh38
rs528707955 Chr.10: 74120267-74120267 on Build GRCh38
rs3829204 Chr.10: 74120023-74120023 on Build GRCh38
rs59061551 Chr.10: 74120031-74120031 on Build GRCh38
rs749090680 Chr.10: 74119905-74119905 on Build GRCh38
rs756405514 Chr.10: 74119876-74119876 on Build GRCh38
rs1052527163 Chr.10: 74120269-74120269 on Build GRCh38
rs914062903 Chr.10: 74120306-74120306 on Build GRCh38
rs946893917 Chr.10: 74120334-74120334 on Build GRCh38
rs981312354 Chr.10: 74120169-74120169 on Build GRCh38
rs1040173424 Chr.10: 74119930-74119930 on Build GRCh38

View Full Product Details