Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86935733-86936274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMRN2
Entrez Gene ID
79812
Gene Name
multimerin 2
Gene Aliases
EMILIN-3, EMILIN3, ENDOGLYX-1
UniGene
Hs.524479
Gene Chromosome Location
Chr.10: 86935540-86971311 on Build GRCh38
Transcripts
NM_024756.2 XM_005270153.1 XM_006717970.3

SNP Detail

SNP ID Chromosome Location
rs769811911 Chr.10: 86936147-86936147 on Build GRCh38
rs763165729 Chr.10: 86936212-86936212 on Build GRCh38
rs764460303 Chr.10: 86936087-86936089 on Build GRCh38
rs896442274 Chr.10: 86935880-86935880 on Build GRCh38
rs766521783 Chr.10: 86936249-86936249 on Build GRCh38
rs1038467445 Chr.10: 86935810-86935810 on Build GRCh38
rs917754320 Chr.10: 86936194-86936197 on Build GRCh38
rs3206734 Chr.10: 86935850-86935850 on Build GRCh38
rs920052487 Chr.10: 86936138-86936138 on Build GRCh38
rs998542119 Chr.10: 86936078-86936078 on Build GRCh38
rs547975208 Chr.10: 86935797-86935797 on Build GRCh38
rs983903646 Chr.10: 86935777-86935777 on Build GRCh38
rs530424245 Chr.10: 86936073-86936073 on Build GRCh38
rs1054473626 Chr.10: 86935962-86935962 on Build GRCh38
rs889164938 Chr.10: 86936001-86936001 on Build GRCh38
rs17408512 Chr.10: 86935949-86935949 on Build GRCh38
rs547960425 Chr.10: 86936043-86936043 on Build GRCh38
rs1044977156 Chr.10: 86936240-86936240 on Build GRCh38
rs761859477 Chr.10: 86936045-86936045 on Build GRCh38
rs1010437692 Chr.10: 86936010-86936010 on Build GRCh38
rs769200504 Chr.10: 86936018-86936018 on Build GRCh38
rs936990507 Chr.10: 86935823-86935823 on Build GRCh38
rs950324360 Chr.10: 86935871-86935871 on Build GRCh38
rs1000997324 Chr.10: 86935953-86935953 on Build GRCh38
rs551761822 Chr.10: 86936098-86936098 on Build GRCh38
rs74997421 Chr.10: 86935767-86935767 on Build GRCh38
rs951328146 Chr.10: 86936127-86936127 on Build GRCh38
rs767967190 Chr.10: 86935897-86935897 on Build GRCh38
rs919732242 Chr.10: 86935818-86935818 on Build GRCh38
rs1044675049 Chr.10: 86935908-86935908 on Build GRCh38
rs974546004 Chr.10: 86936122-86936122 on Build GRCh38
rs777162038 Chr.10: 86936035-86936035 on Build GRCh38
rs530342552 Chr.10: 86935917-86935917 on Build GRCh38
rs773307212 Chr.10: 86936184-86936184 on Build GRCh38
rs929792792 Chr.10: 86936171-86936171 on Build GRCh38
rs982620112 Chr.10: 86936183-86936187 on Build GRCh38
rs371337957 Chr.10: 86935777-86935777 on Build GRCh38
rs577612993 Chr.10: 86936211-86936211 on Build GRCh38
rs991117105 Chr.10: 86935828-86935828 on Build GRCh38
rs755504393 Chr.10: 86936093-86936094 on Build GRCh38
rs3206733 Chr.10: 86935872-86935872 on Build GRCh38
rs1013205397 Chr.10: 86935893-86935893 on Build GRCh38
rs375092242 Chr.10: 86936059-86936059 on Build GRCh38
rs568141852 Chr.10: 86936224-86936224 on Build GRCh38
rs534621439 Chr.10: 86936172-86936172 on Build GRCh38
rs911228680 Chr.10: 86935759-86935759 on Build GRCh38
rs766914886 Chr.10: 86935813-86935816 on Build GRCh38
rs957756957 Chr.10: 86935994-86935994 on Build GRCh38
rs1041953092 Chr.10: 86935887-86935887 on Build GRCh38
rs762552995 Chr.10: 86936011-86936011 on Build GRCh38
rs559717785 Chr.10: 86935846-86935846 on Build GRCh38
rs138136460 Chr.10: 86936109-86936109 on Build GRCh38
rs1032114089 Chr.10: 86935980-86935980 on Build GRCh38
rs750821170 Chr.10: 86936044-86936044 on Build GRCh38

View Full Product Details