Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92606135-92606673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF11
Entrez Gene ID
3832
Gene Name
kinesin family member 11
Gene Aliases
EG5, HKSP, KNSL1, MCLMR, TRIP5
UniGene
Hs.8878
Gene Chromosome Location
Chr.10: 92593068-92655395 on Build GRCh38
Transcripts
NM_004523.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7218022 c.246C>T p.Y82Y Chr.10: 92606654-92606654 on Build GRCh38
4695870 c.238G>T p.D80Y Chr.10: 92606646-92606646 on Build GRCh38
7207411 c.193A>G p.T65A Chr.10: 92606380-92606380 on Build GRCh38
5904928 c.211-6C>T p.? Chr.10: 92606613-92606613 on Build GRCh38
5492679 c.108C>T p.S36S Chr.10: 92606295-92606295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990746054 Chr.10: 92606261-92606261 on Build GRCh38
rs1054875072 Chr.10: 92606471-92606471 on Build GRCh38
rs781603790 Chr.10: 92606585-92606585 on Build GRCh38
rs182023792 Chr.10: 92606624-92606624 on Build GRCh38
rs1029636418 Chr.10: 92606172-92606172 on Build GRCh38
rs11393423 Chr.10: 92606591-92606592 on Build GRCh38
rs776498258 Chr.10: 92606244-92606244 on Build GRCh38
rs543526925 Chr.10: 92606429-92606429 on Build GRCh38
rs777932420 Chr.10: 92606244-92606245 on Build GRCh38
rs754165708 Chr.10: 92606296-92606296 on Build GRCh38
rs770932642 Chr.10: 92606590-92606591 on Build GRCh38
rs775105159 Chr.10: 92606405-92606405 on Build GRCh38
rs780120726 Chr.10: 92606327-92606327 on Build GRCh38
rs763302428 Chr.10: 92606529-92606529 on Build GRCh38
rs765172185 Chr.10: 92606218-92606218 on Build GRCh38
rs200410468 Chr.10: 92606380-92606380 on Build GRCh38
rs552534716 Chr.10: 92606551-92606551 on Build GRCh38
rs762408459 Chr.10: 92606254-92606254 on Build GRCh38
rs759170504 Chr.10: 92606292-92606292 on Build GRCh38
rs770861566 Chr.10: 92606614-92606614 on Build GRCh38
rs1051011411 Chr.10: 92606606-92606606 on Build GRCh38
rs372961115 Chr.10: 92606410-92606410 on Build GRCh38
rs770698062 Chr.10: 92606590-92606590 on Build GRCh38
rs796052144 Chr.10: 92606392-92606392 on Build GRCh38
rs781156148 Chr.10: 92606340-92606340 on Build GRCh38
rs139504305 Chr.10: 92606512-92606512 on Build GRCh38
rs751609310 Chr.10: 92606569-92606569 on Build GRCh38
rs773582910 Chr.10: 92606647-92606647 on Build GRCh38
rs368028003 Chr.10: 92606441-92606441 on Build GRCh38
rs765569740 Chr.10: 92606411-92606411 on Build GRCh38
rs760952528 Chr.10: 92606613-92606614 on Build GRCh38
rs891118071 Chr.10: 92606636-92606636 on Build GRCh38
rs188684233 Chr.10: 92606604-92606604 on Build GRCh38
rs369196517 Chr.10: 92606628-92606628 on Build GRCh38
rs752961144 Chr.10: 92606588-92606588 on Build GRCh38
rs769804344 Chr.10: 92606247-92606247 on Build GRCh38
rs371010146 Chr.10: 92606613-92606613 on Build GRCh38
rs376032374 Chr.10: 92606335-92606335 on Build GRCh38
rs756032812 Chr.10: 92606603-92606603 on Build GRCh38
rs762515248 Chr.10: 92606591-92606594 on Build GRCh38
rs916356707 Chr.10: 92606491-92606491 on Build GRCh38
rs765785937 Chr.10: 92606301-92606301 on Build GRCh38
rs759142342 Chr.10: 92606382-92606382 on Build GRCh38
rs184049283 Chr.10: 92606367-92606367 on Build GRCh38
rs777923201 Chr.10: 92606361-92606361 on Build GRCh38
rs753104089 Chr.10: 92606217-92606217 on Build GRCh38
rs561133245 Chr.10: 92606305-92606305 on Build GRCh38
rs766835030 Chr.10: 92606617-92606619 on Build GRCh38
rs748004032 Chr.10: 92606350-92606350 on Build GRCh38
rs775193169 Chr.10: 92606621-92606621 on Build GRCh38
rs746801517 Chr.10: 92606414-92606417 on Build GRCh38
rs771667854 Chr.10: 92606395-92606395 on Build GRCh38
rs774001058 Chr.10: 92606606-92606606 on Build GRCh38
rs770145180 Chr.10: 92606634-92606634 on Build GRCh38
rs75352237 Chr.10: 92606603-92606604 on Build GRCh38
rs80250278 Chr.10: 92606456-92606456 on Build GRCh38
rs750904039 Chr.10: 92606434-92606434 on Build GRCh38
rs796854274 Chr.10: 92606591-92606592 on Build GRCh38
rs773934211 Chr.10: 92606284-92606284 on Build GRCh38
rs755085897 Chr.10: 92606573-92606573 on Build GRCh38
rs199698837 Chr.10: 92606295-92606295 on Build GRCh38
rs773260058 Chr.10: 92606251-92606251 on Build GRCh38
rs375962985 Chr.10: 92606413-92606413 on Build GRCh38
rs979734876 Chr.10: 92606179-92606179 on Build GRCh38
rs1006936734 Chr.10: 92606203-92606203 on Build GRCh38
rs763388552 Chr.10: 92606649-92606649 on Build GRCh38
rs769680276 Chr.10: 92606351-92606351 on Build GRCh38
rs758886398 Chr.10: 92606222-92606222 on Build GRCh38
rs200980564 Chr.10: 92606246-92606246 on Build GRCh38
rs565058444 Chr.10: 92606591-92606591 on Build GRCh38
rs375931983 Chr.10: 92606571-92606571 on Build GRCh38
rs766396764 Chr.10: 92606449-92606449 on Build GRCh38
rs763865871 Chr.10: 92606604-92606605 on Build GRCh38
rs765910083 Chr.10: 92606262-92606262 on Build GRCh38
rs371359626 Chr.10: 92606173-92606173 on Build GRCh38
rs771543083 Chr.10: 92606617-92606617 on Build GRCh38
rs373135960 Chr.10: 92606310-92606310 on Build GRCh38
rs770362112 Chr.10: 92606373-92606373 on Build GRCh38
rs941186252 Chr.10: 92606493-92606493 on Build GRCh38

View Full Product Details