Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93625219-93625725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE6C
Entrez Gene ID
5146
Gene Name
phosphodiesterase 6C
Gene Aliases
ACHM5, COD4, PDEA2
UniGene
Hs.658121
Gene Chromosome Location
Chr.10: 93612588-93665673 on Build GRCh38
Transcripts
NM_006204.3 XM_017016322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5892095 c.908C>T p.P303L Chr.10: 93625618-93625618 on Build GRCh38
3967229 c.898G>T p.E300* Chr.10: 93625608-93625608 on Build GRCh38
5536866 c.939G>A p.R313R Chr.10: 93625649-93625649 on Build GRCh38
3441702 c.865G>A p.E289K Chr.10: 93625575-93625575 on Build GRCh38
3441703 c.897A>T p.G299G Chr.10: 93625607-93625607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756385811 Chr.10: 93625647-93625647 on Build GRCh38
rs760390582 Chr.10: 93625536-93625536 on Build GRCh38
rs767644884 Chr.10: 93625626-93625626 on Build GRCh38
rs374982904 Chr.10: 93625279-93625279 on Build GRCh38
rs555721116 Chr.10: 93625332-93625332 on Build GRCh38
rs535535161 Chr.10: 93625299-93625299 on Build GRCh38
rs371814026 Chr.10: 93625653-93625653 on Build GRCh38
rs757019766 Chr.10: 93625657-93625657 on Build GRCh38
rs1034040203 Chr.10: 93625459-93625459 on Build GRCh38
rs759590474 Chr.10: 93625623-93625623 on Build GRCh38
rs901828987 Chr.10: 93625402-93625402 on Build GRCh38
rs758212800 Chr.10: 93625673-93625673 on Build GRCh38
rs182347389 Chr.10: 93625465-93625465 on Build GRCh38
rs886047482 Chr.10: 93625658-93625658 on Build GRCh38
rs143162788 Chr.10: 93625615-93625615 on Build GRCh38
rs1026738619 Chr.10: 93625429-93625429 on Build GRCh38
rs772853811 Chr.10: 93625571-93625571 on Build GRCh38
rs971372451 Chr.10: 93625428-93625428 on Build GRCh38
rs763386959 Chr.10: 93625588-93625588 on Build GRCh38
rs145086429 Chr.10: 93625252-93625252 on Build GRCh38
rs75998292 Chr.10: 93625345-93625345 on Build GRCh38
rs374805348 Chr.10: 93625654-93625654 on Build GRCh38
rs1024555196 Chr.10: 93625386-93625386 on Build GRCh38
rs969892464 Chr.10: 93625505-93625505 on Build GRCh38
rs772036185 Chr.10: 93625503-93625503 on Build GRCh38
rs1035610092 Chr.10: 93625524-93625524 on Build GRCh38
rs138970846 Chr.10: 93625583-93625583 on Build GRCh38
rs776392319 Chr.10: 93625685-93625685 on Build GRCh38
rs371628034 Chr.10: 93625528-93625528 on Build GRCh38
rs747744070 Chr.10: 93625448-93625448 on Build GRCh38
rs1001387587 Chr.10: 93625519-93625519 on Build GRCh38
rs560168696 Chr.10: 93625670-93625670 on Build GRCh38
rs907591947 Chr.10: 93625513-93625513 on Build GRCh38
rs976317552 Chr.10: 93625538-93625538 on Build GRCh38
rs904038860 Chr.10: 93625313-93625313 on Build GRCh38
rs1006734569 Chr.10: 93625456-93625456 on Build GRCh38
rs984552653 Chr.10: 93625475-93625475 on Build GRCh38
rs752650713 Chr.10: 93625627-93625627 on Build GRCh38
rs980412665 Chr.10: 93625449-93625449 on Build GRCh38
rs923427888 Chr.10: 93625551-93625551 on Build GRCh38
rs368923723 Chr.10: 93625683-93625683 on Build GRCh38
rs201884878 Chr.10: 93625619-93625619 on Build GRCh38
rs564759104 Chr.10: 93625381-93625381 on Build GRCh38
rs760417465 Chr.10: 93625662-93625662 on Build GRCh38
rs771407674 Chr.10: 93625696-93625696 on Build GRCh38
rs563634712 Chr.10: 93625256-93625256 on Build GRCh38
rs779620671 Chr.10: 93625674-93625674 on Build GRCh38
rs575787738 Chr.10: 93625335-93625335 on Build GRCh38
rs398124218 Chr.10: 93625597-93625597 on Build GRCh38
rs752106542 Chr.10: 93625617-93625617 on Build GRCh38
rs746462386 Chr.10: 93625678-93625678 on Build GRCh38
rs775065186 Chr.10: 93625529-93625529 on Build GRCh38
rs1010031121 Chr.10: 93625349-93625349 on Build GRCh38
rs749735213 Chr.10: 93625689-93625689 on Build GRCh38

View Full Product Details