Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93901815-93902325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35G1
Entrez Gene ID
159371
Gene Name
solute carrier family 35 member G1
Gene Aliases
C10orf60, TMEM20
UniGene
Hs.632085
Gene Chromosome Location
Chr.10: 93893951-93914265 on Build GRCh38
Transcripts
NM_001134658.1 NM_153226.2 XM_011539371.2 XM_011539373.2

SNP Detail

SNP ID Chromosome Location
rs1047528465 Chr.10: 93901967-93901967 on Build GRCh38
rs1043418462 Chr.10: 93902026-93902026 on Build GRCh38
rs562004824 Chr.10: 93902275-93902275 on Build GRCh38
rs1000202563 Chr.10: 93902045-93902045 on Build GRCh38
rs902737078 Chr.10: 93902044-93902044 on Build GRCh38
rs946364690 Chr.10: 93902015-93902015 on Build GRCh38
rs944581556 Chr.10: 93901992-93901992 on Build GRCh38
rs1048500448 Chr.10: 93902012-93902012 on Build GRCh38
rs916698361 Chr.10: 93901900-93901900 on Build GRCh38
rs551765796 Chr.10: 93901840-93901840 on Build GRCh38
rs77596423 Chr.10: 93902209-93902209 on Build GRCh38
rs907483934 Chr.10: 93902085-93902085 on Build GRCh38
rs962770155 Chr.10: 93902256-93902256 on Build GRCh38
rs530826987 Chr.10: 93901897-93901897 on Build GRCh38
rs747792441 Chr.10: 93901993-93901993 on Build GRCh38
rs911733086 Chr.10: 93901978-93901978 on Build GRCh38
rs918305732 Chr.10: 93901990-93901990 on Build GRCh38
rs1021700850 Chr.10: 93902273-93902273 on Build GRCh38
rs544183966 Chr.10: 93901902-93901902 on Build GRCh38
rs542144196 Chr.10: 93901991-93901991 on Build GRCh38
rs991291031 Chr.10: 93901893-93901893 on Build GRCh38
rs1035300753 Chr.10: 93902088-93902088 on Build GRCh38
rs753570625 Chr.10: 93902193-93902193 on Build GRCh38
rs896633545 Chr.10: 93902156-93902156 on Build GRCh38
rs539263459 Chr.10: 93902003-93902003 on Build GRCh38
rs148475687 Chr.10: 93901889-93901889 on Build GRCh38
rs954026336 Chr.10: 93902279-93902279 on Build GRCh38
rs190416173 Chr.10: 93901857-93901857 on Build GRCh38
rs929234524 Chr.10: 93901962-93901962 on Build GRCh38
rs1044651413 Chr.10: 93902037-93902039 on Build GRCh38
rs1011672139 Chr.10: 93902242-93902243 on Build GRCh38
rs891836440 Chr.10: 93902223-93902223 on Build GRCh38
rs550629031 Chr.10: 93901907-93901907 on Build GRCh38
rs974010842 Chr.10: 93902277-93902277 on Build GRCh38
rs1039184287 Chr.10: 93902002-93902002 on Build GRCh38
rs192683321 Chr.10: 93901939-93901939 on Build GRCh38
rs1035609298 Chr.10: 93901860-93901860 on Build GRCh38
rs912500766 Chr.10: 93901908-93901908 on Build GRCh38
rs960796949 Chr.10: 93901890-93901890 on Build GRCh38
rs766181862 Chr.10: 93902070-93902070 on Build GRCh38

View Full Product Details